Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30063637-30064133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDOA
Entrez Gene ID
226
Gene Name
aldolase, fructose-bisphosphate A
Gene Aliases
ALDA, GSD12, HEL-S-87p
UniGene
Hs.513490
Gene Chromosome Location
Chr.16: 30053090-30070420 on Build GRCh38
Transcripts
NM_000034.3 XM_011545768.2

SNP Detail

SNP ID Chromosome Location
rs36029842 Chr.16: 30063861-30063861 on Build GRCh38
rs965697610 Chr.16: 30063846-30063879 on Build GRCh38
rs943786501 Chr.16: 30064012-30064012 on Build GRCh38
rs973124133 Chr.16: 30064100-30064100 on Build GRCh38
rs926344871 Chr.16: 30063781-30063781 on Build GRCh38
rs904595296 Chr.16: 30063904-30063904 on Build GRCh38
rs1051655934 Chr.16: 30063843-30063843 on Build GRCh38
rs529860854 Chr.16: 30063794-30063794 on Build GRCh38
rs970211975 Chr.16: 30063993-30063993 on Build GRCh38
rs1031951266 Chr.16: 30063728-30063728 on Build GRCh38
rs768241580 Chr.16: 30064070-30064070 on Build GRCh38
rs903226082 Chr.16: 30064074-30064074 on Build GRCh38
rs548243129 Chr.16: 30063858-30063858 on Build GRCh38
rs1011558608 Chr.16: 30063824-30063824 on Build GRCh38
rs562655563 Chr.16: 30063784-30063784 on Build GRCh38
rs144671145 Chr.16: 30063946-30063946 on Build GRCh38
rs575936238 Chr.16: 30064058-30064058 on Build GRCh38
rs893122548 Chr.16: 30063805-30063805 on Build GRCh38
rs908225644 Chr.16: 30064007-30064007 on Build GRCh38
rs952649928 Chr.16: 30063986-30063986 on Build GRCh38
rs1051146203 Chr.16: 30063795-30063795 on Build GRCh38
rs902047690 Chr.16: 30063767-30063767 on Build GRCh38
rs77821461 Chr.16: 30064034-30064034 on Build GRCh38
rs956470998 Chr.16: 30063736-30063736 on Build GRCh38
rs191041320 Chr.16: 30064104-30064104 on Build GRCh38
rs763167376 Chr.16: 30063713-30063713 on Build GRCh38
rs538492279 Chr.16: 30064028-30064028 on Build GRCh38
rs1035528956 Chr.16: 30063872-30063872 on Build GRCh38
rs1011841445 Chr.16: 30064050-30064050 on Build GRCh38
rs368816502 Chr.16: 30063778-30063778 on Build GRCh38
rs767655507 Chr.16: 30063936-30063936 on Build GRCh38
rs572970341 Chr.16: 30064081-30064081 on Build GRCh38
rs545165122 Chr.16: 30063683-30063683 on Build GRCh38
rs923449674 Chr.16: 30064024-30064024 on Build GRCh38
rs958823626 Chr.16: 30063922-30063922 on Build GRCh38
rs1041226287 Chr.16: 30064013-30064013 on Build GRCh38
rs904541204 Chr.16: 30063671-30063671 on Build GRCh38
rs1014923254 Chr.16: 30063979-30063979 on Build GRCh38
rs1024536701 Chr.16: 30063991-30063991 on Build GRCh38
rs552539779 Chr.16: 30063948-30063948 on Build GRCh38
rs941727785 Chr.16: 30063792-30063792 on Build GRCh38
rs963092934 Chr.16: 30064060-30064060 on Build GRCh38
rs908276583 Chr.16: 30063710-30063710 on Build GRCh38
rs1017660484 Chr.16: 30063668-30063668 on Build GRCh38
rs554432237 Chr.16: 30064079-30064079 on Build GRCh38
rs1046781309 Chr.16: 30063685-30063685 on Build GRCh38
rs1041699170 Chr.16: 30064062-30064062 on Build GRCh38
rs973458561 Chr.16: 30063797-30063797 on Build GRCh38
rs560213553 Chr.16: 30063708-30063708 on Build GRCh38
rs376372530 Chr.16: 30064105-30064105 on Build GRCh38
rs980225726 Chr.16: 30064019-30064019 on Build GRCh38
rs557673274 Chr.16: 30063853-30063853 on Build GRCh38
rs116317239 Chr.16: 30063964-30063964 on Build GRCh38
rs893363131 Chr.16: 30064035-30064035 on Build GRCh38
rs566611884 Chr.16: 30063934-30063934 on Build GRCh38
rs1040506504 Chr.16: 30063746-30063746 on Build GRCh38
rs943802912 Chr.16: 30063742-30063742 on Build GRCh38
rs536086646 Chr.16: 30063749-30063749 on Build GRCh38
rs982384586 Chr.16: 30063988-30063988 on Build GRCh38
rs575060506 Chr.16: 30064061-30064061 on Build GRCh38
rs762074553 Chr.16: 30063932-30063932 on Build GRCh38
rs764506062 Chr.16: 30063834-30063834 on Build GRCh38
rs970160969 Chr.16: 30063750-30063750 on Build GRCh38
rs536039533 Chr.16: 30064069-30064069 on Build GRCh38
rs776143747 Chr.16: 30063660-30063660 on Build GRCh38
rs1003375008 Chr.16: 30063860-30063860 on Build GRCh38

View Full Product Details