Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30378433-30378930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEPT1
Entrez Gene ID
1731
Gene Name
septin 1
Gene Aliases
DIFF6, LARP, PNUTL3, SEP1
UniGene
Hs.632176
Gene Chromosome Location
Chr.16: 30378133-30382850 on Build GRCh38
Transcripts
NM_052838.4 XM_017022991.1 NM_001214906.1 NM_001214907.1

Gene Symbol
ZNF48
Entrez Gene ID
197407
Gene Name
zinc finger protein 48
Gene Aliases
ZNF553
UniGene
Hs.513501
Gene Chromosome Location
Chr.16: 30378312-30400108 on Build GRCh38
Transcripts
NM_052838.4 XM_017022991.1 NM_001214906.1 NM_001214907.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5789427 c.1009G>A p.D337N Chr.16: 30378618-30378618 on Build GRCh38
703298 c.1047G>A p.K349K Chr.16: 30378491-30378491 on Build GRCh38
6522757 c.1036A>T p.M346L Chr.16: 30378502-30378502 on Build GRCh38
7077364 c.1014A>C p.E338D Chr.16: 30378613-30378613 on Build GRCh38
6144418 c.1003G>C p.E335Q Chr.16: 30378624-30378624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766275412 Chr.16: 30378463-30378463 on Build GRCh38
rs762996705 Chr.16: 30378615-30378615 on Build GRCh38
rs188765492 Chr.16: 30378628-30378628 on Build GRCh38
rs1053587490 Chr.16: 30378454-30378454 on Build GRCh38
rs780363243 Chr.16: 30378688-30378688 on Build GRCh38
rs765670566 Chr.16: 30378656-30378656 on Build GRCh38
rs775601659 Chr.16: 30378494-30378494 on Build GRCh38
rs750727995 Chr.16: 30378659-30378659 on Build GRCh38
rs769007736 Chr.16: 30378598-30378598 on Build GRCh38
rs945305815 Chr.16: 30378498-30378498 on Build GRCh38
rs370036422 Chr.16: 30378589-30378589 on Build GRCh38
rs746342786 Chr.16: 30378588-30378588 on Build GRCh38
rs749998572 Chr.16: 30378661-30378661 on Build GRCh38
rs1032914191 Chr.16: 30378568-30378568 on Build GRCh38
rs774095793 Chr.16: 30378745-30378745 on Build GRCh38
rs542340766 Chr.16: 30378812-30378812 on Build GRCh38
rs1008692747 Chr.16: 30378483-30378483 on Build GRCh38
rs560713592 Chr.16: 30378827-30378827 on Build GRCh38
rs770906252 Chr.16: 30378737-30378737 on Build GRCh38
rs372111888 Chr.16: 30378720-30378720 on Build GRCh38
rs370110193 Chr.16: 30378706-30378706 on Build GRCh38
rs1047308484 Chr.16: 30378770-30378770 on Build GRCh38
rs893632442 Chr.16: 30378472-30378472 on Build GRCh38
rs772853052 Chr.16: 30378612-30378612 on Build GRCh38
rs964876120 Chr.16: 30378503-30378503 on Build GRCh38
rs766212898 Chr.16: 30378624-30378624 on Build GRCh38
rs747058994 Chr.16: 30378595-30378595 on Build GRCh38
rs148773715 Chr.16: 30378847-30378848 on Build GRCh38
rs757647512 Chr.16: 30378651-30378651 on Build GRCh38
rs767151310 Chr.16: 30378567-30378567 on Build GRCh38
rs373429879 Chr.16: 30378550-30378550 on Build GRCh38
rs374758184 Chr.16: 30378600-30378600 on Build GRCh38
rs1038534122 Chr.16: 30378736-30378736 on Build GRCh38
rs957267638 Chr.16: 30378892-30378892 on Build GRCh38
rs200612718 Chr.16: 30378750-30378750 on Build GRCh38
rs534215977 Chr.16: 30378668-30378668 on Build GRCh38
rs1015817631 Chr.16: 30378790-30378790 on Build GRCh38
rs1032884539 Chr.16: 30378844-30378844 on Build GRCh38
rs115079389 Chr.16: 30378870-30378870 on Build GRCh38
rs980567843 Chr.16: 30378886-30378886 on Build GRCh38
rs765282344 Chr.16: 30378540-30378540 on Build GRCh38
rs753751956 Chr.16: 30378543-30378543 on Build GRCh38
rs377366007 Chr.16: 30378594-30378594 on Build GRCh38
rs754556921 Chr.16: 30378457-30378457 on Build GRCh38
rs1003183055 Chr.16: 30378580-30378580 on Build GRCh38
rs989491764 Chr.16: 30378838-30378838 on Build GRCh38
rs201478158 Chr.16: 30378703-30378703 on Build GRCh38
rs757557744 Chr.16: 30378581-30378581 on Build GRCh38
rs779596462 Chr.16: 30378455-30378455 on Build GRCh38
rs751185887 Chr.16: 30378456-30378456 on Build GRCh38
rs760993615 Chr.16: 30378500-30378500 on Build GRCh38
rs202168059 Chr.16: 30378555-30378555 on Build GRCh38
rs574977750 Chr.16: 30378759-30378759 on Build GRCh38
rs758977750 Chr.16: 30378639-30378639 on Build GRCh38
rs941081235 Chr.16: 30378885-30378885 on Build GRCh38
rs930269952 Chr.16: 30378763-30378763 on Build GRCh38
rs751012090 Chr.16: 30378554-30378554 on Build GRCh38
rs976803356 Chr.16: 30378814-30378814 on Build GRCh38
rs556354729 Chr.16: 30378751-30378751 on Build GRCh38
rs774947841 Chr.16: 30378752-30378752 on Build GRCh38
rs147923586 Chr.16: 30378689-30378689 on Build GRCh38
rs368548834 Chr.16: 30378468-30378468 on Build GRCh38
rs779307561 Chr.16: 30378582-30378582 on Build GRCh38
rs749031577 Chr.16: 30378724-30378724 on Build GRCh38
rs1008245192 Chr.16: 30378795-30378795 on Build GRCh38
rs966001302 Chr.16: 30378793-30378793 on Build GRCh38
rs906211907 Chr.16: 30378552-30378552 on Build GRCh38
rs773118369 Chr.16: 30378712-30378712 on Build GRCh38
rs747981524 Chr.16: 30378602-30378602 on Build GRCh38
rs762404144 Chr.16: 30378545-30378545 on Build GRCh38
rs550819100 Chr.16: 30378515-30378515 on Build GRCh38
rs771493820 Chr.16: 30378461-30378461 on Build GRCh38
rs114027820 Chr.16: 30378883-30378883 on Build GRCh38
rs750328205 Chr.16: 30378544-30378544 on Build GRCh38
rs755063266 Chr.16: 30378690-30378690 on Build GRCh38
rs920161834 Chr.16: 30378742-30378742 on Build GRCh38
rs116643497 Chr.16: 30378604-30378604 on Build GRCh38
rs13336099 Chr.16: 30378869-30378869 on Build GRCh38
rs981559339 Chr.16: 30378695-30378695 on Build GRCh38
rs200316804 Chr.16: 30378471-30378471 on Build GRCh38
rs767169336 Chr.16: 30378641-30378641 on Build GRCh38
rs776639677 Chr.16: 30378522-30378522 on Build GRCh38
rs749638833 Chr.16: 30378458-30378458 on Build GRCh38
rs761636402 Chr.16: 30378536-30378536 on Build GRCh38
rs921261287 Chr.16: 30378818-30378818 on Build GRCh38
rs765652384 Chr.16: 30378585-30378585 on Build GRCh38
rs759067671 Chr.16: 30378747-30378747 on Build GRCh38

View Full Product Details