Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30570314-30570854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF688
Entrez Gene ID
146542
Gene Name
zinc finger protein 688
Gene Aliases
-
UniGene
Hs.301463
Gene Chromosome Location
Chr.16: 30569677-30572407 on Build GRCh38
Transcripts
NM_001024683.1 NM_145271.3 XM_005255139.2 XM_005255140.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6049386 c.383T>G p.V128G Chr.16: 30570364-30570364 on Build GRCh38
3817795 c.323A>G p.N108S Chr.16: 30570424-30570424 on Build GRCh38
6049388 c.382G>A p.V128M Chr.16: 30570365-30570365 on Build GRCh38
4060153 c.403C>T p.L135L Chr.16: 30570344-30570344 on Build GRCh38
1377421 c.345G>T p.K115N Chr.16: 30570360-30570360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867058164 Chr.16: 30570501-30570501 on Build GRCh38
rs112208830 Chr.16: 30570438-30570438 on Build GRCh38
rs996804367 Chr.16: 30570814-30570814 on Build GRCh38
rs199950882 Chr.16: 30570761-30570761 on Build GRCh38
rs867543702 Chr.16: 30570777-30570777 on Build GRCh38
rs768034951 Chr.16: 30570388-30570388 on Build GRCh38
rs1026901787 Chr.16: 30570503-30570503 on Build GRCh38
rs114187215 Chr.16: 30570536-30570536 on Build GRCh38
rs996920346 Chr.16: 30570592-30570592 on Build GRCh38
rs762690708 Chr.16: 30570373-30570373 on Build GRCh38
rs995678318 Chr.16: 30570500-30570500 on Build GRCh38
rs3072029 Chr.16: 30570722-30570723 on Build GRCh38
rs958074912 Chr.16: 30570648-30570648 on Build GRCh38
rs958751974 Chr.16: 30570463-30570463 on Build GRCh38
rs532681247 Chr.16: 30570703-30570703 on Build GRCh38
rs977742176 Chr.16: 30570698-30570698 on Build GRCh38
rs772248056 Chr.16: 30570406-30570406 on Build GRCh38
rs750113192 Chr.16: 30570357-30570357 on Build GRCh38
rs754234250 Chr.16: 30570398-30570398 on Build GRCh38
rs571785590 Chr.16: 30570795-30570795 on Build GRCh38
rs779869480 Chr.16: 30570417-30570417 on Build GRCh38
rs943976165 Chr.16: 30570677-30570677 on Build GRCh38
rs887256673 Chr.16: 30570751-30570752 on Build GRCh38
rs565602482 Chr.16: 30570701-30570701 on Build GRCh38
rs763750385 Chr.16: 30570396-30570396 on Build GRCh38
rs538861014 Chr.16: 30570418-30570418 on Build GRCh38
rs779204537 Chr.16: 30570335-30570335 on Build GRCh38
rs1055430992 Chr.16: 30570723-30570742 on Build GRCh38
rs33997546 Chr.16: 30570355-30570355 on Build GRCh38
rs200429164 Chr.16: 30570762-30570769 on Build GRCh38
rs921110465 Chr.16: 30570729-30570729 on Build GRCh38
rs931022710 Chr.16: 30570738-30570738 on Build GRCh38
rs776528968 Chr.16: 30570456-30570461 on Build GRCh38
rs775256598 Chr.16: 30570341-30570341 on Build GRCh38
rs776633326 Chr.16: 30570441-30570441 on Build GRCh38
rs768612233 Chr.16: 30570344-30570344 on Build GRCh38
rs938337905 Chr.16: 30570723-30570734 on Build GRCh38
rs778849434 Chr.16: 30570402-30570402 on Build GRCh38
rs776381852 Chr.16: 30570348-30570348 on Build GRCh38
rs1028370890 Chr.16: 30570625-30570628 on Build GRCh38
rs79717302 Chr.16: 30570546-30570546 on Build GRCh38
rs560953845 Chr.16: 30570560-30570560 on Build GRCh38
rs914878155 Chr.16: 30570785-30570785 on Build GRCh38
rs368601331 Chr.16: 30570707-30570707 on Build GRCh38
rs553344886 Chr.16: 30570389-30570389 on Build GRCh38
rs549842765 Chr.16: 30570611-30570611 on Build GRCh38
rs768295705 Chr.16: 30570443-30570448 on Build GRCh38
rs540232737 Chr.16: 30570664-30570664 on Build GRCh38
rs181336462 Chr.16: 30570756-30570756 on Build GRCh38
rs572938950 Chr.16: 30570629-30570629 on Build GRCh38
rs771908916 Chr.16: 30570337-30570337 on Build GRCh38
rs768293639 Chr.16: 30570434-30570434 on Build GRCh38
rs780523922 Chr.16: 30570681-30570681 on Build GRCh38
rs376932353 Chr.16: 30570434-30570436 on Build GRCh38
rs908452803 Chr.16: 30570669-30570669 on Build GRCh38
rs1004858432 Chr.16: 30570519-30570519 on Build GRCh38
rs1028254701 Chr.16: 30570834-30570834 on Build GRCh38
rs374633072 Chr.16: 30570397-30570397 on Build GRCh38
rs754417217 Chr.16: 30570725-30570725 on Build GRCh38
rs769403192 Chr.16: 30570447-30570447 on Build GRCh38
rs762743839 Chr.16: 30570479-30570479 on Build GRCh38
rs991549875 Chr.16: 30570502-30570502 on Build GRCh38
rs894197178 Chr.16: 30570474-30570474 on Build GRCh38
rs567224880 Chr.16: 30570336-30570336 on Build GRCh38
rs34533623 Chr.16: 30570765-30570766 on Build GRCh38
rs901390887 Chr.16: 30570770-30570770 on Build GRCh38
rs757374797 Chr.16: 30570401-30570401 on Build GRCh38
rs967347883 Chr.16: 30570685-30570685 on Build GRCh38
rs551171864 Chr.16: 30570749-30570749 on Build GRCh38
rs549060120 Chr.16: 30570826-30570826 on Build GRCh38
rs760643468 Chr.16: 30570343-30570343 on Build GRCh38
rs761597026 Chr.16: 30570446-30570446 on Build GRCh38
rs1048585936 Chr.16: 30570751-30570751 on Build GRCh38
rs749459574 Chr.16: 30570456-30570456 on Build GRCh38
rs772818349 Chr.16: 30570459-30570459 on Build GRCh38
rs756829800 Chr.16: 30570624-30570624 on Build GRCh38
rs147838183 Chr.16: 30570405-30570405 on Build GRCh38
rs932367072 Chr.16: 30570739-30570739 on Build GRCh38
rs1020544425 Chr.16: 30570394-30570394 on Build GRCh38
rs1041686443 Chr.16: 30570766-30570767 on Build GRCh38
rs542663913 Chr.16: 30570550-30570550 on Build GRCh38
rs965756559 Chr.16: 30570574-30570574 on Build GRCh38

View Full Product Details