Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31039437-31039935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STX4
Entrez Gene ID
6810
Gene Name
syntaxin 4
Gene Aliases
STX4A, p35-2
UniGene
Hs.83734
Gene Chromosome Location
Chr.16: 31033095-31040168 on Build GRCh38
Transcripts
NM_001272095.1 NM_001272096.1 NM_004604.4 XM_005255521.1 XM_011545925.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6792635 c.703-1delG p.? Chr.16: 31039540-31039540 on Build GRCh38
3113682 c.746C>T p.A249V Chr.16: 31039584-31039584 on Build GRCh38
4060306 c.775G>A p.V259I Chr.16: 31039613-31039613 on Build GRCh38
6848916 c.706G>A p.E236K Chr.16: 31039544-31039544 on Build GRCh38
4060307 c.886G>T p.V296F Chr.16: 31039795-31039795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528546531 Chr.16: 31039885-31039885 on Build GRCh38
rs200012136 Chr.16: 31039549-31039549 on Build GRCh38
rs371550711 Chr.16: 31039496-31039496 on Build GRCh38
rs369325032 Chr.16: 31039699-31039699 on Build GRCh38
rs201229504 Chr.16: 31039807-31039807 on Build GRCh38
rs376701568 Chr.16: 31039557-31039557 on Build GRCh38
rs138235448 Chr.16: 31039745-31039745 on Build GRCh38
rs570754713 Chr.16: 31039815-31039815 on Build GRCh38
rs372521363 Chr.16: 31039841-31039841 on Build GRCh38
rs771745614 Chr.16: 31039759-31039759 on Build GRCh38
rs1037967843 Chr.16: 31039464-31039464 on Build GRCh38
rs951141766 Chr.16: 31039875-31039875 on Build GRCh38
rs757567713 Chr.16: 31039837-31039837 on Build GRCh38
rs768693850 Chr.16: 31039776-31039776 on Build GRCh38
rs756380170 Chr.16: 31039630-31039630 on Build GRCh38
rs746913731 Chr.16: 31039763-31039763 on Build GRCh38
rs771201826 Chr.16: 31039658-31039658 on Build GRCh38
rs776543576 Chr.16: 31039780-31039780 on Build GRCh38
rs142253791 Chr.16: 31039620-31039620 on Build GRCh38
rs781515100 Chr.16: 31039621-31039621 on Build GRCh38
rs1037975946 Chr.16: 31039725-31039725 on Build GRCh38
rs996977992 Chr.16: 31039499-31039499 on Build GRCh38
rs375926870 Chr.16: 31039688-31039688 on Build GRCh38
rs900518933 Chr.16: 31039525-31039525 on Build GRCh38
rs186297007 Chr.16: 31039660-31039660 on Build GRCh38
rs756773896 Chr.16: 31039506-31039506 on Build GRCh38
rs771950436 Chr.16: 31039585-31039585 on Build GRCh38
rs149552887 Chr.16: 31039762-31039762 on Build GRCh38
rs191587609 Chr.16: 31039785-31039785 on Build GRCh38
rs748753141 Chr.16: 31039635-31039637 on Build GRCh38
rs748504322 Chr.16: 31039539-31039539 on Build GRCh38
rs753663211 Chr.16: 31039490-31039490 on Build GRCh38
rs759458097 Chr.16: 31039584-31039584 on Build GRCh38
rs1049474818 Chr.16: 31039530-31039530 on Build GRCh38
rs781601908 Chr.16: 31039531-31039531 on Build GRCh38
rs199709113 Chr.16: 31039592-31039592 on Build GRCh38
rs532998208 Chr.16: 31039552-31039552 on Build GRCh38
rs562489469 Chr.16: 31039554-31039554 on Build GRCh38
rs753558102 Chr.16: 31039505-31039505 on Build GRCh38
rs765803811 Chr.16: 31039798-31039798 on Build GRCh38
rs372737064 Chr.16: 31039670-31039670 on Build GRCh38
rs775241348 Chr.16: 31039669-31039669 on Build GRCh38
rs774245402 Chr.16: 31039847-31039848 on Build GRCh38
rs770229721 Chr.16: 31039540-31039540 on Build GRCh38
rs778576478 Chr.16: 31039511-31039511 on Build GRCh38
rs200786971 Chr.16: 31039599-31039599 on Build GRCh38
rs764353518 Chr.16: 31039753-31039753 on Build GRCh38
rs145848804 Chr.16: 31039624-31039624 on Build GRCh38
rs770931989 Chr.16: 31039556-31039556 on Build GRCh38
rs750133929 Chr.16: 31039703-31039703 on Build GRCh38
rs761558550 Chr.16: 31039603-31039603 on Build GRCh38
rs1032246469 Chr.16: 31039852-31039852 on Build GRCh38
rs376316073 Chr.16: 31039644-31039644 on Build GRCh38
rs778204069 Chr.16: 31039645-31039645 on Build GRCh38
rs893251464 Chr.16: 31039578-31039578 on Build GRCh38
rs769736866 Chr.16: 31039786-31039786 on Build GRCh38
rs760346516 Chr.16: 31039598-31039598 on Build GRCh38
rs750008975 Chr.16: 31039617-31039617 on Build GRCh38
rs755606689 Chr.16: 31039526-31039526 on Build GRCh38
rs765598299 Chr.16: 31039840-31039840 on Build GRCh38
rs1033941416 Chr.16: 31039616-31039616 on Build GRCh38
rs544762593 Chr.16: 31039687-31039687 on Build GRCh38
rs752495327 Chr.16: 31039491-31039491 on Build GRCh38
rs959584523 Chr.16: 31039909-31039909 on Build GRCh38
rs754251503 Chr.16: 31039755-31039755 on Build GRCh38
rs745985110 Chr.16: 31039664-31039664 on Build GRCh38
rs776631760 Chr.16: 31039684-31039684 on Build GRCh38
rs890878699 Chr.16: 31039771-31039771 on Build GRCh38
rs1027088927 Chr.16: 31039861-31039861 on Build GRCh38
rs560039970 Chr.16: 31039758-31039758 on Build GRCh38
rs762675311 Chr.16: 31039793-31039793 on Build GRCh38
rs139037938 Chr.16: 31039612-31039612 on Build GRCh38
rs765930924 Chr.16: 31039709-31039709 on Build GRCh38
rs752043308 Chr.16: 31039524-31039524 on Build GRCh38
rs966581561 Chr.16: 31039609-31039609 on Build GRCh38
rs772224599 Chr.16: 31039668-31039668 on Build GRCh38
rs762324647 Chr.16: 31039708-31039708 on Build GRCh38
rs763694019 Chr.16: 31039501-31039501 on Build GRCh38
rs770465950 Chr.16: 31039848-31039859 on Build GRCh38
rs34576482 Chr.16: 31039808-31039808 on Build GRCh38
rs907389335 Chr.16: 31039773-31039773 on Build GRCh38
rs753079913 Chr.16: 31039729-31039729 on Build GRCh38

View Full Product Details