Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46967482-46968030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA2
Entrez Gene ID
10294
Gene Name
DnaJ heat shock protein family (Hsp40) member A2
Gene Aliases
CPR3, DJ3, DJA2, DNAJ, DNJ3, HIRIP4, PRO3015, RDJ2
UniGene
Hs.368078
Gene Chromosome Location
Chr.16: 46955362-46973714 on Build GRCh38
Transcripts
NM_005880.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3691018 c.504C>T p.R168R Chr.16: 46967586-46967586 on Build GRCh38
970805 c.448G>T p.G150C Chr.16: 46967642-46967642 on Build GRCh38
5603414 c.526C>T p.P176S Chr.16: 46967564-46967564 on Build GRCh38
3509724 c.445C>T p.Q149* Chr.16: 46967645-46967645 on Build GRCh38
5740638 c.488G>A p.R163Q Chr.16: 46967602-46967602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898482969 Chr.16: 46967898-46967898 on Build GRCh38
rs1051440908 Chr.16: 46967868-46967868 on Build GRCh38
rs898618979 Chr.16: 46967982-46967982 on Build GRCh38
rs969478506 Chr.16: 46967970-46967970 on Build GRCh38
rs780723242 Chr.16: 46967535-46967535 on Build GRCh38
rs1031285554 Chr.16: 46967941-46967941 on Build GRCh38
rs750010396 Chr.16: 46967652-46967652 on Build GRCh38
rs909106903 Chr.16: 46967763-46967763 on Build GRCh38
rs772277874 Chr.16: 46967599-46967599 on Build GRCh38
rs996822616 Chr.16: 46967983-46967983 on Build GRCh38
rs937654701 Chr.16: 46967949-46967949 on Build GRCh38
rs955565112 Chr.16: 46967700-46967700 on Build GRCh38
rs2034598 Chr.16: 46967714-46967714 on Build GRCh38
rs575291050 Chr.16: 46967640-46967640 on Build GRCh38
rs563442946 Chr.16: 46967609-46967609 on Build GRCh38
rs955818883 Chr.16: 46967923-46967923 on Build GRCh38
rs777006758 Chr.16: 46967927-46967927 on Build GRCh38
rs1038414465 Chr.16: 46967976-46967976 on Build GRCh38
rs773304512 Chr.16: 46967601-46967601 on Build GRCh38
rs919202344 Chr.16: 46967987-46967987 on Build GRCh38
rs148966873 Chr.16: 46967887-46967887 on Build GRCh38
rs918045730 Chr.16: 46967727-46967727 on Build GRCh38
rs986946348 Chr.16: 46967924-46967924 on Build GRCh38
rs962903907 Chr.16: 46967598-46967598 on Build GRCh38
rs761096342 Chr.16: 46967768-46967768 on Build GRCh38
rs777158867 Chr.16: 46967613-46967613 on Build GRCh38
rs879181822 Chr.16: 46967883-46967883 on Build GRCh38
rs956689886 Chr.16: 46967947-46967947 on Build GRCh38
rs760999558 Chr.16: 46967607-46967607 on Build GRCh38
rs913614609 Chr.16: 46967704-46967704 on Build GRCh38
rs749201446 Chr.16: 46967574-46967574 on Build GRCh38
rs567180063 Chr.16: 46967553-46967553 on Build GRCh38
rs1028566093 Chr.16: 46967914-46967914 on Build GRCh38
rs542243176 Chr.16: 46967788-46967788 on Build GRCh38
rs745927844 Chr.16: 46967590-46967590 on Build GRCh38
rs112975508 Chr.16: 46967935-46967935 on Build GRCh38
rs917803614 Chr.16: 46967957-46967957 on Build GRCh38
rs886258940 Chr.16: 46967972-46967972 on Build GRCh38
rs750150488 Chr.16: 46967543-46967543 on Build GRCh38
rs896431328 Chr.16: 46967754-46967754 on Build GRCh38
rs564235428 Chr.16: 46967929-46967929 on Build GRCh38
rs764372182 Chr.16: 46967902-46967902 on Build GRCh38
rs760084433 Chr.16: 46967619-46967619 on Build GRCh38
rs941891704 Chr.16: 46967973-46967973 on Build GRCh38
rs1004268079 Chr.16: 46967881-46967881 on Build GRCh38
rs949346757 Chr.16: 46967958-46967958 on Build GRCh38
rs1017002520 Chr.16: 46967893-46967893 on Build GRCh38
rs925065483 Chr.16: 46967942-46967942 on Build GRCh38
rs566350491 Chr.16: 46967997-46967997 on Build GRCh38
rs766109639 Chr.16: 46967667-46967667 on Build GRCh38
rs140371161 Chr.16: 46967588-46967588 on Build GRCh38
rs1035994972 Chr.16: 46967791-46967793 on Build GRCh38
rs529471972 Chr.16: 46967701-46967701 on Build GRCh38
rs373057510 Chr.16: 46967680-46967680 on Build GRCh38
rs1048024158 Chr.16: 46967792-46967792 on Build GRCh38
rs756868978 Chr.16: 46967513-46967513 on Build GRCh38
rs765997210 Chr.16: 46967952-46967952 on Build GRCh38
rs987406955 Chr.16: 46967703-46967703 on Build GRCh38
rs897474077 Chr.16: 46967791-46967791 on Build GRCh38
rs765847997 Chr.16: 46967624-46967624 on Build GRCh38
rs114801127 Chr.16: 46967776-46967776 on Build GRCh38
rs561798918 Chr.16: 46967989-46967989 on Build GRCh38
rs779007173 Chr.16: 46967585-46967585 on Build GRCh38
rs755809808 Chr.16: 46967660-46967660 on Build GRCh38
rs773304184 Chr.16: 46967961-46967961 on Build GRCh38
rs534125595 Chr.16: 46967587-46967587 on Build GRCh38
rs377753297 Chr.16: 46967673-46967673 on Build GRCh38
rs537901431 Chr.16: 46967720-46967720 on Build GRCh38
rs751026855 Chr.16: 46967634-46967634 on Build GRCh38
rs556258836 Chr.16: 46967723-46967723 on Build GRCh38
rs966419349 Chr.16: 46967934-46967934 on Build GRCh38
rs779863241 Chr.16: 46967559-46967559 on Build GRCh38
rs34724036 Chr.16: 46967769-46967770 on Build GRCh38
rs975623207 Chr.16: 46967608-46967608 on Build GRCh38
rs904067265 Chr.16: 46967930-46967930 on Build GRCh38
rs753663895 Chr.16: 46967669-46967669 on Build GRCh38
rs543517724 Chr.16: 46967984-46967984 on Build GRCh38
rs1047529917 Chr.16: 46967960-46967960 on Build GRCh38
rs1055050277 Chr.16: 46967744-46967744 on Build GRCh38
rs570719544 Chr.16: 46967718-46967718 on Build GRCh38
rs768603261 Chr.16: 46967582-46967582 on Build GRCh38
rs545673731 Chr.16: 46967892-46967892 on Build GRCh38
rs553761064 Chr.16: 46967822-46967822 on Build GRCh38
rs887170751 Chr.16: 46967860-46967860 on Build GRCh38
rs373164180 Chr.16: 46967726-46967726 on Build GRCh38
rs937685792 Chr.16: 46967737-46967737 on Build GRCh38
rs1006985897 Chr.16: 46967895-46967895 on Build GRCh38
rs910279461 Chr.16: 46967998-46967998 on Build GRCh38
rs772107935 Chr.16: 46967622-46967622 on Build GRCh38

View Full Product Details