Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 55871539-55872050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES5A
Entrez Gene ID
221223
Gene Name
carboxylesterase 5A
Gene Aliases
CAUXIN, CES4C1, CES5, CES7, HEL126
UniGene
Hs.350800
Gene Chromosome Location
Chr.16: 55846154-55956031 on Build GRCh38
Transcripts
NM_001143685.1 NM_001190158.1 NM_145024.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6144784 c.280T>A p.C94S Chr.16: 55871762-55871762 on Build GRCh38
6894847 c.416C>A p.P139H Chr.16: 55871626-55871626 on Build GRCh38
6894846 c.503C>A p.P168H Chr.16: 55871626-55871626 on Build GRCh38
5036227 c.329T>C p.V110A Chr.16: 55871713-55871713 on Build GRCh38
3818119 c.465C>G p.I155M Chr.16: 55871664-55871664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527503380 Chr.16: 55871915-55871915 on Build GRCh38
rs981814367 Chr.16: 55871963-55871963 on Build GRCh38
rs571308497 Chr.16: 55872013-55872013 on Build GRCh38
rs766662841 Chr.16: 55871620-55871620 on Build GRCh38
rs773621525 Chr.16: 55871609-55871609 on Build GRCh38
rs80178015 Chr.16: 55871703-55871703 on Build GRCh38
rs757390984 Chr.16: 55871793-55871793 on Build GRCh38
rs780278163 Chr.16: 55871596-55871596 on Build GRCh38
rs117887370 Chr.16: 55871624-55871624 on Build GRCh38
rs368224955 Chr.16: 55871607-55871607 on Build GRCh38
rs915486462 Chr.16: 55872024-55872024 on Build GRCh38
rs543903403 Chr.16: 55871646-55871646 on Build GRCh38
rs749037841 Chr.16: 55871791-55871791 on Build GRCh38
rs901286690 Chr.16: 55871888-55871888 on Build GRCh38
rs752977508 Chr.16: 55871704-55871704 on Build GRCh38
rs199716261 Chr.16: 55871626-55871626 on Build GRCh38
rs865854672 Chr.16: 55871627-55871627 on Build GRCh38
rs768790708 Chr.16: 55871598-55871598 on Build GRCh38
rs143198507 Chr.16: 55871648-55871648 on Build GRCh38
rs748298244 Chr.16: 55871601-55871601 on Build GRCh38
rs781695639 Chr.16: 55871716-55871716 on Build GRCh38
rs370635715 Chr.16: 55871715-55871715 on Build GRCh38
rs555322337 Chr.16: 55871847-55871847 on Build GRCh38
rs966978863 Chr.16: 55872026-55872026 on Build GRCh38
rs761050788 Chr.16: 55871616-55871616 on Build GRCh38
rs186095802 Chr.16: 55871935-55871935 on Build GRCh38
rs755880941 Chr.16: 55871645-55871645 on Build GRCh38
rs757227876 Chr.16: 55871859-55871859 on Build GRCh38
rs753666614 Chr.16: 55871699-55871699 on Build GRCh38
rs533154273 Chr.16: 55871710-55871710 on Build GRCh38
rs1026145878 Chr.16: 55871936-55871936 on Build GRCh38
rs766399380 Chr.16: 55871889-55871889 on Build GRCh38
rs758534289 Chr.16: 55871781-55871781 on Build GRCh38
rs772960947 Chr.16: 55871838-55871838 on Build GRCh38
rs149280225 Chr.16: 55871670-55871670 on Build GRCh38
rs899050009 Chr.16: 55871934-55871934 on Build GRCh38
rs529456613 Chr.16: 55871600-55871600 on Build GRCh38
rs1033362179 Chr.16: 55872001-55872001 on Build GRCh38
rs138909689 Chr.16: 55871878-55871878 on Build GRCh38
rs141931680 Chr.16: 55871996-55871996 on Build GRCh38
rs75888384 Chr.16: 55871914-55871914 on Build GRCh38
rs752605615 Chr.16: 55871777-55871777 on Build GRCh38
rs533130569 Chr.16: 55871658-55871658 on Build GRCh38
rs760526464 Chr.16: 55871696-55871696 on Build GRCh38
rs759343877 Chr.16: 55871776-55871776 on Build GRCh38
rs867444664 Chr.16: 55871946-55871946 on Build GRCh38
rs147327784 Chr.16: 55871748-55871748 on Build GRCh38
rs78002815 Chr.16: 55871856-55871856 on Build GRCh38
rs759940407 Chr.16: 55871666-55871666 on Build GRCh38
rs577855139 Chr.16: 55871819-55871819 on Build GRCh38
rs373871562 Chr.16: 55871833-55871834 on Build GRCh38
rs746388180 Chr.16: 55871809-55871809 on Build GRCh38
rs1042526420 Chr.16: 55871887-55871887 on Build GRCh38
rs78418359 Chr.16: 55871689-55871689 on Build GRCh38
rs748197821 Chr.16: 55871653-55871653 on Build GRCh38
rs181260793 Chr.16: 55871894-55871894 on Build GRCh38
rs970476775 Chr.16: 55871943-55871943 on Build GRCh38
rs201475444 Chr.16: 55871597-55871597 on Build GRCh38
rs766399736 Chr.16: 55871697-55871697 on Build GRCh38
rs771604902 Chr.16: 55871826-55871826 on Build GRCh38
rs112216945 Chr.16: 55871571-55871571 on Build GRCh38
rs1026493218 Chr.16: 55871832-55871832 on Build GRCh38
rs780397072 Chr.16: 55871820-55871820 on Build GRCh38
rs762845235 Chr.16: 55871855-55871855 on Build GRCh38
rs773935287 Chr.16: 55871737-55871737 on Build GRCh38
rs750308188 Chr.16: 55871692-55871692 on Build GRCh38
rs938264879 Chr.16: 55871693-55871693 on Build GRCh38
rs755896830 Chr.16: 55871589-55871589 on Build GRCh38
rs566269786 Chr.16: 55871842-55871842 on Build GRCh38
rs772066729 Chr.16: 55871654-55871654 on Build GRCh38
rs143432962 Chr.16: 55871676-55871676 on Build GRCh38
rs959050508 Chr.16: 55872005-55872005 on Build GRCh38
rs138444171 Chr.16: 55871661-55871661 on Build GRCh38
rs781629490 Chr.16: 55871798-55871798 on Build GRCh38
rs369304717 Chr.16: 55871635-55871635 on Build GRCh38
rs764424332 Chr.16: 55871786-55871786 on Build GRCh38
rs141262535 Chr.16: 55871843-55871843 on Build GRCh38
rs373286926 Chr.16: 55871652-55871652 on Build GRCh38
rs760284149 Chr.16: 55871759-55871759 on Build GRCh38
rs55703803 Chr.16: 55871569-55871569 on Build GRCh38
rs560882062 Chr.16: 55871981-55871981 on Build GRCh38
rs541523688 Chr.16: 55871901-55871901 on Build GRCh38
rs539219013 Chr.16: 55871835-55871835 on Build GRCh38
rs200930153 Chr.16: 55871785-55871785 on Build GRCh38
rs531185987 Chr.16: 55871986-55871986 on Build GRCh38
rs771828202 Chr.16: 55871743-55871743 on Build GRCh38
rs757803224 Chr.16: 55871765-55871765 on Build GRCh38
rs3859108 Chr.16: 55871724-55871724 on Build GRCh38
rs766954207 Chr.16: 55871681-55871681 on Build GRCh38
rs200734661 Chr.16: 55871673-55871673 on Build GRCh38
rs142239690 Chr.16: 55871576-55871576 on Build GRCh38
rs776729237 Chr.16: 55871768-55871768 on Build GRCh38
rs148234688 Chr.16: 55871659-55871659 on Build GRCh38
rs117055708 Chr.16: 55871841-55871841 on Build GRCh38
rs745647794 Chr.16: 55871577-55871577 on Build GRCh38
rs72810507 Chr.16: 55871764-55871764 on Build GRCh38
rs749723780 Chr.16: 55871822-55871822 on Build GRCh38
rs758432457 Chr.16: 55871706-55871706 on Build GRCh38
rs373069588 Chr.16: 55871637-55871637 on Build GRCh38
rs768269547 Chr.16: 55871667-55871667 on Build GRCh38
rs751560300 Chr.16: 55871789-55871789 on Build GRCh38
rs559093225 Chr.16: 55871890-55871890 on Build GRCh38
rs77612013 Chr.16: 55871738-55871738 on Build GRCh38
rs372330298 Chr.16: 55871723-55871723 on Build GRCh38
rs544955880 Chr.16: 55871605-55871605 on Build GRCh38
rs146474777 Chr.16: 55871625-55871625 on Build GRCh38
rs777709900 Chr.16: 55871709-55871709 on Build GRCh38
rs998194673 Chr.16: 55871733-55871733 on Build GRCh38
rs755239329 Chr.16: 55871649-55871649 on Build GRCh38
rs375083402 Chr.16: 55871643-55871643 on Build GRCh38

View Full Product Details