Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58116545-58117037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP20
Entrez Gene ID
29105
Gene Name
cilia and flagella associated protein 20
Gene Aliases
C16orf80, EVORF, GTL3, fSAP23
UniGene
Hs.532755
Gene Chromosome Location
Chr.16: 58113588-58129425 on Build GRCh38
Transcripts
NM_013242.2 NR_136632.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6614584 c.99C>T p.H33H Chr.16: 58116937-58116937 on Build GRCh38
6055172 c.89G>C p.R30P Chr.16: 58116947-58116947 on Build GRCh38
1640511 c.164+7T>C p.? Chr.16: 58116865-58116865 on Build GRCh38
1709232 c.131C>T p.S44F Chr.16: 58116905-58116905 on Build GRCh38
4061510 c.88C>T p.R30W Chr.16: 58116948-58116948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912245391 Chr.16: 58116630-58116630 on Build GRCh38
rs890189707 Chr.16: 58116765-58116765 on Build GRCh38
rs924929062 Chr.16: 58116591-58116591 on Build GRCh38
rs969227889 Chr.16: 58116818-58116818 on Build GRCh38
rs1039382768 Chr.16: 58116848-58116848 on Build GRCh38
rs900080525 Chr.16: 58116917-58116917 on Build GRCh38
rs762167347 Chr.16: 58116980-58116980 on Build GRCh38
rs751641684 Chr.16: 58116921-58116921 on Build GRCh38
rs768014590 Chr.16: 58116992-58116992 on Build GRCh38
rs565611994 Chr.16: 58116672-58116672 on Build GRCh38
rs146983372 Chr.16: 58116814-58116814 on Build GRCh38
rs969207274 Chr.16: 58116798-58116798 on Build GRCh38
rs995390670 Chr.16: 58116699-58116699 on Build GRCh38
rs928045153 Chr.16: 58116581-58116581 on Build GRCh38
rs199567935 Chr.16: 58116996-58116996 on Build GRCh38
rs199717197 Chr.16: 58116970-58116970 on Build GRCh38
rs867094284 Chr.16: 58116905-58116905 on Build GRCh38
rs764578076 Chr.16: 58116975-58116975 on Build GRCh38
rs558423067 Chr.16: 58116937-58116937 on Build GRCh38
rs960035866 Chr.16: 58116758-58116758 on Build GRCh38
rs569612903 Chr.16: 58116844-58116844 on Build GRCh38
rs375028807 Chr.16: 58116872-58116872 on Build GRCh38
rs536685889 Chr.16: 58116894-58116894 on Build GRCh38
rs1004983605 Chr.16: 58116773-58116773 on Build GRCh38
rs1052429073 Chr.16: 58116622-58116622 on Build GRCh38
rs978977136 Chr.16: 58116813-58116813 on Build GRCh38
rs754628799 Chr.16: 58116880-58116880 on Build GRCh38
rs548014135 Chr.16: 58116704-58116704 on Build GRCh38
rs764641089 Chr.16: 58116846-58116846 on Build GRCh38
rs915206240 Chr.16: 58116889-58116889 on Build GRCh38
rs762314571 Chr.16: 58117001-58117001 on Build GRCh38
rs754717000 Chr.16: 58116997-58116997 on Build GRCh38
rs776364234 Chr.16: 58116955-58116955 on Build GRCh38
rs1014564416 Chr.16: 58116776-58116776 on Build GRCh38
rs1048684472 Chr.16: 58116710-58116710 on Build GRCh38
rs758312713 Chr.16: 58116897-58116897 on Build GRCh38
rs1035545281 Chr.16: 58116742-58116742 on Build GRCh38
rs868427039 Chr.16: 58116861-58116861 on Build GRCh38
rs759796352 Chr.16: 58116835-58116835 on Build GRCh38
rs375373333 Chr.16: 58116829-58116829 on Build GRCh38
rs1056987735 Chr.16: 58117016-58117016 on Build GRCh38
rs113807913 Chr.16: 58116669-58116669 on Build GRCh38
rs887058393 Chr.16: 58116724-58116724 on Build GRCh38
rs111955536 Chr.16: 58116819-58116819 on Build GRCh38
rs924762364 Chr.16: 58116816-58116816 on Build GRCh38
rs943646740 Chr.16: 58116647-58116647 on Build GRCh38
rs778596857 Chr.16: 58116893-58116893 on Build GRCh38
rs374561290 Chr.16: 58116847-58116847 on Build GRCh38
rs746844193 Chr.16: 58116924-58116924 on Build GRCh38
rs761193487 Chr.16: 58116797-58116797 on Build GRCh38
rs894061098 Chr.16: 58116624-58116624 on Build GRCh38
rs748397056 Chr.16: 58116592-58116592 on Build GRCh38
rs973307758 Chr.16: 58116792-58116792 on Build GRCh38
rs939820671 Chr.16: 58117016-58117016 on Build GRCh38
rs769976697 Chr.16: 58116961-58116961 on Build GRCh38
rs777751037 Chr.16: 58116923-58116923 on Build GRCh38
rs775072906 Chr.16: 58116830-58116830 on Build GRCh38
rs946965096 Chr.16: 58116627-58116627 on Build GRCh38
rs370808916 Chr.16: 58116855-58116855 on Build GRCh38
rs141340242 Chr.16: 58116741-58116741 on Build GRCh38
rs113679510 Chr.16: 58116693-58116693 on Build GRCh38
rs544107070 Chr.16: 58116573-58116573 on Build GRCh38
rs746124389 Chr.16: 58116958-58116958 on Build GRCh38
rs998345218 Chr.16: 58116716-58116716 on Build GRCh38
rs1015008563 Chr.16: 58117006-58117006 on Build GRCh38
rs545060310 Chr.16: 58116574-58116574 on Build GRCh38
rs767928662 Chr.16: 58116871-58116871 on Build GRCh38
rs368336221 Chr.16: 58116969-58116969 on Build GRCh38
rs528724157 Chr.16: 58116791-58116791 on Build GRCh38

View Full Product Details