Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66844818-66845311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA7
Entrez Gene ID
766
Gene Name
carbonic anhydrase 7
Gene Aliases
CAVII
UniGene
Hs.37014
Gene Chromosome Location
Chr.16: 66844379-66854149 on Build GRCh38
Transcripts
NM_001014435.1 NM_005182.2 XM_005256136.3 XM_011523311.1 XM_011523312.1

SNP Detail

SNP ID Chromosome Location
rs1027650240 Chr.16: 66844947-66844947 on Build GRCh38
rs1014795265 Chr.16: 66844964-66844964 on Build GRCh38
rs562224660 Chr.16: 66844856-66844856 on Build GRCh38
rs533207496 Chr.16: 66845038-66845038 on Build GRCh38
rs887764579 Chr.16: 66845249-66845249 on Build GRCh38
rs1004858563 Chr.16: 66845250-66845250 on Build GRCh38
rs891939313 Chr.16: 66844912-66844912 on Build GRCh38
rs1006799561 Chr.16: 66844870-66844870 on Build GRCh38
rs926214194 Chr.16: 66844881-66844881 on Build GRCh38
rs544394844 Chr.16: 66844923-66844923 on Build GRCh38
rs745373929 Chr.16: 66845166-66845166 on Build GRCh38
rs936247735 Chr.16: 66844888-66844888 on Build GRCh38
rs769427804 Chr.16: 66845176-66845176 on Build GRCh38
rs1036706002 Chr.16: 66845282-66845282 on Build GRCh38
rs950189203 Chr.16: 66844926-66844926 on Build GRCh38
rs957639793 Chr.16: 66845102-66845102 on Build GRCh38
rs1007240452 Chr.16: 66845177-66845177 on Build GRCh38
rs979026628 Chr.16: 66844880-66844880 on Build GRCh38
rs965188071 Chr.16: 66844883-66844883 on Build GRCh38
rs991069410 Chr.16: 66845075-66845075 on Build GRCh38
rs1046255144 Chr.16: 66844932-66844932 on Build GRCh38
rs1019486203 Chr.16: 66844875-66844875 on Build GRCh38
rs997335858 Chr.16: 66844968-66844968 on Build GRCh38
rs1029161861 Chr.16: 66844981-66844981 on Build GRCh38
rs564250341 Chr.16: 66844982-66844982 on Build GRCh38
rs938293101 Chr.16: 66845006-66845006 on Build GRCh38
rs182123538 Chr.16: 66845113-66845113 on Build GRCh38
rs972536033 Chr.16: 66844886-66844886 on Build GRCh38
rs1053253641 Chr.16: 66844893-66844893 on Build GRCh38
rs531185411 Chr.16: 66844869-66844869 on Build GRCh38
rs1004786475 Chr.16: 66844963-66844963 on Build GRCh38
rs989008067 Chr.16: 66845109-66845109 on Build GRCh38
rs918145775 Chr.16: 66845091-66845091 on Build GRCh38
rs996670718 Chr.16: 66845203-66845203 on Build GRCh38
rs1049115834 Chr.16: 66845247-66845247 on Build GRCh38
rs905910052 Chr.16: 66844933-66844933 on Build GRCh38
rs924946424 Chr.16: 66845057-66845057 on Build GRCh38
rs529254965 Chr.16: 66845204-66845204 on Build GRCh38
rs887734784 Chr.16: 66844960-66844960 on Build GRCh38
rs746939354 Chr.16: 66844871-66844871 on Build GRCh38
rs926973418 Chr.16: 66844995-66844995 on Build GRCh38
rs979100136 Chr.16: 66845008-66845008 on Build GRCh38
rs965846531 Chr.16: 66844848-66844848 on Build GRCh38
rs1045867404 Chr.16: 66845139-66845139 on Build GRCh38
rs1025409349 Chr.16: 66844910-66844910 on Build GRCh38
rs779873916 Chr.16: 66845196-66845196 on Build GRCh38
rs1042499429 Chr.16: 66845114-66845114 on Build GRCh38
rs901058656 Chr.16: 66845260-66845260 on Build GRCh38
rs757195485 Chr.16: 66844942-66844942 on Build GRCh38
rs902608534 Chr.16: 66845118-66845118 on Build GRCh38
rs12102712 Chr.16: 66845138-66845138 on Build GRCh38
rs966284713 Chr.16: 66844966-66844966 on Build GRCh38
rs953122135 Chr.16: 66844994-66844994 on Build GRCh38

View Full Product Details