Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66886279-66886791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDP2
Entrez Gene ID
57546
Gene Name
pyruvate dehyrogenase phosphatase catalytic subunit 2
Gene Aliases
PPM2B, PPM2C2
UniGene
Hs.632214
Gene Chromosome Location
Chr.16: 66880397-66891101 on Build GRCh38
Transcripts
NM_020786.2 XM_005256064.4 XM_005256066.4 XM_005256067.4 XM_005256068.4 XM_017023506.1

SNP Detail

SNP ID Chromosome Location
rs963828920 Chr.16: 66886446-66886446 on Build GRCh38
rs749694985 Chr.16: 66886503-66886503 on Build GRCh38
rs1054766739 Chr.16: 66886614-66886614 on Build GRCh38
rs895257431 Chr.16: 66886309-66886309 on Build GRCh38
rs531026993 Chr.16: 66886427-66886427 on Build GRCh38
rs1007336515 Chr.16: 66886405-66886406 on Build GRCh38
rs981474505 Chr.16: 66886571-66886571 on Build GRCh38
rs991643971 Chr.16: 66886539-66886539 on Build GRCh38
rs749772024 Chr.16: 66886516-66886516 on Build GRCh38
rs570935684 Chr.16: 66886491-66886491 on Build GRCh38
rs950079232 Chr.16: 66886550-66886550 on Build GRCh38
rs934564379 Chr.16: 66886383-66886383 on Build GRCh38
rs898554478 Chr.16: 66886608-66886608 on Build GRCh38
rs1025998856 Chr.16: 66886414-66886414 on Build GRCh38
rs1017319487 Chr.16: 66886406-66886406 on Build GRCh38
rs769219321 Chr.16: 66886511-66886511 on Build GRCh38
rs903108980 Chr.16: 66886331-66886331 on Build GRCh38
rs550796293 Chr.16: 66886450-66886450 on Build GRCh38
rs994586638 Chr.16: 66886645-66886645 on Build GRCh38
rs769308230 Chr.16: 66886520-66886520 on Build GRCh38
rs756536680 Chr.16: 66886422-66886422 on Build GRCh38
rs11861556 Chr.16: 66886549-66886549 on Build GRCh38
rs568387467 Chr.16: 66886416-66886416 on Build GRCh38
rs924216572 Chr.16: 66886532-66886532 on Build GRCh38
rs1038801182 Chr.16: 66886735-66886735 on Build GRCh38
rs772177177 Chr.16: 66886592-66886592 on Build GRCh38
rs991962654 Chr.16: 66886307-66886307 on Build GRCh38
rs573289988 Chr.16: 66886385-66886385 on Build GRCh38
rs1016924375 Chr.16: 66886365-66886365 on Build GRCh38
rs548474284 Chr.16: 66886394-66886394 on Build GRCh38
rs989892088 Chr.16: 66886467-66886467 on Build GRCh38
rs28398401 Chr.16: 66886607-66886607 on Build GRCh38
rs184521572 Chr.16: 66886316-66886316 on Build GRCh38
rs372166645 Chr.16: 66886415-66886416 on Build GRCh38
rs1017127428 Chr.16: 66886530-66886530 on Build GRCh38
rs780636633 Chr.16: 66886424-66886424 on Build GRCh38
rs1045738849 Chr.16: 66886319-66886319 on Build GRCh38
rs1056543944 Chr.16: 66886306-66886306 on Build GRCh38
rs914464106 Chr.16: 66886712-66886712 on Build GRCh38
rs1025633652 Chr.16: 66886659-66886659 on Build GRCh38
rs1056596615 Chr.16: 66886551-66886551 on Build GRCh38
rs1004138791 Chr.16: 66886305-66886305 on Build GRCh38
rs528841581 Chr.16: 66886370-66886370 on Build GRCh38
rs78861024 Chr.16: 66886720-66886720 on Build GRCh38
rs915943341 Chr.16: 66886311-66886311 on Build GRCh38
rs943213092 Chr.16: 66886713-66886713 on Build GRCh38
rs901611086 Chr.16: 66886750-66886750 on Build GRCh38
rs994141623 Chr.16: 66886407-66886407 on Build GRCh38
rs370604800 Chr.16: 66886388-66886388 on Build GRCh38
rs951668120 Chr.16: 66886500-66886500 on Build GRCh38
rs748292579 Chr.16: 66886585-66886585 on Build GRCh38

View Full Product Details