Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66995543-66996031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES4A
Entrez Gene ID
283848
Gene Name
carboxylesterase 4A
Gene Aliases
CES6, CES8
UniGene
Hs.346947
Gene Chromosome Location
Chr.16: 66988589-67009758 on Build GRCh38
Transcripts
NM_173815.6 XM_005255896.4 XM_011523021.2 XM_011523023.2 XM_011523024.2 XM_011523025.2 XM_011523026.2 XM_011523027.2 XM_011523028.2 XM_011523029.2 XM_011523030.2 XM_017023160.1 XM_017023161.1 XM_017023162.1 XM_017023163.1 XM_017023164.1 XM_017023165.1 XM_017023166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1579480 c.163C>T p.P55S Chr.16: 66995732-66995732 on Build GRCh38
5544684 c.120G>A p.Q40Q Chr.16: 66995689-66995689 on Build GRCh38
70159 c.251A>C p.Y84S Chr.16: 66995820-66995820 on Build GRCh38
5971851 c.118C>G p.Q40E Chr.16: 66995687-66995687 on Build GRCh38
2994058 c.254C>A p.P85Q Chr.16: 66995823-66995823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368774734 Chr.16: 66995650-66995650 on Build GRCh38
rs748351246 Chr.16: 66995742-66995742 on Build GRCh38
rs190027245 Chr.16: 66995716-66995716 on Build GRCh38
rs778459412 Chr.16: 66995591-66995591 on Build GRCh38
rs779116353 Chr.16: 66995832-66995832 on Build GRCh38
rs181090965 Chr.16: 66995837-66995837 on Build GRCh38
rs1030259395 Chr.16: 66995717-66995717 on Build GRCh38
rs752250953 Chr.16: 66995661-66995661 on Build GRCh38
rs746274234 Chr.16: 66995703-66995703 on Build GRCh38
rs769432095 Chr.16: 66995865-66995865 on Build GRCh38
rs547202214 Chr.16: 66995974-66995974 on Build GRCh38
rs780357459 Chr.16: 66995718-66995718 on Build GRCh38
rs758035761 Chr.16: 66995593-66995593 on Build GRCh38
rs374816669 Chr.16: 66995579-66995579 on Build GRCh38
rs559191657 Chr.16: 66995881-66995881 on Build GRCh38
rs746350982 Chr.16: 66995601-66995601 on Build GRCh38
rs774606089 Chr.16: 66995740-66995740 on Build GRCh38
rs543139450 Chr.16: 66995801-66995801 on Build GRCh38
rs1005517357 Chr.16: 66995933-66995933 on Build GRCh38
rs746674439 Chr.16: 66995595-66995595 on Build GRCh38
rs1034991786 Chr.16: 66995707-66995707 on Build GRCh38
rs1038795866 Chr.16: 66995937-66995937 on Build GRCh38
rs897210819 Chr.16: 66995939-66995939 on Build GRCh38
rs756374290 Chr.16: 66995821-66995821 on Build GRCh38
rs1042167676 Chr.16: 66995859-66995859 on Build GRCh38
rs759320354 Chr.16: 66995764-66995764 on Build GRCh38
rs199679268 Chr.16: 66995589-66995589 on Build GRCh38
rs750095223 Chr.16: 66995817-66995817 on Build GRCh38
rs374790706 Chr.16: 66995953-66995953 on Build GRCh38
rs767140273 Chr.16: 66995809-66995809 on Build GRCh38
rs371877898 Chr.16: 66995592-66995592 on Build GRCh38
rs767264840 Chr.16: 66995780-66995780 on Build GRCh38
rs762852498 Chr.16: 66995879-66995879 on Build GRCh38
rs944409678 Chr.16: 66995916-66995916 on Build GRCh38
rs565055593 Chr.16: 66995962-66995962 on Build GRCh38
rs61749482 Chr.16: 66995784-66995784 on Build GRCh38
rs538562443 Chr.16: 66995603-66995603 on Build GRCh38
rs747273397 Chr.16: 66995863-66995863 on Build GRCh38
rs375849213 Chr.16: 66995599-66995599 on Build GRCh38
rs774657354 Chr.16: 66995637-66995637 on Build GRCh38
rs368938557 Chr.16: 66995780-66995780 on Build GRCh38
rs891287651 Chr.16: 66995975-66995975 on Build GRCh38
rs376340766 Chr.16: 66995823-66995823 on Build GRCh38
rs765087870 Chr.16: 66995772-66995772 on Build GRCh38
rs144460906 Chr.16: 66995700-66995700 on Build GRCh38
rs776476618 Chr.16: 66995753-66995753 on Build GRCh38
rs578173809 Chr.16: 66995681-66995681 on Build GRCh38
rs374808886 Chr.16: 66995668-66995668 on Build GRCh38
rs895073469 Chr.16: 66995927-66995927 on Build GRCh38
rs763009751 Chr.16: 66995615-66995615 on Build GRCh38
rs775625714 Chr.16: 66995613-66995613 on Build GRCh38
rs759497902 Chr.16: 66995653-66995653 on Build GRCh38
rs756940311 Chr.16: 66995694-66995694 on Build GRCh38
rs559135863 Chr.16: 66995762-66995762 on Build GRCh38
rs766238040 Chr.16: 66995648-66995648 on Build GRCh38
rs911541683 Chr.16: 66995883-66995883 on Build GRCh38
rs773244767 Chr.16: 66995857-66995857 on Build GRCh38
rs763613217 Chr.16: 66995682-66995682 on Build GRCh38
rs377408075 Chr.16: 66995824-66995824 on Build GRCh38
rs762151263 Chr.16: 66995641-66995641 on Build GRCh38
rs757557968 Chr.16: 66995863-66995864 on Build GRCh38
rs1055039806 Chr.16: 66995899-66995899 on Build GRCh38
rs372106864 Chr.16: 66995708-66995708 on Build GRCh38
rs369754930 Chr.16: 66995850-66995850 on Build GRCh38
rs570712906 Chr.16: 66995842-66995843 on Build GRCh38
rs772475063 Chr.16: 66995747-66995747 on Build GRCh38
rs574184459 Chr.16: 66995789-66995789 on Build GRCh38
rs757841884 Chr.16: 66995664-66995664 on Build GRCh38
rs375062290 Chr.16: 66995693-66995693 on Build GRCh38
rs749827189 Chr.16: 66995721-66995721 on Build GRCh38
rs762645374 Chr.16: 66995701-66995701 on Build GRCh38
rs187002887 Chr.16: 66995973-66995973 on Build GRCh38
rs905135906 Chr.16: 66995678-66995678 on Build GRCh38
rs771234444 Chr.16: 66995598-66995598 on Build GRCh38
rs751170926 Chr.16: 66995687-66995687 on Build GRCh38
rs775392623 Chr.16: 66995869-66995869 on Build GRCh38
rs369605074 Chr.16: 66995712-66995712 on Build GRCh38
rs939205520 Chr.16: 66995893-66995893 on Build GRCh38
rs768862443 Chr.16: 66995636-66995636 on Build GRCh38
rs753761130 Chr.16: 66995582-66995582 on Build GRCh38
rs768807367 Chr.16: 66995739-66995739 on Build GRCh38
rs373178908 Chr.16: 66995785-66995785 on Build GRCh38
rs372259602 Chr.16: 66995656-66995656 on Build GRCh38
rs777472258 Chr.16: 66995594-66995594 on Build GRCh38
rs914971487 Chr.16: 66995840-66995840 on Build GRCh38

View Full Product Details