Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67148531-67149039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT9
Entrez Gene ID
84752
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 9
Gene Aliases
-
UniGene
Hs.744158
Gene Chromosome Location
Chr.16: 67148102-67150999 on Build GRCh38
Transcripts
NM_033309.2 NM_001320540.1 NM_001320541.1 NM_001320542.1 NM_001320543.1 NM_025187.4 XM_006721284.3 XM_011523350.2 XM_017023729.1

Gene Symbol
C16orf70
Entrez Gene ID
80262
Gene Name
chromosome 16 open reading frame 70
Gene Aliases
C16orf6, LIN10, lin-10
UniGene
Hs.513666
Gene Chromosome Location
Chr.16: 67109881-67148544 on Build GRCh38
Transcripts
NM_033309.2 NM_001320540.1 NM_001320541.1 NM_001320542.1 NM_001320543.1 NM_025187.4 XM_006721284.3 XM_011523350.2 XM_017023729.1

SNP Detail

SNP ID Chromosome Location
rs932843919 Chr.16: 67148743-67148743 on Build GRCh38
rs1010723056 Chr.16: 67148695-67148699 on Build GRCh38
rs928880468 Chr.16: 67148861-67148862 on Build GRCh38
rs949016057 Chr.16: 67148591-67148591 on Build GRCh38
rs946489518 Chr.16: 67148891-67148891 on Build GRCh38
rs756113022 Chr.16: 67148639-67148639 on Build GRCh38
rs997203214 Chr.16: 67148769-67148770 on Build GRCh38
rs955339711 Chr.16: 67148797-67148797 on Build GRCh38
rs778246083 Chr.16: 67148636-67148636 on Build GRCh38
rs368296957 Chr.16: 67149017-67149017 on Build GRCh38
rs368523989 Chr.16: 67148695-67148695 on Build GRCh38
rs1013301318 Chr.16: 67148605-67148605 on Build GRCh38
rs188607326 Chr.16: 67149010-67149010 on Build GRCh38
rs1002462735 Chr.16: 67148980-67148980 on Build GRCh38
rs996698342 Chr.16: 67148953-67148953 on Build GRCh38
rs1000963083 Chr.16: 67148559-67148559 on Build GRCh38
rs145270821 Chr.16: 67148859-67148860 on Build GRCh38
rs1029581540 Chr.16: 67148783-67148783 on Build GRCh38
rs944130324 Chr.16: 67148706-67148706 on Build GRCh38
rs934901279 Chr.16: 67148846-67148846 on Build GRCh38
rs982757273 Chr.16: 67148798-67148798 on Build GRCh38
rs771438722 Chr.16: 67148801-67148801 on Build GRCh38
rs556660421 Chr.16: 67148943-67148943 on Build GRCh38
rs891058824 Chr.16: 67149009-67149009 on Build GRCh38
rs554469225 Chr.16: 67148813-67148813 on Build GRCh38
rs574304990 Chr.16: 67148881-67148881 on Build GRCh38
rs980616799 Chr.16: 67148641-67148641 on Build GRCh38
rs919859671 Chr.16: 67148660-67148660 on Build GRCh38
rs899623398 Chr.16: 67148910-67148910 on Build GRCh38
rs1052151211 Chr.16: 67148619-67148619 on Build GRCh38
rs1024725932 Chr.16: 67148624-67148624 on Build GRCh38
rs908514321 Chr.16: 67148702-67148702 on Build GRCh38
rs956456884 Chr.16: 67148602-67148602 on Build GRCh38
rs916098198 Chr.16: 67148820-67148820 on Build GRCh38
rs147313315 Chr.16: 67148877-67148892 on Build GRCh38
rs1024758514 Chr.16: 67148893-67148893 on Build GRCh38
rs534502858 Chr.16: 67148725-67148725 on Build GRCh38
rs146150540 Chr.16: 67148984-67148984 on Build GRCh38
rs904791179 Chr.16: 67148815-67148815 on Build GRCh38
rs939002579 Chr.16: 67148610-67148610 on Build GRCh38
rs962791522 Chr.16: 67148814-67148814 on Build GRCh38
rs1052109313 Chr.16: 67148860-67148860 on Build GRCh38
rs770732587 Chr.16: 67148742-67148742 on Build GRCh38
rs921342272 Chr.16: 67148728-67148728 on Build GRCh38
rs183615064 Chr.16: 67148900-67148900 on Build GRCh38
rs572824879 Chr.16: 67148715-67148715 on Build GRCh38
rs892174285 Chr.16: 67148863-67148863 on Build GRCh38
rs75897856 Chr.16: 67148599-67148599 on Build GRCh38
rs976424662 Chr.16: 67148872-67148872 on Build GRCh38
rs892233292 Chr.16: 67148693-67148693 on Build GRCh38
rs777674653 Chr.16: 67148690-67148690 on Build GRCh38
rs775823959 Chr.16: 67148999-67148999 on Build GRCh38
rs948995275 Chr.16: 67148837-67148837 on Build GRCh38

View Full Product Details