Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71635733-71636233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARVELD3
Entrez Gene ID
91862
Gene Name
MARVEL domain containing 3
Gene Aliases
MARVD3, MRVLDC3
UniGene
Hs.513706
Gene Chromosome Location
Chr.16: 71626137-71641965 on Build GRCh38
Transcripts
NM_001017967.3 NM_001271329.1 NM_052858.5 XM_011523449.2

SNP Detail

SNP ID Chromosome Location
rs767615594 Chr.16: 71636160-71636160 on Build GRCh38
rs574534586 Chr.16: 71636127-71636127 on Build GRCh38
rs913336003 Chr.16: 71635975-71635975 on Build GRCh38
rs575997016 Chr.16: 71636128-71636128 on Build GRCh38
rs1029264172 Chr.16: 71635823-71635823 on Build GRCh38
rs183261118 Chr.16: 71636103-71636103 on Build GRCh38
rs1035369648 Chr.16: 71636151-71636151 on Build GRCh38
rs370562143 Chr.16: 71635868-71635868 on Build GRCh38
rs1031465726 Chr.16: 71636102-71636102 on Build GRCh38
rs890504537 Chr.16: 71636081-71636081 on Build GRCh38
rs970019295 Chr.16: 71636145-71636145 on Build GRCh38
rs990737535 Chr.16: 71636088-71636088 on Build GRCh38
rs1037990116 Chr.16: 71635925-71635925 on Build GRCh38
rs866445793 Chr.16: 71635984-71635984 on Build GRCh38
rs570198238 Chr.16: 71635910-71635910 on Build GRCh38
rs938457775 Chr.16: 71635829-71635829 on Build GRCh38
rs1011436023 Chr.16: 71636112-71636112 on Build GRCh38
rs900488141 Chr.16: 71636008-71636008 on Build GRCh38
rs376362936 Chr.16: 71635810-71635810 on Build GRCh38
rs1051786088 Chr.16: 71636057-71636057 on Build GRCh38
rs762681501 Chr.16: 71635784-71635784 on Build GRCh38
rs1021521569 Chr.16: 71636144-71636144 on Build GRCh38
rs192508525 Chr.16: 71635799-71635799 on Build GRCh38
rs765883593 Chr.16: 71635793-71635793 on Build GRCh38
rs533516832 Chr.16: 71635832-71635832 on Build GRCh38
rs1005345605 Chr.16: 71635770-71635770 on Build GRCh38
rs931998025 Chr.16: 71636034-71636034 on Build GRCh38
rs983991026 Chr.16: 71635830-71635830 on Build GRCh38
rs979858958 Chr.16: 71635791-71635791 on Build GRCh38
rs1050771464 Chr.16: 71636205-71636205 on Build GRCh38
rs1056303 Chr.16: 71636163-71636163 on Build GRCh38
rs980145917 Chr.16: 71636150-71636150 on Build GRCh38
rs756080923 Chr.16: 71636192-71636192 on Build GRCh38
rs953689049 Chr.16: 71635849-71635849 on Build GRCh38
rs548826137 Chr.16: 71635778-71635778 on Build GRCh38
rs971488603 Chr.16: 71635985-71635985 on Build GRCh38
rs879424734 Chr.16: 71636162-71636163 on Build GRCh38
rs112433481 Chr.16: 71636090-71636090 on Build GRCh38
rs751439134 Chr.16: 71635858-71635858 on Build GRCh38
rs997834692 Chr.16: 71635822-71635822 on Build GRCh38

View Full Product Details