Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71649604-71650130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHLPP2
Entrez Gene ID
23035
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
Gene Aliases
PHLPPL, PPM3B
UniGene
Hs.709458
Gene Chromosome Location
Chr.16: 71644924-71724701 on Build GRCh38
Transcripts
NM_001289003.1 NM_015020.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1609630 c.2928A>C p.Q976H Chr.16: 71649934-71649934 on Build GRCh38
1663485 c.2942T>A p.I981N Chr.16: 71649920-71649920 on Build GRCh38
1519652 c.2914C>T p.P972S Chr.16: 71649948-71649948 on Build GRCh38
1479069 c.3200_3201insC p.A1068fs*6 Chr.16: 71649661-71649662 on Build GRCh38
5342843 c.2921C>T p.T974I Chr.16: 71649941-71649941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368274198 Chr.16: 71649927-71649927 on Build GRCh38
rs150749372 Chr.16: 71649839-71649839 on Build GRCh38
rs746973319 Chr.16: 71649791-71649791 on Build GRCh38
rs968278880 Chr.16: 71649648-71649649 on Build GRCh38
rs773669194 Chr.16: 71649661-71649662 on Build GRCh38
rs779777756 Chr.16: 71649747-71649747 on Build GRCh38
rs764231210 Chr.16: 71649715-71649715 on Build GRCh38
rs771226206 Chr.16: 71650077-71650080 on Build GRCh38
rs138193971 Chr.16: 71649647-71649647 on Build GRCh38
rs748209892 Chr.16: 71650092-71650092 on Build GRCh38
rs542079837 Chr.16: 71650100-71650100 on Build GRCh38
rs746749301 Chr.16: 71649858-71649858 on Build GRCh38
rs966582380 Chr.16: 71649829-71649829 on Build GRCh38
rs775549369 Chr.16: 71649799-71649799 on Build GRCh38
rs771798945 Chr.16: 71649712-71649712 on Build GRCh38
rs577401843 Chr.16: 71649978-71649978 on Build GRCh38
rs994892561 Chr.16: 71649889-71649889 on Build GRCh38
rs751946290 Chr.16: 71649717-71649717 on Build GRCh38
rs143977373 Chr.16: 71649855-71649855 on Build GRCh38
rs778863666 Chr.16: 71650008-71650008 on Build GRCh38
rs750151844 Chr.16: 71650073-71650073 on Build GRCh38
rs139007505 Chr.16: 71649840-71649840 on Build GRCh38
rs577952264 Chr.16: 71650088-71650088 on Build GRCh38
rs770308351 Chr.16: 71649649-71649649 on Build GRCh38
rs143949615 Chr.16: 71649960-71649960 on Build GRCh38
rs148247641 Chr.16: 71649779-71649779 on Build GRCh38
rs555796455 Chr.16: 71650085-71650085 on Build GRCh38
rs375283006 Chr.16: 71649797-71649797 on Build GRCh38
rs61733127 Chr.16: 71649815-71649815 on Build GRCh38
rs768776946 Chr.16: 71649871-71649871 on Build GRCh38
rs28619971 Chr.16: 71649837-71649837 on Build GRCh38
rs780945216 Chr.16: 71649991-71649991 on Build GRCh38
rs375263749 Chr.16: 71650059-71650059 on Build GRCh38
rs768901557 Chr.16: 71649803-71649803 on Build GRCh38
rs371852267 Chr.16: 71650006-71650006 on Build GRCh38
rs966991891 Chr.16: 71649905-71649905 on Build GRCh38
rs147192446 Chr.16: 71649778-71649778 on Build GRCh38
rs755624704 Chr.16: 71649657-71649657 on Build GRCh38
rs1045589860 Chr.16: 71649734-71649734 on Build GRCh38
rs929252693 Chr.16: 71650097-71650097 on Build GRCh38
rs182647340 Chr.16: 71650053-71650053 on Build GRCh38
rs142287063 Chr.16: 71649856-71649856 on Build GRCh38
rs770864666 Chr.16: 71649860-71649860 on Build GRCh38
rs774480827 Chr.16: 71649894-71649894 on Build GRCh38
rs374030439 Chr.16: 71649741-71649741 on Build GRCh38
rs549003726 Chr.16: 71649743-71649743 on Build GRCh38
rs374864964 Chr.16: 71649812-71649812 on Build GRCh38
rs894273700 Chr.16: 71649681-71649681 on Build GRCh38
rs763310095 Chr.16: 71649952-71649953 on Build GRCh38
rs772983033 Chr.16: 71649672-71649672 on Build GRCh38
rs748004499 Chr.16: 71649781-71649781 on Build GRCh38
rs760182767 Chr.16: 71650027-71650027 on Build GRCh38
rs765481727 Chr.16: 71649951-71649951 on Build GRCh38
rs768419363 Chr.16: 71649665-71649665 on Build GRCh38
rs758544612 Chr.16: 71650096-71650096 on Build GRCh38
rs774480161 Chr.16: 71649811-71649811 on Build GRCh38
rs765346195 Chr.16: 71649702-71649702 on Build GRCh38
rs925298558 Chr.16: 71649940-71649940 on Build GRCh38
rs779692183 Chr.16: 71649660-71649660 on Build GRCh38
rs749007521 Chr.16: 71649664-71649664 on Build GRCh38
rs747200736 Chr.16: 71650014-71650014 on Build GRCh38
rs767022708 Chr.16: 71649936-71649936 on Build GRCh38
rs751331392 Chr.16: 71649967-71649967 on Build GRCh38
rs748202550 Chr.16: 71650010-71650010 on Build GRCh38
rs762007955 Chr.16: 71649902-71649902 on Build GRCh38
rs753334495 Chr.16: 71649767-71649767 on Build GRCh38
rs140175974 Chr.16: 71649944-71649944 on Build GRCh38
rs534322297 Chr.16: 71649903-71649903 on Build GRCh38
rs771803100 Chr.16: 71649787-71649787 on Build GRCh38
rs896010584 Chr.16: 71649817-71649817 on Build GRCh38
rs377731854 Chr.16: 71650069-71650069 on Build GRCh38
rs558747968 Chr.16: 71649937-71649937 on Build GRCh38
rs918013782 Chr.16: 71649968-71649968 on Build GRCh38
rs550126046 Chr.16: 71649718-71649718 on Build GRCh38
rs771471036 Chr.16: 71650016-71650016 on Build GRCh38
rs761074301 Chr.16: 71649935-71649935 on Build GRCh38
rs760629537 Chr.16: 71649697-71649697 on Build GRCh38
rs777601856 Chr.16: 71649790-71649790 on Build GRCh38
rs756045415 Chr.16: 71650002-71650002 on Build GRCh38
rs368234798 Chr.16: 71649745-71649745 on Build GRCh38
rs753422591 Chr.16: 71649825-71649825 on Build GRCh38
rs985997309 Chr.16: 71649737-71649737 on Build GRCh38
rs757087212 Chr.16: 71649990-71649990 on Build GRCh38
rs751265135 Chr.16: 71650063-71650063 on Build GRCh38
rs764710771 Chr.16: 71649946-71649946 on Build GRCh38
rs774295206 Chr.16: 71649667-71649667 on Build GRCh38
rs187345250 Chr.16: 71649881-71649881 on Build GRCh38
rs371257120 Chr.16: 71650084-71650084 on Build GRCh38
rs772067328 Chr.16: 71649671-71649671 on Build GRCh38
rs145787660 Chr.16: 71650022-71650022 on Build GRCh38
rs949847674 Chr.16: 71650012-71650012 on Build GRCh38
rs778535377 Chr.16: 71649852-71649852 on Build GRCh38
rs757987779 Chr.16: 71649947-71649947 on Build GRCh38
rs531387188 Chr.16: 71649669-71649669 on Build GRCh38
rs772207777 Chr.16: 71650011-71650011 on Build GRCh38
rs765705432 Chr.16: 71650042-71650042 on Build GRCh38
rs750796940 Chr.16: 71649813-71649813 on Build GRCh38
rs745867331 Chr.16: 71649994-71649994 on Build GRCh38
rs775530751 Chr.16: 71649706-71649706 on Build GRCh38
rs1055612670 Chr.16: 71649674-71649674 on Build GRCh38
rs781071469 Chr.16: 71649861-71649861 on Build GRCh38
rs762959444 Chr.16: 71649800-71649800 on Build GRCh38
rs767371734 Chr.16: 71650071-71650071 on Build GRCh38
rs745777414 Chr.16: 71649867-71649867 on Build GRCh38

View Full Product Details