Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71929080-71929576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IST1
Entrez Gene ID
9798
Gene Name
IST1, ESCRT-III associated factor
Gene Aliases
OLC1
UniGene
Hs.232194
Gene Chromosome Location
Chr.16: 71894408-71930639 on Build GRCh38
Transcripts
NM_001270975.1 NM_001270976.1 NM_001270977.1 NM_001270978.1 NM_001270979.1 NM_014761.3 NM_181536.1

Gene Symbol
PKD1L3
Entrez Gene ID
342372
Gene Name
polycystin 1 like 3, transient receptor potential channel interacting
Gene Aliases
-
UniGene
Hs.675286
Gene Chromosome Location
Chr.16: 71929538-72000401 on Build GRCh38
Transcripts
NM_001270975.1 NM_001270976.1 NM_001270977.1 NM_001270978.1 NM_001270979.1 NM_014761.3 NM_181536.1

SNP Detail

SNP ID Chromosome Location
rs949488282 Chr.16: 71929473-71929473 on Build GRCh38
rs769042089 Chr.16: 71929126-71929126 on Build GRCh38
rs763793190 Chr.16: 71929466-71929466 on Build GRCh38
rs1028150138 Chr.16: 71929351-71929351 on Build GRCh38
rs988124954 Chr.16: 71929465-71929465 on Build GRCh38
rs1045293078 Chr.16: 71929221-71929221 on Build GRCh38
rs962604710 Chr.16: 71929406-71929406 on Build GRCh38
rs538631546 Chr.16: 71929494-71929494 on Build GRCh38
rs551989334 Chr.16: 71929501-71929501 on Build GRCh38
rs986207389 Chr.16: 71929206-71929206 on Build GRCh38
rs996185186 Chr.16: 71929244-71929244 on Build GRCh38
rs1024519056 Chr.16: 71929389-71929389 on Build GRCh38
rs944691002 Chr.16: 71929474-71929474 on Build GRCh38
rs952237174 Chr.16: 71929456-71929456 on Build GRCh38
rs1040939103 Chr.16: 71929232-71929232 on Build GRCh38
rs1004292004 Chr.16: 71929534-71929534 on Build GRCh38
rs910640765 Chr.16: 71929472-71929472 on Build GRCh38
rs112083024 Chr.16: 71929173-71929173 on Build GRCh38
rs1042975233 Chr.16: 71929125-71929125 on Build GRCh38
rs935347930 Chr.16: 71929450-71929450 on Build GRCh38
rs546686418 Chr.16: 71929303-71929303 on Build GRCh38
rs773562611 Chr.16: 71929282-71929282 on Build GRCh38
rs930720083 Chr.16: 71929184-71929184 on Build GRCh38
rs894111075 Chr.16: 71929365-71929365 on Build GRCh38
rs965062279 Chr.16: 71929301-71929301 on Build GRCh38
rs986155213 Chr.16: 71929457-71929457 on Build GRCh38
rs764846964 Chr.16: 71929535-71929535 on Build GRCh38
rs760064231 Chr.16: 71929526-71929526 on Build GRCh38
rs972322534 Chr.16: 71929407-71929407 on Build GRCh38
rs1034126958 Chr.16: 71929363-71929363 on Build GRCh38
rs548477342 Chr.16: 71929138-71929138 on Build GRCh38
rs866992596 Chr.16: 71929207-71929207 on Build GRCh38
rs766865783 Chr.16: 71929328-71929328 on Build GRCh38
rs1041571751 Chr.16: 71929321-71929321 on Build GRCh38
rs532242468 Chr.16: 71929416-71929416 on Build GRCh38
rs7195988 Chr.16: 71929283-71929283 on Build GRCh38
rs569251164 Chr.16: 71929537-71929537 on Build GRCh38
rs571446641 Chr.16: 71929432-71929432 on Build GRCh38
rs147352269 Chr.16: 71929502-71929502 on Build GRCh38
rs887330701 Chr.16: 71929509-71929509 on Build GRCh38
rs1014343302 Chr.16: 71929380-71929380 on Build GRCh38
rs760152973 Chr.16: 71929374-71929374 on Build GRCh38
rs866378106 Chr.16: 71929358-71929358 on Build GRCh38
rs993069193 Chr.16: 71929112-71929112 on Build GRCh38
rs944722680 Chr.16: 71929227-71929227 on Build GRCh38
rs770283292 Chr.16: 71929217-71929217 on Build GRCh38
rs569907636 Chr.16: 71929352-71929352 on Build GRCh38
rs1027765930 Chr.16: 71929419-71929419 on Build GRCh38
rs753482486 Chr.16: 71929484-71929484 on Build GRCh38
rs886790164 Chr.16: 71929266-71929266 on Build GRCh38
rs1019322705 Chr.16: 71929287-71929287 on Build GRCh38
rs972643332 Chr.16: 71929153-71929153 on Build GRCh38
rs962200821 Chr.16: 71929144-71929144 on Build GRCh38
rs901093813 Chr.16: 71929277-71929277 on Build GRCh38
rs748584204 Chr.16: 71929180-71929180 on Build GRCh38

View Full Product Details