Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75481726-75482220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST6
Entrez Gene ID
4166
Gene Name
carbohydrate sulfotransferase 6
Gene Aliases
MCDC1
UniGene
Hs.655622
Gene Chromosome Location
Chr.16: 75473124-75495419 on Build GRCh38
Transcripts
NM_021615.4 XM_005255955.4 XM_011523085.2

SNP Detail

SNP ID Chromosome Location
rs919943067 Chr.16: 75481935-75481935 on Build GRCh38
rs1001415167 Chr.16: 75481889-75481889 on Build GRCh38
rs911371186 Chr.16: 75481970-75481970 on Build GRCh38
rs190632251 Chr.16: 75481926-75481926 on Build GRCh38
rs1014035519 Chr.16: 75482123-75482123 on Build GRCh38
rs945406549 Chr.16: 75482165-75482165 on Build GRCh38
rs1017024642 Chr.16: 75481745-75481745 on Build GRCh38
rs1005731220 Chr.16: 75482043-75482043 on Build GRCh38
rs766269041 Chr.16: 75482155-75482155 on Build GRCh38
rs889968794 Chr.16: 75481827-75481827 on Build GRCh38
rs944172584 Chr.16: 75481986-75481986 on Build GRCh38
rs1043080275 Chr.16: 75482076-75482076 on Build GRCh38
rs1021328261 Chr.16: 75481867-75481867 on Build GRCh38
rs992645006 Chr.16: 75481925-75481925 on Build GRCh38
rs1007953702 Chr.16: 75481829-75481829 on Build GRCh38
rs111319091 Chr.16: 75482133-75482133 on Build GRCh38
rs1054788794 Chr.16: 75481994-75481995 on Build GRCh38
rs961721678 Chr.16: 75482159-75482159 on Build GRCh38
rs954842068 Chr.16: 75482115-75482115 on Build GRCh38
rs973142727 Chr.16: 75481934-75481934 on Build GRCh38
rs3884080 Chr.16: 75482019-75482020 on Build GRCh38
rs939998703 Chr.16: 75481780-75481780 on Build GRCh38
rs891570308 Chr.16: 75481848-75481848 on Build GRCh38
rs183203023 Chr.16: 75482040-75482040 on Build GRCh38
rs147406533 Chr.16: 75482018-75482019 on Build GRCh38
rs1018204661 Chr.16: 75482145-75482145 on Build GRCh38
rs945276761 Chr.16: 75481885-75481885 on Build GRCh38
rs534647925 Chr.16: 75481831-75481831 on Build GRCh38
rs1037064648 Chr.16: 75481784-75481784 on Build GRCh38
rs557102064 Chr.16: 75481879-75481879 on Build GRCh38
rs551337641 Chr.16: 75481833-75481833 on Build GRCh38
rs775808193 Chr.16: 75481921-75481921 on Build GRCh38
rs920164350 Chr.16: 75481790-75481790 on Build GRCh38
rs989976152 Chr.16: 75482160-75482161 on Build GRCh38
rs544249772 Chr.16: 75482150-75482150 on Build GRCh38
rs927912847 Chr.16: 75482189-75482189 on Build GRCh38
rs977959105 Chr.16: 75482176-75482176 on Build GRCh38
rs1027374837 Chr.16: 75482175-75482175 on Build GRCh38
rs34105319 Chr.16: 75482020-75482021 on Build GRCh38
rs1034211245 Chr.16: 75482101-75482101 on Build GRCh38
rs1040835650 Chr.16: 75481838-75481838 on Build GRCh38
rs904201418 Chr.16: 75482079-75482079 on Build GRCh38
rs777308766 Chr.16: 75481757-75481757 on Build GRCh38
rs537330126 Chr.16: 75481847-75481847 on Build GRCh38
rs532378441 Chr.16: 75481949-75481949 on Build GRCh38
rs746169691 Chr.16: 75481801-75481801 on Build GRCh38
rs771460173 Chr.16: 75481774-75481774 on Build GRCh38
rs1049885647 Chr.16: 75481821-75481821 on Build GRCh38
rs896821890 Chr.16: 75482055-75482055 on Build GRCh38
rs961519214 Chr.16: 75481932-75481932 on Build GRCh38
rs985724161 Chr.16: 75481960-75481960 on Build GRCh38
rs945816334 Chr.16: 75482049-75482049 on Build GRCh38
rs371427598 Chr.16: 75481863-75481863 on Build GRCh38
rs187929339 Chr.16: 75482044-75482044 on Build GRCh38
rs886052322 Chr.16: 75481821-75481821 on Build GRCh38
rs575557449 Chr.16: 75482111-75482111 on Build GRCh38
rs774593319 Chr.16: 75481797-75481797 on Build GRCh38
rs1001382535 Chr.16: 75482099-75482099 on Build GRCh38
rs536853758 Chr.16: 75482010-75482010 on Build GRCh38
rs762811065 Chr.16: 75482153-75482153 on Build GRCh38
rs1014799078 Chr.16: 75482138-75482138 on Build GRCh38
rs963640269 Chr.16: 75481753-75481753 on Build GRCh38
rs993743227 Chr.16: 75482083-75482083 on Build GRCh38
rs1034116607 Chr.16: 75481909-75481909 on Build GRCh38
rs1026451374 Chr.16: 75481789-75481789 on Build GRCh38
rs553892780 Chr.16: 75482052-75482052 on Build GRCh38
rs545422669 Chr.16: 75482051-75482051 on Build GRCh38
rs114012239 Chr.16: 75481878-75481878 on Build GRCh38
rs561066093 Chr.16: 75482160-75482160 on Build GRCh38
rs749832523 Chr.16: 75481771-75481771 on Build GRCh38
rs191088770 Chr.16: 75482054-75482054 on Build GRCh38
rs886052323 Chr.16: 75481866-75481866 on Build GRCh38
rs987551960 Chr.16: 75481796-75481796 on Build GRCh38

View Full Product Details