Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75540978-75541500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM231
Entrez Gene ID
79583
Gene Name
transmembrane protein 231
Gene Aliases
ALYE870, JBTS20, MKS11, PRO1886
UniGene
Hs.156784
Gene Chromosome Location
Chr.16: 75538117-75556286 on Build GRCh38
Transcripts
NM_001077416.2 NM_001077418.2 NR_074083.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5971931 c.800C>T p.A267V Chr.16: 75541407-75541407 on Build GRCh38
4579275 c.797G>C p.R266T Chr.16: 75541410-75541410 on Build GRCh38
5893889 c.842C>T p.P281L Chr.16: 75541365-75541365 on Build GRCh38
4062858 c.790G>A p.V264M Chr.16: 75541417-75541417 on Build GRCh38
5896281 c.772C>T p.P258S Chr.16: 75541435-75541435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780481430 Chr.16: 75541042-75541042 on Build GRCh38
rs1007468725 Chr.16: 75541237-75541237 on Build GRCh38
rs996376809 Chr.16: 75541173-75541173 on Build GRCh38
rs201237881 Chr.16: 75541465-75541465 on Build GRCh38
rs1020757759 Chr.16: 75541361-75541361 on Build GRCh38
rs747920221 Chr.16: 75541433-75541433 on Build GRCh38
rs551336750 Chr.16: 75541230-75541230 on Build GRCh38
rs1004834188 Chr.16: 75541110-75541110 on Build GRCh38
rs888818002 Chr.16: 75541223-75541223 on Build GRCh38
rs763365146 Chr.16: 75541306-75541306 on Build GRCh38
rs918979596 Chr.16: 75541178-75541178 on Build GRCh38
rs886039807 Chr.16: 75541466-75541466 on Build GRCh38
rs369010440 Chr.16: 75541371-75541371 on Build GRCh38
rs781284313 Chr.16: 75541107-75541107 on Build GRCh38
rs967746060 Chr.16: 75541005-75541006 on Build GRCh38
rs746055228 Chr.16: 75541447-75541447 on Build GRCh38
rs989041053 Chr.16: 75541363-75541363 on Build GRCh38
rs1000562089 Chr.16: 75541186-75541187 on Build GRCh38
rs369180727 Chr.16: 75541057-75541057 on Build GRCh38
rs978704864 Chr.16: 75541083-75541083 on Build GRCh38
rs753867687 Chr.16: 75541415-75541415 on Build GRCh38
rs63219162 Chr.16: 75541181-75541181 on Build GRCh38
rs769897831 Chr.16: 75541448-75541448 on Build GRCh38
rs146398551 Chr.16: 75541010-75541010 on Build GRCh38
rs758122684 Chr.16: 75541440-75541440 on Build GRCh38
rs746025189 Chr.16: 75541393-75541393 on Build GRCh38
rs771853785 Chr.16: 75541455-75541455 on Build GRCh38
rs1049132130 Chr.16: 75541101-75541101 on Build GRCh38
rs771137248 Chr.16: 75541378-75541378 on Build GRCh38
rs930407134 Chr.16: 75541187-75541187 on Build GRCh38
rs923606211 Chr.16: 75541039-75541039 on Build GRCh38
rs901687287 Chr.16: 75541163-75541163 on Build GRCh38
rs971651334 Chr.16: 75541452-75541452 on Build GRCh38
rs751248751 Chr.16: 75541317-75541317 on Build GRCh38
rs766954527 Chr.16: 75541323-75541323 on Build GRCh38
rs886434544 Chr.16: 75541106-75541106 on Build GRCh38
rs1029129159 Chr.16: 75541005-75541005 on Build GRCh38
rs58545518 Chr.16: 75541198-75541199 on Build GRCh38
rs776577271 Chr.16: 75541384-75541384 on Build GRCh38
rs533530608 Chr.16: 75541181-75541181 on Build GRCh38
rs958968993 Chr.16: 75541105-75541105 on Build GRCh38
rs1040668371 Chr.16: 75541136-75541136 on Build GRCh38
rs188282170 Chr.16: 75541012-75541012 on Build GRCh38
rs139657175 Chr.16: 75541056-75541056 on Build GRCh38
rs199813223 Chr.16: 75541410-75541410 on Build GRCh38
rs143003749 Chr.16: 75541373-75541373 on Build GRCh38
rs756891705 Chr.16: 75541318-75541318 on Build GRCh38
rs78753303 Chr.16: 75541179-75541179 on Build GRCh38
rs139236786 Chr.16: 75541405-75541405 on Build GRCh38
rs903638635 Chr.16: 75541181-75541187 on Build GRCh38
rs754926460 Chr.16: 75541417-75541417 on Build GRCh38
rs749893185 Chr.16: 75541326-75541326 on Build GRCh38
rs765968316 Chr.16: 75541414-75541414 on Build GRCh38
rs917418692 Chr.16: 75541133-75541133 on Build GRCh38
rs930634683 Chr.16: 75541085-75541085 on Build GRCh38
rs763415666 Chr.16: 75541400-75541400 on Build GRCh38
rs200581224 Chr.16: 75541474-75541474 on Build GRCh38
rs764417023 Chr.16: 75541308-75541308 on Build GRCh38
rs776336619 Chr.16: 75541180-75541181 on Build GRCh38
rs909239818 Chr.16: 75541166-75541166 on Build GRCh38
rs750043051 Chr.16: 75541406-75541406 on Build GRCh38
rs775972268 Chr.16: 75541302-75541302 on Build GRCh38
rs747228664 Chr.16: 75541377-75541377 on Build GRCh38
rs1028744067 Chr.16: 75541177-75541177 on Build GRCh38
rs778875107 Chr.16: 75541424-75541424 on Build GRCh38
rs950155038 Chr.16: 75541147-75541147 on Build GRCh38
rs1024949621 Chr.16: 75541374-75541374 on Build GRCh38
rs528641330 Chr.16: 75541055-75541055 on Build GRCh38
rs184604234 Chr.16: 75541242-75541242 on Build GRCh38
rs925977893 Chr.16: 75541090-75541090 on Build GRCh38
rs192828186 Chr.16: 75541131-75541131 on Build GRCh38
rs776025526 Chr.16: 75541396-75541396 on Build GRCh38
rs530120683 Chr.16: 75541256-75541256 on Build GRCh38
rs369627585 Chr.16: 75541402-75541402 on Build GRCh38
rs769850804 Chr.16: 75541395-75541395 on Build GRCh38
rs757260161 Chr.16: 75541349-75541349 on Build GRCh38
rs967455443 Chr.16: 75541076-75541076 on Build GRCh38
rs116304500 Chr.16: 75541046-75541046 on Build GRCh38
rs917448087 Chr.16: 75541050-75541050 on Build GRCh38
rs992192698 Chr.16: 75541121-75541121 on Build GRCh38
rs780063686 Chr.16: 75541458-75541458 on Build GRCh38
rs1048704527 Chr.16: 75541221-75541221 on Build GRCh38
rs749217269 Chr.16: 75541283-75541283 on Build GRCh38
rs1036548824 Chr.16: 75541175-75541175 on Build GRCh38
rs781532766 Chr.16: 75541443-75541443 on Build GRCh38
rs756011907 Chr.16: 75541340-75541340 on Build GRCh38
rs901771300 Chr.16: 75541284-75541284 on Build GRCh38
rs146210288 Chr.16: 75541401-75541401 on Build GRCh38

View Full Product Details