Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 77195453-77195952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MON1B
Entrez Gene ID
22879
Gene Name
MON1 homolog B, secretory trafficking associated
Gene Aliases
HSRG1, SAND2, SRG1
UniGene
Hs.513743
Gene Chromosome Location
Chr.16: 77190919-77199646 on Build GRCh38
Transcripts
NM_001286639.1 NM_001286640.1 NM_014940.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5015409 c.1312C>T p.P438S Chr.16: 77195551-77195551 on Build GRCh38
6714225 c.1414G>A p.V472M Chr.16: 77195653-77195653 on Build GRCh38
7124335 c.1319G>A p.S440N Chr.16: 77195558-77195558 on Build GRCh38
5519406 c.1373G>A p.R458H Chr.16: 77195612-77195612 on Build GRCh38
6190362 c.1299_1304delTGAGCT p.L435_E436delLE Chr.16: 77195538-77195543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535689958 Chr.16: 77195599-77195599 on Build GRCh38
rs769027511 Chr.16: 77195585-77195585 on Build GRCh38
rs983919029 Chr.16: 77195882-77195882 on Build GRCh38
rs35595738 Chr.16: 77195898-77195899 on Build GRCh38
rs555921616 Chr.16: 77195629-77195629 on Build GRCh38
rs754849470 Chr.16: 77195553-77195553 on Build GRCh38
rs748557982 Chr.16: 77195671-77195671 on Build GRCh38
rs75533076 Chr.16: 77195890-77195890 on Build GRCh38
rs745968801 Chr.16: 77195573-77195573 on Build GRCh38
rs952235757 Chr.16: 77195636-77195636 on Build GRCh38
rs1004152603 Chr.16: 77195746-77195746 on Build GRCh38
rs967254149 Chr.16: 77195855-77195855 on Build GRCh38
rs199600735 Chr.16: 77195598-77195598 on Build GRCh38
rs939825681 Chr.16: 77195895-77195895 on Build GRCh38
rs755639920 Chr.16: 77195538-77195538 on Build GRCh38
rs776212026 Chr.16: 77195703-77195703 on Build GRCh38
rs780235256 Chr.16: 77195643-77195643 on Build GRCh38
rs753950747 Chr.16: 77195536-77195536 on Build GRCh38
rs182520517 Chr.16: 77195867-77195867 on Build GRCh38
rs767304693 Chr.16: 77195592-77195592 on Build GRCh38
rs74578006 Chr.16: 77195594-77195594 on Build GRCh38
rs906218827 Chr.16: 77195762-77195762 on Build GRCh38
rs769222989 Chr.16: 77195654-77195654 on Build GRCh38
rs1019565 Chr.16: 77195862-77195862 on Build GRCh38
rs756424459 Chr.16: 77195729-77195729 on Build GRCh38
rs746124611 Chr.16: 77195638-77195638 on Build GRCh38
rs572676092 Chr.16: 77195639-77195639 on Build GRCh38
rs539419806 Chr.16: 77195549-77195549 on Build GRCh38
rs937700161 Chr.16: 77195927-77195927 on Build GRCh38
rs763254600 Chr.16: 77195486-77195486 on Build GRCh38
rs1024267863 Chr.16: 77195600-77195600 on Build GRCh38
rs747931127 Chr.16: 77195560-77195560 on Build GRCh38
rs769820253 Chr.16: 77195575-77195575 on Build GRCh38
rs774554424 Chr.16: 77195516-77195516 on Build GRCh38
rs781332889 Chr.16: 77195572-77195572 on Build GRCh38
rs201465107 Chr.16: 77195578-77195578 on Build GRCh38
rs374484182 Chr.16: 77195492-77195492 on Build GRCh38
rs551675960 Chr.16: 77195880-77195880 on Build GRCh38
rs190219071 Chr.16: 77195795-77195795 on Build GRCh38
rs563414415 Chr.16: 77195861-77195861 on Build GRCh38
rs774785037 Chr.16: 77195586-77195586 on Build GRCh38
rs770488807 Chr.16: 77195702-77195702 on Build GRCh38
rs570840558 Chr.16: 77195480-77195480 on Build GRCh38
rs1014171177 Chr.16: 77195579-77195579 on Build GRCh38
rs143886376 Chr.16: 77195825-77195825 on Build GRCh38
rs752500795 Chr.16: 77195619-77195619 on Build GRCh38
rs914161600 Chr.16: 77195604-77195604 on Build GRCh38
rs374239495 Chr.16: 77195693-77195693 on Build GRCh38
rs767214918 Chr.16: 77195528-77195528 on Build GRCh38
rs543404100 Chr.16: 77195833-77195833 on Build GRCh38
rs765015648 Chr.16: 77195612-77195612 on Build GRCh38
rs966389964 Chr.16: 77195908-77195908 on Build GRCh38
rs762214589 Chr.16: 77195590-77195590 on Build GRCh38
rs753276223 Chr.16: 77195602-77195602 on Build GRCh38
rs368413301 Chr.16: 77195725-77195725 on Build GRCh38
rs778827471 Chr.16: 77195559-77195559 on Build GRCh38
rs975846768 Chr.16: 77195898-77195898 on Build GRCh38
rs776016012 Chr.16: 77195485-77195485 on Build GRCh38
rs765239007 Chr.16: 77195719-77195719 on Build GRCh38
rs772609925 Chr.16: 77195674-77195674 on Build GRCh38
rs758910465 Chr.16: 77195704-77195704 on Build GRCh38
rs1028348590 Chr.16: 77195631-77195631 on Build GRCh38
rs1021133600 Chr.16: 77195847-77195847 on Build GRCh38
rs749321723 Chr.16: 77195653-77195653 on Build GRCh38
rs71394301 Chr.16: 77195537-77195537 on Build GRCh38
rs371113497 Chr.16: 77195488-77195488 on Build GRCh38
rs531731361 Chr.16: 77195866-77195866 on Build GRCh38
rs757321096 Chr.16: 77195548-77195549 on Build GRCh38
rs142851781 Chr.16: 77195582-77195583 on Build GRCh38
rs762818609 Chr.16: 77195817-77195817 on Build GRCh38
rs754576869 Chr.16: 77195516-77195519 on Build GRCh38
rs779456878 Chr.16: 77195739-77195739 on Build GRCh38
rs60108557 Chr.16: 77195870-77195870 on Build GRCh38
rs368499093 Chr.16: 77195581-77195581 on Build GRCh38
rs374570202 Chr.16: 77195502-77195502 on Build GRCh38
rs774953462 Chr.16: 77195665-77195665 on Build GRCh38
rs781423019 Chr.16: 77195630-77195630 on Build GRCh38
rs74027174 Chr.16: 77195906-77195906 on Build GRCh38
rs1055369869 Chr.16: 77195775-77195775 on Build GRCh38
rs761613792 Chr.16: 77195527-77195527 on Build GRCh38
rs12599030 Chr.16: 77195879-77195879 on Build GRCh38
rs757473363 Chr.16: 77195565-77195565 on Build GRCh38
rs1048213043 Chr.16: 77195731-77195731 on Build GRCh38
rs370803909 Chr.16: 77195611-77195611 on Build GRCh38
rs62049563 Chr.16: 77195727-77195727 on Build GRCh38
rs773690280 Chr.16: 77195687-77195687 on Build GRCh38
rs1039143949 Chr.16: 77195718-77195718 on Build GRCh38
rs765963556 Chr.16: 77195601-77195601 on Build GRCh38
rs374361534 Chr.16: 77195529-77195529 on Build GRCh38
rs2303259 Chr.16: 77195760-77195760 on Build GRCh38
rs751340853 Chr.16: 77195728-77195728 on Build GRCh38
rs752586684 Chr.16: 77195724-77195724 on Build GRCh38
rs2017197 Chr.16: 77195772-77195772 on Build GRCh38
rs1014305515 Chr.16: 77195834-77195834 on Build GRCh38
rs906459646 Chr.16: 77195919-77195919 on Build GRCh38
rs957791998 Chr.16: 77195820-77195820 on Build GRCh38

View Full Product Details