Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84012733-84013270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC38A8
Entrez Gene ID
146167
Gene Name
solute carrier family 38 member 8
Gene Aliases
FVH2
UniGene
Gene Chromosome Location
Chr.16: 84009667-84043372 on Build GRCh38
Transcripts
NM_001080442.2 XM_017022946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
974312 c.1194G>T p.E398D Chr.16: 84013021-84013021 on Build GRCh38
974314 c.1166T>C p.L389S Chr.16: 84013049-84013049 on Build GRCh38
6782027 c.1163-5delT p.? Chr.16: 84013057-84013057 on Build GRCh38
974313 c.1192G>A p.E398K Chr.16: 84013023-84013023 on Build GRCh38
3691217 c.1191C>T p.V397V Chr.16: 84013024-84013024 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs140910258 Chr.16: 84013081-84013081 on Build GRCh38
rs370482252 Chr.16: 84012959-84012959 on Build GRCh38
rs767040255 Chr.16: 84013022-84013022 on Build GRCh38
rs140783318 Chr.16: 84013023-84013023 on Build GRCh38
rs111375709 Chr.16: 84012785-84012785 on Build GRCh38
rs903298698 Chr.16: 84013054-84013054 on Build GRCh38
rs568953778 Chr.16: 84013082-84013082 on Build GRCh38
rs79659184 Chr.16: 84013062-84013062 on Build GRCh38
rs200490244 Chr.16: 84012962-84012962 on Build GRCh38
rs11864211 Chr.16: 84013157-84013157 on Build GRCh38
rs984590706 Chr.16: 84013159-84013159 on Build GRCh38
rs771227689 Chr.16: 84013005-84013005 on Build GRCh38
rs562707340 Chr.16: 84013175-84013175 on Build GRCh38
rs931745349 Chr.16: 84012847-84012847 on Build GRCh38
rs974488018 Chr.16: 84012882-84012882 on Build GRCh38
rs984928821 Chr.16: 84013030-84013030 on Build GRCh38
rs183629554 Chr.16: 84013197-84013197 on Build GRCh38
rs13335951 Chr.16: 84013248-84013248 on Build GRCh38
rs368391947 Chr.16: 84012972-84012972 on Build GRCh38
rs948354315 Chr.16: 84012928-84012928 on Build GRCh38
rs375620631 Chr.16: 84013007-84013007 on Build GRCh38
rs564832832 Chr.16: 84012764-84012764 on Build GRCh38
rs144724564 Chr.16: 84013133-84013133 on Build GRCh38
rs531844037 Chr.16: 84013189-84013189 on Build GRCh38
rs569246606 Chr.16: 84012805-84012805 on Build GRCh38
rs749095541 Chr.16: 84013042-84013042 on Build GRCh38
rs201194139 Chr.16: 84013010-84013010 on Build GRCh38
rs935850857 Chr.16: 84012887-84012887 on Build GRCh38
rs34004368 Chr.16: 84013152-84013153 on Build GRCh38
rs747043282 Chr.16: 84013031-84013031 on Build GRCh38
rs752108516 Chr.16: 84013101-84013101 on Build GRCh38
rs763652400 Chr.16: 84012974-84012974 on Build GRCh38
rs764024391 Chr.16: 84013020-84013020 on Build GRCh38
rs1045286889 Chr.16: 84012936-84012936 on Build GRCh38
rs773172752 Chr.16: 84012766-84012766 on Build GRCh38
rs769472836 Chr.16: 84013036-84013036 on Build GRCh38
rs74032385 Chr.16: 84013152-84013152 on Build GRCh38
rs999299344 Chr.16: 84012921-84012921 on Build GRCh38
rs13336030 Chr.16: 84013164-84013164 on Build GRCh38
rs11864162 Chr.16: 84012885-84012885 on Build GRCh38
rs751209781 Chr.16: 84012976-84012976 on Build GRCh38
rs778256736 Chr.16: 84013142-84013142 on Build GRCh38
rs199825635 Chr.16: 84012992-84012992 on Build GRCh38
rs561107010 Chr.16: 84012968-84012968 on Build GRCh38
rs138662310 Chr.16: 84013208-84013208 on Build GRCh38
rs1024818233 Chr.16: 84012915-84012915 on Build GRCh38
rs920315397 Chr.16: 84013083-84013083 on Build GRCh38
rs377566753 Chr.16: 84012949-84012949 on Build GRCh38
rs751854628 Chr.16: 84012853-84012853 on Build GRCh38
rs11864095 Chr.16: 84012894-84012894 on Build GRCh38
rs928123886 Chr.16: 84013181-84013181 on Build GRCh38
rs772270380 Chr.16: 84012996-84012996 on Build GRCh38
rs560766320 Chr.16: 84012951-84012951 on Build GRCh38
rs373981480 Chr.16: 84012958-84012958 on Build GRCh38
rs1031036693 Chr.16: 84012842-84012842 on Build GRCh38
rs940360240 Chr.16: 84012866-84012866 on Build GRCh38
rs551460024 Chr.16: 84013044-84013044 on Build GRCh38
rs910089872 Chr.16: 84013033-84013033 on Build GRCh38
rs756407374 Chr.16: 84012990-84012990 on Build GRCh38
rs888964877 Chr.16: 84013117-84013117 on Build GRCh38
rs542526554 Chr.16: 84012878-84012878 on Build GRCh38
rs934464384 Chr.16: 84013126-84013126 on Build GRCh38
rs879227849 Chr.16: 84013057-84013057 on Build GRCh38
rs977414128 Chr.16: 84012820-84012820 on Build GRCh38
rs563027385 Chr.16: 84012960-84012960 on Build GRCh38
rs537507964 Chr.16: 84013085-84013085 on Build GRCh38
rs993452101 Chr.16: 84013224-84013224 on Build GRCh38
rs948215115 Chr.16: 84013050-84013050 on Build GRCh38
rs367869251 Chr.16: 84013067-84013067 on Build GRCh38
rs765695393 Chr.16: 84013024-84013024 on Build GRCh38
rs745941398 Chr.16: 84013073-84013073 on Build GRCh38
rs775199543 Chr.16: 84013037-84013037 on Build GRCh38
rs560480773 Chr.16: 84012789-84012789 on Build GRCh38
rs555478976 Chr.16: 84012895-84012895 on Build GRCh38
rs75919326 Chr.16: 84013241-84013241 on Build GRCh38
rs11864146 Chr.16: 84013110-84013110 on Build GRCh38
rs989305705 Chr.16: 84012854-84012854 on Build GRCh38
rs867840710 Chr.16: 84013153-84013153 on Build GRCh38
rs11149615 Chr.16: 84012807-84012807 on Build GRCh38
rs765043988 Chr.16: 84012963-84012963 on Build GRCh38
rs756247928 Chr.16: 84012840-84012840 on Build GRCh38
rs760404306 Chr.16: 84013056-84013057 on Build GRCh38
rs767108917 Chr.16: 84013061-84013061 on Build GRCh38
rs911619049 Chr.16: 84012865-84012865 on Build GRCh38
rs777931373 Chr.16: 84012997-84012997 on Build GRCh38
rs376582553 Chr.16: 84013064-84013064 on Build GRCh38
rs546371698 Chr.16: 84013210-84013210 on Build GRCh38
rs372732804 Chr.16: 84012939-84012939 on Build GRCh38
rs539350242 Chr.16: 84013013-84013013 on Build GRCh38
rs778443799 Chr.16: 84013026-84013026 on Build GRCh38
rs550481608 Chr.16: 84012818-84012818 on Build GRCh38
rs564492410 Chr.16: 84012933-84012933 on Build GRCh38
rs780278855 Chr.16: 84013075-84013075 on Build GRCh38
rs540883811 Chr.16: 84012944-84012944 on Build GRCh38
rs915156138 Chr.16: 84012872-84012872 on Build GRCh38
rs747096444 Chr.16: 84013000-84013000 on Build GRCh38
rs770575237 Chr.16: 84013086-84013086 on Build GRCh38
rs781399880 Chr.16: 84013072-84013072 on Build GRCh38
rs934521336 Chr.16: 84013218-84013218 on Build GRCh38
rs763386898 Chr.16: 84013095-84013095 on Build GRCh38
rs386793311 Chr.16: 84012805-84012807 on Build GRCh38
rs1012673217 Chr.16: 84012819-84012819 on Build GRCh38
rs1014280742 Chr.16: 84012808-84012808 on Build GRCh38
rs762428151 Chr.16: 84012973-84012973 on Build GRCh38
rs752720800 Chr.16: 84012967-84012967 on Build GRCh38
rs776461882 Chr.16: 84013090-84013090 on Build GRCh38
rs932715459 Chr.16: 84013225-84013225 on Build GRCh38
rs779499799 Chr.16: 84012953-84012953 on Build GRCh38
rs368021455 Chr.16: 84013077-84013077 on Build GRCh38
rs373404229 Chr.16: 84013078-84013078 on Build GRCh38
rs1024369868 Chr.16: 84012810-84012810 on Build GRCh38
rs533344193 Chr.16: 84013006-84013006 on Build GRCh38
rs889178841 Chr.16: 84013205-84013205 on Build GRCh38
rs544815533 Chr.16: 84012919-84012919 on Build GRCh38
rs891110517 Chr.16: 84013244-84013244 on Build GRCh38
rs770843695 Chr.16: 84013191-84013191 on Build GRCh38
rs775199077 Chr.16: 84012913-84012913 on Build GRCh38
rs775276596 Chr.16: 84013011-84013011 on Build GRCh38
rs987646028 Chr.16: 84012857-84012857 on Build GRCh38
rs948242772 Chr.16: 84013171-84013171 on Build GRCh38
rs549547902 Chr.16: 84012804-84012804 on Build GRCh38
rs768533885 Chr.16: 84013043-84013043 on Build GRCh38
rs147996147 Chr.16: 84012772-84012772 on Build GRCh38
rs372130068 Chr.16: 84013068-84013068 on Build GRCh38
rs921417378 Chr.16: 84012845-84012845 on Build GRCh38
rs888678625 Chr.16: 84012806-84012806 on Build GRCh38
rs991149067 Chr.16: 84013200-84013200 on Build GRCh38
rs949986979 Chr.16: 84013116-84013116 on Build GRCh38
rs746158836 Chr.16: 84013034-84013034 on Build GRCh38
rs76584708 Chr.16: 84013211-84013211 on Build GRCh38
rs1043822834 Chr.16: 84013204-84013204 on Build GRCh38
rs754105161 Chr.16: 84012986-84012986 on Build GRCh38
rs543610535 Chr.16: 84012817-84012817 on Build GRCh38
rs1003621343 Chr.16: 84012828-84012828 on Build GRCh38
rs776227695 Chr.16: 84012961-84012961 on Build GRCh38
rs750804604 Chr.16: 84012985-84012985 on Build GRCh38
rs11864037 Chr.16: 84012827-84012827 on Build GRCh38
rs971960547 Chr.16: 84013147-84013147 on Build GRCh38
rs549558425 Chr.16: 84013121-84013121 on Build GRCh38
rs748140565 Chr.16: 84012955-84012955 on Build GRCh38
rs944464881 Chr.16: 84012771-84012771 on Build GRCh38
rs11864124 Chr.16: 84012797-84012797 on Build GRCh38
rs756838232 Chr.16: 84012982-84012982 on Build GRCh38
rs79490100 Chr.16: 84013063-84013063 on Build GRCh38
rs142352848 Chr.16: 84013032-84013032 on Build GRCh38
rs756303937 Chr.16: 84013074-84013074 on Build GRCh38
rs575230279 Chr.16: 84012904-84012904 on Build GRCh38
rs939630089 Chr.16: 84013182-84013182 on Build GRCh38
rs764064787 Chr.16: 84012834-84012834 on Build GRCh38
rs377031283 Chr.16: 84013069-84013069 on Build GRCh38
rs994311376 Chr.16: 84012795-84012795 on Build GRCh38
rs1026640437 Chr.16: 84013237-84013237 on Build GRCh38
rs570746289 Chr.16: 84012824-84012824 on Build GRCh38
rs1050066471 Chr.16: 84012763-84012763 on Build GRCh38
rs953350582 Chr.16: 84012873-84012873 on Build GRCh38
rs1048524290 Chr.16: 84012905-84012905 on Build GRCh38
rs753059428 Chr.16: 84013065-84013065 on Build GRCh38
rs565375069 Chr.16: 84013201-84013201 on Build GRCh38
rs371927266 Chr.16: 84013055-84013055 on Build GRCh38
rs147305708 Chr.16: 84013021-84013021 on Build GRCh38
rs901657234 Chr.16: 84013217-84013217 on Build GRCh38

View Full Product Details