Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84063184-84063680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBTPS1
Entrez Gene ID
8720
Gene Name
membrane bound transcription factor peptidase, site 1
Gene Aliases
PCSK8, S1P, SKI-1
UniGene
Hs.75890
Gene Chromosome Location
Chr.16: 84053761-84116943 on Build GRCh38
Transcripts
NM_003791.3 XM_017023816.1 XM_017023817.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4989509 c.2470A>G p.N824D Chr.16: 84063407-84063407 on Build GRCh38
4063241 c.2525T>C p.V842A Chr.16: 84063352-84063352 on Build GRCh38
5534345 c.2500C>T p.P834S Chr.16: 84063377-84063377 on Build GRCh38
272157 c.2476C>T p.P826S Chr.16: 84063401-84063401 on Build GRCh38
974325 c.2448G>A p.K816K Chr.16: 84063429-84063429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898073325 Chr.16: 84063395-84063395 on Build GRCh38
rs763878760 Chr.16: 84063404-84063404 on Build GRCh38
rs754910212 Chr.16: 84063437-84063437 on Build GRCh38
rs758961781 Chr.16: 84063451-84063451 on Build GRCh38
rs541813989 Chr.16: 84063257-84063257 on Build GRCh38
rs75546706 Chr.16: 84063469-84063469 on Build GRCh38
rs372976515 Chr.16: 84063301-84063301 on Build GRCh38
rs191659420 Chr.16: 84063534-84063534 on Build GRCh38
rs776951947 Chr.16: 84063570-84063570 on Build GRCh38
rs772622114 Chr.16: 84063334-84063334 on Build GRCh38
rs996821573 Chr.16: 84063649-84063649 on Build GRCh38
rs540173110 Chr.16: 84063208-84063208 on Build GRCh38
rs138967616 Chr.16: 84063397-84063397 on Build GRCh38
rs374397087 Chr.16: 84063225-84063225 on Build GRCh38
rs370380325 Chr.16: 84063399-84063399 on Build GRCh38
rs376198517 Chr.16: 84063490-84063490 on Build GRCh38
rs759742244 Chr.16: 84063389-84063389 on Build GRCh38
rs781122328 Chr.16: 84063461-84063461 on Build GRCh38
rs747921323 Chr.16: 84063497-84063498 on Build GRCh38
rs567703063 Chr.16: 84063538-84063538 on Build GRCh38
rs199864699 Chr.16: 84063273-84063273 on Build GRCh38
rs750092763 Chr.16: 84063347-84063347 on Build GRCh38
rs35417190 Chr.16: 84063416-84063416 on Build GRCh38
rs761471353 Chr.16: 84063411-84063411 on Build GRCh38
rs774209924 Chr.16: 84063482-84063482 on Build GRCh38
rs916911082 Chr.16: 84063598-84063598 on Build GRCh38
rs370199960 Chr.16: 84063569-84063569 on Build GRCh38
rs914891643 Chr.16: 84063523-84063523 on Build GRCh38
rs987521295 Chr.16: 84063521-84063521 on Build GRCh38
rs1028946543 Chr.16: 84063499-84063499 on Build GRCh38
rs752959333 Chr.16: 84063444-84063444 on Build GRCh38
rs894095043 Chr.16: 84063636-84063636 on Build GRCh38
rs778078965 Chr.16: 84063452-84063452 on Build GRCh38
rs907703579 Chr.16: 84063229-84063229 on Build GRCh38
rs781227786 Chr.16: 84063467-84063467 on Build GRCh38
rs369701756 Chr.16: 84063276-84063276 on Build GRCh38
rs573201668 Chr.16: 84063251-84063251 on Build GRCh38
rs776428151 Chr.16: 84063281-84063281 on Build GRCh38
rs200761583 Chr.16: 84063293-84063293 on Build GRCh38
rs772240932 Chr.16: 84063268-84063268 on Build GRCh38
rs987303959 Chr.16: 84063219-84063219 on Build GRCh38
rs749915110 Chr.16: 84063497-84063497 on Build GRCh38
rs117609228 Chr.16: 84063616-84063616 on Build GRCh38
rs3215186 Chr.16: 84063528-84063531 on Build GRCh38
rs1027930162 Chr.16: 84063618-84063618 on Build GRCh38
rs200166892 Chr.16: 84063470-84063470 on Build GRCh38
rs369928148 Chr.16: 84063300-84063300 on Build GRCh38
rs747506713 Chr.16: 84063386-84063386 on Build GRCh38
rs763690401 Chr.16: 84063318-84063318 on Build GRCh38
rs755449425 Chr.16: 84063353-84063353 on Build GRCh38
rs898026252 Chr.16: 84063609-84063609 on Build GRCh38
rs762587906 Chr.16: 84063474-84063474 on Build GRCh38
rs768489393 Chr.16: 84063475-84063475 on Build GRCh38
rs544699754 Chr.16: 84063345-84063345 on Build GRCh38
rs991211487 Chr.16: 84063574-84063574 on Build GRCh38
rs888880297 Chr.16: 84063535-84063535 on Build GRCh38
rs954825567 Chr.16: 84063505-84063505 on Build GRCh38
rs954773589 Chr.16: 84063207-84063207 on Build GRCh38
rs775061695 Chr.16: 84063264-84063264 on Build GRCh38
rs751083921 Chr.16: 84063405-84063405 on Build GRCh38
rs762968189 Chr.16: 84063400-84063400 on Build GRCh38
rs927871196 Chr.16: 84063484-84063484 on Build GRCh38
rs776655277 Chr.16: 84063549-84063550 on Build GRCh38
rs372134252 Chr.16: 84063272-84063272 on Build GRCh38
rs145091497 Chr.16: 84063380-84063380 on Build GRCh38
rs758247595 Chr.16: 84063313-84063313 on Build GRCh38
rs887751313 Chr.16: 84063625-84063625 on Build GRCh38
rs889277286 Chr.16: 84063350-84063350 on Build GRCh38
rs940491978 Chr.16: 84063230-84063230 on Build GRCh38
rs563500548 Chr.16: 84063362-84063362 on Build GRCh38
rs973618663 Chr.16: 84063561-84063561 on Build GRCh38
rs771056877 Chr.16: 84063464-84063464 on Build GRCh38
rs920486582 Chr.16: 84063266-84063266 on Build GRCh38
rs1014920849 Chr.16: 84063522-84063522 on Build GRCh38
rs376241284 Chr.16: 84063368-84063368 on Build GRCh38
rs761816416 Chr.16: 84063487-84063487 on Build GRCh38
rs58311755 Chr.16: 84063531-84063534 on Build GRCh38
rs374428264 Chr.16: 84063291-84063291 on Build GRCh38
rs779368224 Chr.16: 84063432-84063432 on Build GRCh38
rs894034840 Chr.16: 84063476-84063476 on Build GRCh38
rs184821080 Chr.16: 84063622-84063622 on Build GRCh38
rs934974997 Chr.16: 84063237-84063237 on Build GRCh38
rs559343209 Chr.16: 84063457-84063457 on Build GRCh38
rs767118837 Chr.16: 84063413-84063413 on Build GRCh38
rs751561549 Chr.16: 84063421-84063422 on Build GRCh38
rs996214139 Chr.16: 84063488-84063488 on Build GRCh38
rs755794928 Chr.16: 84063424-84063424 on Build GRCh38
rs756984033 Chr.16: 84063341-84063341 on Build GRCh38
rs780921067 Chr.16: 84063346-84063346 on Build GRCh38
rs151124229 Chr.16: 84063306-84063306 on Build GRCh38
rs748732082 Chr.16: 84063355-84063355 on Build GRCh38
rs935541670 Chr.16: 84063621-84063621 on Build GRCh38
rs949667620 Chr.16: 84063606-84063606 on Build GRCh38
rs550259956 Chr.16: 84063611-84063611 on Build GRCh38
rs747037664 Chr.16: 84063375-84063375 on Build GRCh38
rs528092317 Chr.16: 84063525-84063525 on Build GRCh38

View Full Product Details