Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84849259-84849673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISPLD2
Entrez Gene ID
83716
Gene Name
cysteine rich secretory protein LCCL domain containing 2
Gene Aliases
CRISP11, LCRISP2
UniGene
Hs.513779
Gene Chromosome Location
Chr.16: 84819981-84909510 on Build GRCh38
Transcripts
NM_031476.3 XM_005256190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4432605 c.393G>C p.W131C Chr.16: 84849418-84849418 on Build GRCh38
4795570 c.442C>T p.P148S Chr.16: 84849467-84849467 on Build GRCh38
6402634 c.424T>C p.Y142H Chr.16: 84849449-84849449 on Build GRCh38
370469 c.365G>T p.R122L Chr.16: 84849390-84849390 on Build GRCh38
6288846 c.471C>T p.P157P Chr.16: 84849496-84849496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048112820 Chr.16: 84849301-84849301 on Build GRCh38
rs200388957 Chr.16: 84849544-84849544 on Build GRCh38
rs781147155 Chr.16: 84849337-84849337 on Build GRCh38
rs773387182 Chr.16: 84849502-84849502 on Build GRCh38
rs199977606 Chr.16: 84849444-84849444 on Build GRCh38
rs71384840 Chr.16: 84849590-84849591 on Build GRCh38
rs893726673 Chr.16: 84849628-84849628 on Build GRCh38
rs749787415 Chr.16: 84849482-84849482 on Build GRCh38
rs751747915 Chr.16: 84849393-84849393 on Build GRCh38
rs4783087 Chr.16: 84849368-84849368 on Build GRCh38
rs752270042 Chr.16: 84849524-84849524 on Build GRCh38
rs4783086 Chr.16: 84849305-84849305 on Build GRCh38
rs534098980 Chr.16: 84849282-84849282 on Build GRCh38
rs140532417 Chr.16: 84849589-84849590 on Build GRCh38
rs1049709880 Chr.16: 84849625-84849625 on Build GRCh38
rs745779699 Chr.16: 84849341-84849341 on Build GRCh38
rs780473680 Chr.16: 84849471-84849471 on Build GRCh38
rs950067163 Chr.16: 84849312-84849312 on Build GRCh38
rs111755787 Chr.16: 84849591-84849592 on Build GRCh38
rs184896897 Chr.16: 84849359-84849359 on Build GRCh38
rs72528201 Chr.16: 84849592-84849593 on Build GRCh38
rs541125181 Chr.16: 84849572-84849572 on Build GRCh38
rs138518275 Chr.16: 84849391-84849391 on Build GRCh38
rs544696581 Chr.16: 84849308-84849308 on Build GRCh38
rs920651416 Chr.16: 84849283-84849283 on Build GRCh38
rs750229143 Chr.16: 84849453-84849453 on Build GRCh38
rs200626683 Chr.16: 84849504-84849504 on Build GRCh38
rs374229408 Chr.16: 84849362-84849362 on Build GRCh38
rs556628775 Chr.16: 84849306-84849306 on Build GRCh38
rs553922667 Chr.16: 84849286-84849286 on Build GRCh38
rs200222373 Chr.16: 84849503-84849503 on Build GRCh38
rs1048164869 Chr.16: 84849425-84849425 on Build GRCh38
rs774416911 Chr.16: 84849490-84849490 on Build GRCh38
rs371094053 Chr.16: 84849349-84849349 on Build GRCh38
rs145249439 Chr.16: 84849447-84849447 on Build GRCh38
rs768682092 Chr.16: 84849487-84849487 on Build GRCh38
rs746169819 Chr.16: 84849436-84849436 on Build GRCh38
rs779980635 Chr.16: 84849404-84849404 on Build GRCh38
rs756045553 Chr.16: 84849399-84849399 on Build GRCh38
rs748762552 Chr.16: 84849351-84849351 on Build GRCh38
rs759194460 Chr.16: 84849567-84849567 on Build GRCh38
rs939800149 Chr.16: 84849582-84849582 on Build GRCh38
rs202232857 Chr.16: 84849505-84849505 on Build GRCh38
rs372363505 Chr.16: 84849452-84849452 on Build GRCh38
rs755254132 Chr.16: 84849433-84849433 on Build GRCh38
rs1055777569 Chr.16: 84849289-84849289 on Build GRCh38
rs777328564 Chr.16: 84849385-84849385 on Build GRCh38
rs377296604 Chr.16: 84849458-84849458 on Build GRCh38
rs879101956 Chr.16: 84849514-84849514 on Build GRCh38
rs997133249 Chr.16: 84849314-84849314 on Build GRCh38
rs1010791620 Chr.16: 84849638-84849638 on Build GRCh38
rs146382941 Chr.16: 84849424-84849424 on Build GRCh38
rs8061351 Chr.16: 84849496-84849496 on Build GRCh38
rs374941256 Chr.16: 84849545-84849545 on Build GRCh38
rs766262174 Chr.16: 84849454-84849454 on Build GRCh38
rs773475049 Chr.16: 84849553-84849553 on Build GRCh38
rs373081281 Chr.16: 84849548-84849548 on Build GRCh38
rs949995417 Chr.16: 84849480-84849480 on Build GRCh38
rs748774112 Chr.16: 84849405-84849405 on Build GRCh38
rs760108981 Chr.16: 84849387-84849387 on Build GRCh38
rs201126463 Chr.16: 84849538-84849538 on Build GRCh38
rs775437927 Chr.16: 84849390-84849390 on Build GRCh38
rs1026929149 Chr.16: 84849557-84849557 on Build GRCh38
rs765424522 Chr.16: 84849389-84849389 on Build GRCh38
rs931082264 Chr.16: 84849295-84849295 on Build GRCh38
rs368401038 Chr.16: 84849443-84849443 on Build GRCh38
rs990657480 Chr.16: 84849588-84849589 on Build GRCh38
rs973911233 Chr.16: 84849428-84849428 on Build GRCh38
rs761818985 Chr.16: 84849492-84849492 on Build GRCh38
rs573927241 Chr.16: 84849303-84849303 on Build GRCh38
rs759515960 Chr.16: 84849512-84849512 on Build GRCh38
rs778473162 Chr.16: 84849415-84849415 on Build GRCh38
rs748283814 Chr.16: 84849531-84849531 on Build GRCh38
rs922316860 Chr.16: 84849614-84849614 on Build GRCh38
rs1041168521 Chr.16: 84849599-84849599 on Build GRCh38
rs138500867 Chr.16: 84849457-84849457 on Build GRCh38
rs142163042 Chr.16: 84849358-84849358 on Build GRCh38
rs774011086 Chr.16: 84849363-84849363 on Build GRCh38
rs908001672 Chr.16: 84849580-84849580 on Build GRCh38
rs575954621 Chr.16: 84849307-84849307 on Build GRCh38
rs915956405 Chr.16: 84849610-84849610 on Build GRCh38
rs368770898 Chr.16: 84849526-84849526 on Build GRCh38
rs571422849 Chr.16: 84849281-84849281 on Build GRCh38
rs142350120 Chr.16: 84849558-84849558 on Build GRCh38
rs541161963 Chr.16: 84849366-84849366 on Build GRCh38
rs777510550 Chr.16: 84849460-84849460 on Build GRCh38
rs181171638 Chr.16: 84849528-84849528 on Build GRCh38
rs746266269 Chr.16: 84849465-84849465 on Build GRCh38
rs995818119 Chr.16: 84849556-84849556 on Build GRCh38
rs747782464 Chr.16: 84849419-84849419 on Build GRCh38
rs1049460685 Chr.16: 84849535-84849535 on Build GRCh38
rs770164349 Chr.16: 84849466-84849466 on Build GRCh38
rs561055307 Chr.16: 84849367-84849367 on Build GRCh38
rs770225087 Chr.16: 84849441-84849441 on Build GRCh38
rs901093113 Chr.16: 84849313-84849313 on Build GRCh38
rs527475737 Chr.16: 84849494-84849494 on Build GRCh38
rs531695495 Chr.16: 84849396-84849396 on Build GRCh38
rs199841126 Chr.16: 84849537-84849537 on Build GRCh38
rs550003217 Chr.16: 84849378-84849378 on Build GRCh38
rs111313372 Chr.16: 84849641-84849641 on Build GRCh38
rs1019988698 Chr.16: 84849311-84849311 on Build GRCh38
rs764209964 Chr.16: 84849450-84849450 on Build GRCh38
rs932293406 Chr.16: 84849615-84849615 on Build GRCh38
rs111592124 Chr.16: 84849355-84849355 on Build GRCh38
rs149652155 Chr.16: 84849464-84849464 on Build GRCh38
rs765200419 Chr.16: 84849520-84849520 on Build GRCh38
rs776491731 Chr.16: 84849562-84849562 on Build GRCh38
rs753780800 Chr.16: 84849507-84849507 on Build GRCh38
rs750132587 Chr.16: 84849367-84849368 on Build GRCh38
rs374775251 Chr.16: 84849549-84849549 on Build GRCh38
rs930734441 Chr.16: 84849285-84849285 on Build GRCh38
rs765087699 Chr.16: 84849365-84849366 on Build GRCh38
rs140214661 Chr.16: 84849397-84849397 on Build GRCh38
rs760941315 Chr.16: 84849554-84849554 on Build GRCh38
rs754474807 Chr.16: 84849406-84849406 on Build GRCh38
rs755676968 Chr.16: 84849345-84849345 on Build GRCh38
rs769215026 Chr.16: 84849546-84849546 on Build GRCh38
rs902319078 Chr.16: 84849646-84849646 on Build GRCh38
rs111566349 Chr.16: 84849350-84849350 on Build GRCh38
rs781236822 Chr.16: 84849490-84849490 on Build GRCh38
rs1028173989 Chr.16: 84849324-84849324 on Build GRCh38

View Full Product Details