Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 87330411-87330912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO31
Entrez Gene ID
79791
Gene Name
F-box protein 31
Gene Aliases
FBX14, FBXO14, Fbx31, MRT45, pp2386
UniGene
Hs.567582
Gene Chromosome Location
Chr.16: 87326987-87392107 on Build GRCh38
Transcripts
NM_001282683.1 NM_024735.4

SNP Detail

SNP ID Chromosome Location
rs911891072 Chr.16: 87330491-87330491 on Build GRCh38
rs951059771 Chr.16: 87330504-87330504 on Build GRCh38
rs1003181818 Chr.16: 87330629-87330629 on Build GRCh38
rs1039021365 Chr.16: 87330511-87330511 on Build GRCh38
rs548858918 Chr.16: 87330443-87330443 on Build GRCh38
rs892021738 Chr.16: 87330587-87330587 on Build GRCh38
rs946270162 Chr.16: 87330885-87330885 on Build GRCh38
rs538691472 Chr.16: 87330494-87330494 on Build GRCh38
rs1042258200 Chr.16: 87330880-87330880 on Build GRCh38
rs1020676178 Chr.16: 87330551-87330551 on Build GRCh38
rs569690880 Chr.16: 87330747-87330747 on Build GRCh38
rs142295558 Chr.16: 87330578-87330578 on Build GRCh38
rs79224744 Chr.16: 87330882-87330882 on Build GRCh38
rs530366893 Chr.16: 87330435-87330435 on Build GRCh38
rs983589987 Chr.16: 87330758-87330758 on Build GRCh38
rs1010257913 Chr.16: 87330683-87330683 on Build GRCh38
rs1006969943 Chr.16: 87330717-87330717 on Build GRCh38
rs983774785 Chr.16: 87330810-87330810 on Build GRCh38
rs927480428 Chr.16: 87330545-87330545 on Build GRCh38
rs938874289 Chr.16: 87330867-87330867 on Build GRCh38
rs932552689 Chr.16: 87330811-87330811 on Build GRCh38
rs1003317668 Chr.16: 87330883-87330883 on Build GRCh38
rs1021729108 Chr.16: 87330433-87330433 on Build GRCh38
rs959874384 Chr.16: 87330698-87330698 on Build GRCh38
rs968764056 Chr.16: 87330434-87330434 on Build GRCh38
rs189017025 Chr.16: 87330528-87330528 on Build GRCh38
rs568627687 Chr.16: 87330696-87330696 on Build GRCh38
rs927667786 Chr.16: 87330853-87330853 on Build GRCh38
rs764633828 Chr.16: 87330492-87330492 on Build GRCh38
rs571387352 Chr.16: 87330490-87330490 on Build GRCh38
rs536202763 Chr.16: 87330536-87330536 on Build GRCh38
rs920798601 Chr.16: 87330454-87330454 on Build GRCh38
rs1047837718 Chr.16: 87330487-87330487 on Build GRCh38
rs17771103 Chr.16: 87330772-87330772 on Build GRCh38
rs139375640 Chr.16: 87330825-87330825 on Build GRCh38
rs140176268 Chr.16: 87330450-87330450 on Build GRCh38
rs1057383238 Chr.16: 87330586-87330586 on Build GRCh38
rs920754492 Chr.16: 87330751-87330751 on Build GRCh38
rs973956381 Chr.16: 87330453-87330453 on Build GRCh38
rs1054014359 Chr.16: 87330532-87330532 on Build GRCh38
rs548162745 Chr.16: 87330477-87330477 on Build GRCh38
rs950953358 Chr.16: 87330755-87330755 on Build GRCh38
rs976695432 Chr.16: 87330460-87330460 on Build GRCh38
rs866298968 Chr.16: 87330771-87330771 on Build GRCh38
rs544290904 Chr.16: 87330793-87330793 on Build GRCh38
rs528074254 Chr.16: 87330459-87330459 on Build GRCh38
rs893716178 Chr.16: 87330859-87330859 on Build GRCh38
rs899721574 Chr.16: 87330513-87330513 on Build GRCh38
rs750291144 Chr.16: 87330706-87330706 on Build GRCh38
rs996796463 Chr.16: 87330515-87330515 on Build GRCh38
rs911946512 Chr.16: 87330759-87330759 on Build GRCh38
rs921983937 Chr.16: 87330794-87330794 on Build GRCh38
rs980548150 Chr.16: 87330835-87330835 on Build GRCh38
rs550373969 Chr.16: 87330681-87330681 on Build GRCh38
rs951277612 Chr.16: 87330789-87330789 on Build GRCh38
rs552342120 Chr.16: 87330461-87330461 on Build GRCh38
rs8063861 Chr.16: 87330784-87330784 on Build GRCh38
rs77403777 Chr.16: 87330510-87330510 on Build GRCh38
rs945158889 Chr.16: 87330872-87330872 on Build GRCh38
rs758359206 Chr.16: 87330752-87330752 on Build GRCh38
rs554830652 Chr.16: 87330572-87330572 on Build GRCh38
rs1033334931 Chr.16: 87330884-87330884 on Build GRCh38
rs893674025 Chr.16: 87330535-87330535 on Build GRCh38
rs1033364295 Chr.16: 87330633-87330633 on Build GRCh38
rs150871757 Chr.16: 87330634-87330634 on Build GRCh38

View Full Product Details