Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 87644540-87645039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JPH3
Entrez Gene ID
57338
Gene Name
junctophilin 3
Gene Aliases
CAGL237, HDL2, JP-3, JP3, TNRC22
UniGene
Hs.592068
Gene Chromosome Location
Chr.16: 87601835-87698156 on Build GRCh38
Transcripts
NM_020655.3 NR_073379.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
974628 c.922G>A p.G308R Chr.16: 87644797-87644797 on Build GRCh38
5003260 c.956G>A p.R319Q Chr.16: 87644831-87644831 on Build GRCh38
1709498 c.1063C>T p.R355W Chr.16: 87644938-87644938 on Build GRCh38
6145788 c.1066G>T p.A356S Chr.16: 87644941-87644941 on Build GRCh38
1735527 c.904G>T p.V302L Chr.16: 87644779-87644779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751551858 Chr.16: 87644779-87644779 on Build GRCh38
rs763168979 Chr.16: 87644620-87644620 on Build GRCh38
rs762639695 Chr.16: 87644828-87644828 on Build GRCh38
rs554628915 Chr.16: 87644788-87644788 on Build GRCh38
rs538911482 Chr.16: 87644849-87644849 on Build GRCh38
rs757427388 Chr.16: 87644911-87644911 on Build GRCh38
rs766775504 Chr.16: 87644832-87644832 on Build GRCh38
rs34442559 Chr.16: 87644613-87644613 on Build GRCh38
rs199599956 Chr.16: 87644660-87644660 on Build GRCh38
rs558669673 Chr.16: 87645015-87645015 on Build GRCh38
rs139015848 Chr.16: 87644775-87644775 on Build GRCh38
rs762909625 Chr.16: 87644870-87644870 on Build GRCh38
rs571536520 Chr.16: 87644703-87644703 on Build GRCh38
rs34975147 Chr.16: 87644970-87644970 on Build GRCh38
rs775157268 Chr.16: 87644826-87644826 on Build GRCh38
rs367750188 Chr.16: 87644709-87644709 on Build GRCh38
rs200634233 Chr.16: 87644990-87644990 on Build GRCh38
rs375292901 Chr.16: 87644710-87644710 on Build GRCh38
rs772600210 Chr.16: 87644609-87644609 on Build GRCh38
rs747066238 Chr.16: 87644850-87644850 on Build GRCh38
rs142273999 Chr.16: 87644642-87644642 on Build GRCh38
rs370880429 Chr.16: 87644939-87644939 on Build GRCh38
rs553005716 Chr.16: 87644697-87644697 on Build GRCh38
rs139022605 Chr.16: 87644565-87644565 on Build GRCh38
rs201202798 Chr.16: 87644881-87644881 on Build GRCh38
rs147468725 Chr.16: 87644905-87644905 on Build GRCh38
rs148131421 Chr.16: 87644830-87644830 on Build GRCh38
rs765608904 Chr.16: 87644619-87644619 on Build GRCh38
rs543680188 Chr.16: 87644735-87644735 on Build GRCh38
rs775908489 Chr.16: 87644657-87644657 on Build GRCh38
rs761450944 Chr.16: 87644831-87644831 on Build GRCh38
rs148709536 Chr.16: 87644631-87644631 on Build GRCh38
rs764738017 Chr.16: 87644708-87644709 on Build GRCh38
rs780773717 Chr.16: 87644789-87644789 on Build GRCh38
rs865800701 Chr.16: 87644878-87644878 on Build GRCh38
rs375238392 Chr.16: 87644991-87644991 on Build GRCh38
rs753117810 Chr.16: 87644837-87644837 on Build GRCh38
rs777642426 Chr.16: 87644561-87644561 on Build GRCh38
rs113233906 Chr.16: 87644757-87644757 on Build GRCh38
rs144550250 Chr.16: 87644908-87644908 on Build GRCh38
rs926962070 Chr.16: 87644766-87644766 on Build GRCh38
rs770113757 Chr.16: 87644653-87644653 on Build GRCh38
rs201133025 Chr.16: 87644567-87644567 on Build GRCh38
rs532653949 Chr.16: 87644770-87644770 on Build GRCh38
rs781773749 Chr.16: 87644700-87644700 on Build GRCh38
rs1027449834 Chr.16: 87644943-87644943 on Build GRCh38
rs746694210 Chr.16: 87644979-87644979 on Build GRCh38
rs17857118 Chr.16: 87645002-87645002 on Build GRCh38
rs377378251 Chr.16: 87644938-87644938 on Build GRCh38
rs199593264 Chr.16: 87644942-87644942 on Build GRCh38
rs773264027 Chr.16: 87644800-87644800 on Build GRCh38
rs963256027 Chr.16: 87644945-87644945 on Build GRCh38
rs8051490 Chr.16: 87644907-87644907 on Build GRCh38
rs751664775 Chr.16: 87644664-87644664 on Build GRCh38
rs771680407 Chr.16: 87644935-87644935 on Build GRCh38
rs879114912 Chr.16: 87644722-87644722 on Build GRCh38
rs150955715 Chr.16: 87644968-87644968 on Build GRCh38
rs760072867 Chr.16: 87644698-87644698 on Build GRCh38
rs150628659 Chr.16: 87644862-87644862 on Build GRCh38
rs1007163373 Chr.16: 87644898-87644898 on Build GRCh38
rs200310769 Chr.16: 87644676-87644676 on Build GRCh38
rs758075712 Chr.16: 87645017-87645017 on Build GRCh38
rs3751725 Chr.16: 87644559-87644559 on Build GRCh38
rs200494558 Chr.16: 87644953-87644953 on Build GRCh38
rs755951557 Chr.16: 87644677-87644677 on Build GRCh38
rs35304352 Chr.16: 87644796-87644796 on Build GRCh38
rs368992633 Chr.16: 87644808-87644808 on Build GRCh38
rs149347940 Chr.16: 87644868-87644868 on Build GRCh38
rs199967240 Chr.16: 87644995-87644995 on Build GRCh38
rs763262123 Chr.16: 87644658-87644658 on Build GRCh38
rs200626151 Chr.16: 87644649-87644649 on Build GRCh38
rs770946209 Chr.16: 87644853-87644853 on Build GRCh38
rs144628082 Chr.16: 87644772-87644772 on Build GRCh38
rs1010227661 Chr.16: 87644999-87644999 on Build GRCh38
rs147830852 Chr.16: 87644715-87644715 on Build GRCh38
rs144950183 Chr.16: 87644745-87644745 on Build GRCh38
rs917417412 Chr.16: 87644753-87644753 on Build GRCh38
rs367865537 Chr.16: 87644716-87644716 on Build GRCh38
rs150019020 Chr.16: 87644614-87644614 on Build GRCh38
rs756262017 Chr.16: 87644804-87644804 on Build GRCh38
rs376157795 Chr.16: 87644844-87644844 on Build GRCh38
rs142274475 Chr.16: 87644778-87644778 on Build GRCh38
rs144295273 Chr.16: 87644622-87644622 on Build GRCh38
rs144403955 Chr.16: 87644696-87644696 on Build GRCh38
rs149023008 Chr.16: 87644769-87644769 on Build GRCh38
rs369581247 Chr.16: 87644895-87644895 on Build GRCh38
rs772099172 Chr.16: 87644764-87644764 on Build GRCh38
rs147853977 Chr.16: 87644910-87644910 on Build GRCh38
rs759416796 Chr.16: 87644815-87644815 on Build GRCh38
rs773282602 Chr.16: 87644768-87644768 on Build GRCh38
rs748773642 Chr.16: 87644639-87644639 on Build GRCh38
rs768392713 Chr.16: 87644871-87644871 on Build GRCh38
rs951622602 Chr.16: 87644805-87644805 on Build GRCh38
rs951600749 Chr.16: 87644712-87644712 on Build GRCh38
rs567320827 Chr.16: 87644802-87644802 on Build GRCh38
rs747743618 Chr.16: 87644932-87644932 on Build GRCh38
rs144601410 Chr.16: 87644814-87644814 on Build GRCh38
rs35538359 Chr.16: 87644740-87644740 on Build GRCh38
rs955237917 Chr.16: 87644714-87644714 on Build GRCh38
rs748340460 Chr.16: 87644762-87644762 on Build GRCh38
rs920163940 Chr.16: 87644571-87644571 on Build GRCh38
rs780181019 Chr.16: 87644925-87644925 on Build GRCh38
rs779032493 Chr.16: 87645010-87645010 on Build GRCh38
rs749892195 Chr.16: 87644768-87644769 on Build GRCh38
rs202066813 Chr.16: 87644973-87644973 on Build GRCh38
rs763479212 Chr.16: 87644952-87644952 on Build GRCh38
rs9934222 Chr.16: 87644643-87644643 on Build GRCh38
rs756677356 Chr.16: 87644785-87644785 on Build GRCh38
rs200030601 Chr.16: 87644728-87644728 on Build GRCh38
rs61737926 Chr.16: 87644718-87644718 on Build GRCh38
rs370806682 Chr.16: 87644558-87644558 on Build GRCh38
rs761581216 Chr.16: 87644894-87644894 on Build GRCh38
rs772295091 Chr.16: 87644799-87644799 on Build GRCh38
rs761599199 Chr.16: 87644665-87644665 on Build GRCh38
rs376022547 Chr.16: 87644650-87644650 on Build GRCh38
rs767548668 Chr.16: 87644670-87644670 on Build GRCh38
rs369550086 Chr.16: 87644733-87644733 on Build GRCh38
rs749721814 Chr.16: 87644747-87644747 on Build GRCh38
rs35084631 Chr.16: 87644928-87644928 on Build GRCh38
rs142136496 Chr.16: 87644608-87644608 on Build GRCh38
rs140676785 Chr.16: 87644688-87644688 on Build GRCh38
rs748335498 Chr.16: 87644797-87644797 on Build GRCh38
rs754329526 Chr.16: 87644833-87644833 on Build GRCh38
rs1055743372 Chr.16: 87644916-87644916 on Build GRCh38
rs781163343 Chr.16: 87644861-87644861 on Build GRCh38
rs774198567 Chr.16: 87644890-87644890 on Build GRCh38
rs778836120 Chr.16: 87644971-87644971 on Build GRCh38
rs751131621 Chr.16: 87644784-87644784 on Build GRCh38
rs769594945 Chr.16: 87644823-87644823 on Build GRCh38
rs766984416 Chr.16: 87644904-87644904 on Build GRCh38
rs566359692 Chr.16: 87645001-87645001 on Build GRCh38
rs565470642 Chr.16: 87644739-87644739 on Build GRCh38
rs1039614803 Chr.16: 87644899-87644899 on Build GRCh38
rs751937922 Chr.16: 87644954-87644954 on Build GRCh38
rs764074006 Chr.16: 87644621-87644621 on Build GRCh38
rs754027911 Chr.16: 87645004-87645004 on Build GRCh38
rs750215162 Chr.16: 87644671-87644671 on Build GRCh38
rs1019784866 Chr.16: 87644809-87644809 on Build GRCh38
rs144594225 Chr.16: 87644982-87644982 on Build GRCh38
rs371703523 Chr.16: 87644719-87644719 on Build GRCh38
rs760635261 Chr.16: 87644721-87644721 on Build GRCh38
rs772774074 Chr.16: 87644937-87644937 on Build GRCh38
rs200608117 Chr.16: 87644983-87644983 on Build GRCh38
rs375201712 Chr.16: 87644969-87644969 on Build GRCh38
rs572969321 Chr.16: 87644906-87644906 on Build GRCh38
rs565191182 Chr.16: 87644595-87644595 on Build GRCh38
rs767702486 Chr.16: 87644965-87644965 on Build GRCh38
rs112449440 Chr.16: 87644730-87644730 on Build GRCh38
rs34200070 Chr.16: 87644865-87644865 on Build GRCh38
rs142622776 Chr.16: 87644955-87644955 on Build GRCh38
rs755421426 Chr.16: 87644793-87644793 on Build GRCh38
rs1045061295 Chr.16: 87644758-87644758 on Build GRCh38
rs147884034 Chr.16: 87644974-87644974 on Build GRCh38
rs372821727 Chr.16: 87644918-87644918 on Build GRCh38
rs769767671 Chr.16: 87644919-87644919 on Build GRCh38
rs749070969 Chr.16: 87644926-87644926 on Build GRCh38
rs745483239 Chr.16: 87644583-87644583 on Build GRCh38
rs752113370 Chr.16: 87644737-87644737 on Build GRCh38
rs781280078 Chr.16: 87644917-87644917 on Build GRCh38
rs770238690 Chr.16: 87644704-87644704 on Build GRCh38
rs371378713 Chr.16: 87644627-87644627 on Build GRCh38
rs548027626 Chr.16: 87644781-87644781 on Build GRCh38
rs8051448 Chr.16: 87644835-87644835 on Build GRCh38
rs761265274 Chr.16: 87644610-87644610 on Build GRCh38
rs201066038 Chr.16: 87644579-87644579 on Build GRCh38
rs371908265 Chr.16: 87644989-87644989 on Build GRCh38
rs774674671 Chr.16: 87644760-87644760 on Build GRCh38
rs755750862 Chr.16: 87644632-87644632 on Build GRCh38
rs767533433 Chr.16: 87644792-87644792 on Build GRCh38
rs766095139 Chr.16: 87644724-87644724 on Build GRCh38
rs980718623 Chr.16: 87644933-87644933 on Build GRCh38
rs779721921 Chr.16: 87644637-87644637 on Build GRCh38
rs747404678 Chr.16: 87644645-87644645 on Build GRCh38
rs142310848 Chr.16: 87644691-87644691 on Build GRCh38
rs372386767 Chr.16: 87644834-87644834 on Build GRCh38
rs202199160 Chr.16: 87644587-87644587 on Build GRCh38
rs188752914 Chr.16: 87644994-87644994 on Build GRCh38
rs979266176 Chr.16: 87644612-87644612 on Build GRCh38
rs111568864 Chr.16: 87645003-87645003 on Build GRCh38
rs779846817 Chr.16: 87644679-87644679 on Build GRCh38
rs780784674 Chr.16: 87644630-87644630 on Build GRCh38
rs563154923 Chr.16: 87644931-87644931 on Build GRCh38

View Full Product Details