Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 87689870-87690361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JPH3
Entrez Gene ID
57338
Gene Name
junctophilin 3
Gene Aliases
CAGL237, HDL2, JP-3, JP3, TNRC22
UniGene
Hs.592068
Gene Chromosome Location
Chr.16: 87601835-87698156 on Build GRCh38
Transcripts
NM_020655.3 NR_073379.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4590162 c.1726A>G p.T576A Chr.16: 87690086-87690086 on Build GRCh38
6859745 c.1633G>A p.G545S Chr.16: 87689993-87689993 on Build GRCh38
5571653 c.1743G>A p.V581V Chr.16: 87690103-87690103 on Build GRCh38
5034679 c.1740C>T p.P580P Chr.16: 87690100-87690100 on Build GRCh38
4591703 c.1732T>G p.S578A Chr.16: 87690092-87690092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753688162 Chr.16: 87690192-87690192 on Build GRCh38
rs751414231 Chr.16: 87690077-87690077 on Build GRCh38
rs760254773 Chr.16: 87690168-87690168 on Build GRCh38
rs139833288 Chr.16: 87690187-87690187 on Build GRCh38
rs775198774 Chr.16: 87690306-87690306 on Build GRCh38
rs761731152 Chr.16: 87690164-87690165 on Build GRCh38
rs765897521 Chr.16: 87690182-87690182 on Build GRCh38
rs759067110 Chr.16: 87690268-87690268 on Build GRCh38
rs748627807 Chr.16: 87690108-87690108 on Build GRCh38
rs997510082 Chr.16: 87690272-87690272 on Build GRCh38
rs570976769 Chr.16: 87690087-87690087 on Build GRCh38
rs762805375 Chr.16: 87690076-87690076 on Build GRCh38
rs970453336 Chr.16: 87690158-87690158 on Build GRCh38
rs770390129 Chr.16: 87689896-87689896 on Build GRCh38
rs780668828 Chr.16: 87690014-87690014 on Build GRCh38
rs767952095 Chr.16: 87690320-87690320 on Build GRCh38
rs765252754 Chr.16: 87690058-87690058 on Build GRCh38
rs1020256184 Chr.16: 87689974-87689974 on Build GRCh38
rs773247332 Chr.16: 87690195-87690218 on Build GRCh38
rs190793959 Chr.16: 87689929-87689929 on Build GRCh38
rs750976547 Chr.16: 87689994-87689994 on Build GRCh38
rs752043597 Chr.16: 87690270-87690270 on Build GRCh38
rs778971502 Chr.16: 87690290-87690290 on Build GRCh38
rs924968124 Chr.16: 87689992-87689992 on Build GRCh38
rs776540937 Chr.16: 87690049-87690049 on Build GRCh38
rs186820422 Chr.16: 87690052-87690052 on Build GRCh38
rs771342408 Chr.16: 87690047-87690047 on Build GRCh38
rs375693785 Chr.16: 87690331-87690331 on Build GRCh38
rs748628691 Chr.16: 87690029-87690029 on Build GRCh38
rs868543694 Chr.16: 87690178-87690178 on Build GRCh38
rs771254621 Chr.16: 87690124-87690124 on Build GRCh38
rs372525495 Chr.16: 87690109-87690109 on Build GRCh38
rs748262681 Chr.16: 87690335-87690335 on Build GRCh38
rs926563309 Chr.16: 87690025-87690025 on Build GRCh38
rs533405611 Chr.16: 87690067-87690067 on Build GRCh38
rs900349372 Chr.16: 87689942-87689942 on Build GRCh38
rs746825843 Chr.16: 87690301-87690301 on Build GRCh38
rs775448005 Chr.16: 87690145-87690145 on Build GRCh38
rs866437377 Chr.16: 87690084-87690084 on Build GRCh38
rs893138182 Chr.16: 87689991-87689991 on Build GRCh38
rs780177994 Chr.16: 87690224-87690224 on Build GRCh38
rs543238672 Chr.16: 87689965-87689965 on Build GRCh38
rs201193066 Chr.16: 87690263-87690263 on Build GRCh38
rs759383771 Chr.16: 87689986-87689986 on Build GRCh38
rs376342781 Chr.16: 87690150-87690150 on Build GRCh38
rs755013193 Chr.16: 87690285-87690285 on Build GRCh38
rs983080109 Chr.16: 87690059-87690059 on Build GRCh38
rs779463840 Chr.16: 87690098-87690098 on Build GRCh38
rs373134894 Chr.16: 87690146-87690146 on Build GRCh38
rs1003287541 Chr.16: 87690317-87690317 on Build GRCh38
rs34478943 Chr.16: 87690131-87690132 on Build GRCh38
rs371557680 Chr.16: 87690194-87690194 on Build GRCh38
rs781608423 Chr.16: 87690273-87690273 on Build GRCh38
rs755603959 Chr.16: 87690091-87690091 on Build GRCh38
rs772495537 Chr.16: 87690031-87690031 on Build GRCh38
rs776905301 Chr.16: 87689973-87689973 on Build GRCh38
rs373622670 Chr.16: 87690246-87690246 on Build GRCh38
rs776382406 Chr.16: 87690303-87690303 on Build GRCh38
rs767240603 Chr.16: 87690151-87690151 on Build GRCh38
rs556010482 Chr.16: 87690271-87690271 on Build GRCh38
rs17853661 Chr.16: 87690294-87690294 on Build GRCh38
rs749538454 Chr.16: 87690283-87690283 on Build GRCh38
rs199695995 Chr.16: 87689905-87689905 on Build GRCh38
rs1052306184 Chr.16: 87689891-87689891 on Build GRCh38
rs201622335 Chr.16: 87689914-87689914 on Build GRCh38
rs747217021 Chr.16: 87690046-87690046 on Build GRCh38
rs34767155 Chr.16: 87690232-87690232 on Build GRCh38
rs920244359 Chr.16: 87689897-87689897 on Build GRCh38
rs375241947 Chr.16: 87690293-87690293 on Build GRCh38
rs765983495 Chr.16: 87690262-87690262 on Build GRCh38
rs764781615 Chr.16: 87690305-87690305 on Build GRCh38
rs142631971 Chr.16: 87690013-87690013 on Build GRCh38
rs769969398 Chr.16: 87690221-87690221 on Build GRCh38
rs199869682 Chr.16: 87690281-87690281 on Build GRCh38
rs750056330 Chr.16: 87690163-87690163 on Build GRCh38
rs957652501 Chr.16: 87690142-87690142 on Build GRCh38
rs756328772 Chr.16: 87690274-87690274 on Build GRCh38
rs541878565 Chr.16: 87689938-87689938 on Build GRCh38
rs879325847 Chr.16: 87689893-87689893 on Build GRCh38
rs764871450 Chr.16: 87690269-87690269 on Build GRCh38
rs755209651 Chr.16: 87690328-87690328 on Build GRCh38
rs769602156 Chr.16: 87690171-87690173 on Build GRCh38
rs370731271 Chr.16: 87690126-87690126 on Build GRCh38
rs778472211 Chr.16: 87690202-87690202 on Build GRCh38
rs866951277 Chr.16: 87690092-87690092 on Build GRCh38
rs767200310 Chr.16: 87690081-87690081 on Build GRCh38
rs766762923 Chr.16: 87690327-87690327 on Build GRCh38
rs781426384 Chr.16: 87690125-87690125 on Build GRCh38
rs1008839686 Chr.16: 87690103-87690103 on Build GRCh38
rs763905837 Chr.16: 87689996-87689996 on Build GRCh38
rs761773582 Chr.16: 87690243-87690243 on Build GRCh38
rs776294816 Chr.16: 87689998-87690009 on Build GRCh38
rs772935872 Chr.16: 87690259-87690259 on Build GRCh38
rs970922443 Chr.16: 87690312-87690312 on Build GRCh38
rs748061937 Chr.16: 87690291-87690291 on Build GRCh38
rs201403180 Chr.16: 87690326-87690326 on Build GRCh38
rs775456087 Chr.16: 87690075-87690075 on Build GRCh38
rs765259221 Chr.16: 87689988-87689988 on Build GRCh38
rs1041641005 Chr.16: 87690265-87690265 on Build GRCh38
rs183515054 Chr.16: 87690005-87690005 on Build GRCh38
rs143854485 Chr.16: 87690220-87690220 on Build GRCh38
rs747517451 Chr.16: 87690195-87690195 on Build GRCh38
rs1008438932 Chr.16: 87689968-87689968 on Build GRCh38
rs759710426 Chr.16: 87690055-87690055 on Build GRCh38
rs758161832 Chr.16: 87689995-87689995 on Build GRCh38
rs975505654 Chr.16: 87689989-87689989 on Build GRCh38
rs780687326 Chr.16: 87689898-87689898 on Build GRCh38
rs748590824 Chr.16: 87689925-87689925 on Build GRCh38
rs757654078 Chr.16: 87690203-87690203 on Build GRCh38
rs755412585 Chr.16: 87690023-87690023 on Build GRCh38
rs150787562 Chr.16: 87690228-87690228 on Build GRCh38
rs750778818 Chr.16: 87690321-87690321 on Build GRCh38
rs547825767 Chr.16: 87690017-87690017 on Build GRCh38
rs571960172 Chr.16: 87689924-87689924 on Build GRCh38
rs781413111 Chr.16: 87690209-87690209 on Build GRCh38
rs747415043 Chr.16: 87690118-87690118 on Build GRCh38
rs866181370 Chr.16: 87690101-87690101 on Build GRCh38
rs1047097758 Chr.16: 87690080-87690080 on Build GRCh38
rs900120702 Chr.16: 87690119-87690119 on Build GRCh38
rs773723020 Chr.16: 87690037-87690037 on Build GRCh38
rs368142343 Chr.16: 87690302-87690302 on Build GRCh38
rs931857765 Chr.16: 87690117-87690117 on Build GRCh38
rs538000829 Chr.16: 87690127-87690127 on Build GRCh38
rs995549378 Chr.16: 87689955-87689955 on Build GRCh38
rs779526331 Chr.16: 87690028-87690028 on Build GRCh38
rs893488074 Chr.16: 87690280-87690280 on Build GRCh38
rs907535551 Chr.16: 87690064-87690064 on Build GRCh38
rs1047508420 Chr.16: 87689910-87689910 on Build GRCh38
rs760368196 Chr.16: 87690260-87690260 on Build GRCh38
rs773024520 Chr.16: 87690295-87690295 on Build GRCh38
rs200796473 Chr.16: 87689962-87689962 on Build GRCh38
rs758938441 Chr.16: 87690200-87690200 on Build GRCh38
rs774104417 Chr.16: 87690149-87690149 on Build GRCh38

View Full Product Details