Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88657837-88658348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MVD
Entrez Gene ID
4597
Gene Name
mevalonate diphosphate decarboxylase
Gene Aliases
FP17780, MDDase, MPD, POROK7
UniGene
Hs.252457
Gene Chromosome Location
Chr.16: 88651935-88663153 on Build GRCh38
Transcripts
NM_002461.2 XM_011523086.2 XM_011523087.2 XM_011523088.2 XM_011523089.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4063498 c.176A>G p.D59G Chr.16: 88657995-88657995 on Build GRCh38
1189338 c.151A>G p.T51A Chr.16: 88658020-88658020 on Build GRCh38
117909 c.226C>T p.Q76* Chr.16: 88657945-88657945 on Build GRCh38
4938631 c.183C>A p.T61T Chr.16: 88657988-88657988 on Build GRCh38
4534781 c.211G>A p.E71K Chr.16: 88657960-88657960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984673655 Chr.16: 88658279-88658279 on Build GRCh38
rs771595297 Chr.16: 88657866-88657866 on Build GRCh38
rs764961928 Chr.16: 88658010-88658010 on Build GRCh38
rs751341258 Chr.16: 88657933-88657941 on Build GRCh38
rs761271641 Chr.16: 88658115-88658115 on Build GRCh38
rs887488599 Chr.16: 88658234-88658234 on Build GRCh38
rs746646920 Chr.16: 88658057-88658057 on Build GRCh38
rs1029915834 Chr.16: 88658268-88658268 on Build GRCh38
rs200434281 Chr.16: 88657975-88657975 on Build GRCh38
rs757341385 Chr.16: 88657900-88657900 on Build GRCh38
rs199644137 Chr.16: 88657916-88657916 on Build GRCh38
rs887405693 Chr.16: 88657918-88657918 on Build GRCh38
rs760020426 Chr.16: 88657868-88657868 on Build GRCh38
rs781276323 Chr.16: 88657902-88657902 on Build GRCh38
rs766384128 Chr.16: 88658006-88658006 on Build GRCh38
rs754779116 Chr.16: 88657963-88657963 on Build GRCh38
rs1007028029 Chr.16: 88658281-88658281 on Build GRCh38
rs947888259 Chr.16: 88658195-88658195 on Build GRCh38
rs756168102 Chr.16: 88657947-88657947 on Build GRCh38
rs748590880 Chr.16: 88658075-88658075 on Build GRCh38
rs923985519 Chr.16: 88658160-88658160 on Build GRCh38
rs562660029 Chr.16: 88658165-88658165 on Build GRCh38
rs765725508 Chr.16: 88657880-88657880 on Build GRCh38
rs11556525 Chr.16: 88658013-88658013 on Build GRCh38
rs991990396 Chr.16: 88658053-88658053 on Build GRCh38
rs375979090 Chr.16: 88657996-88657996 on Build GRCh38
rs752585992 Chr.16: 88658028-88658028 on Build GRCh38
rs143980554 Chr.16: 88657943-88657943 on Build GRCh38
rs747786145 Chr.16: 88657967-88657967 on Build GRCh38
rs145827664 Chr.16: 88658185-88658185 on Build GRCh38
rs772737070 Chr.16: 88657926-88657926 on Build GRCh38
rs775789539 Chr.16: 88658083-88658083 on Build GRCh38
rs374265631 Chr.16: 88657864-88657864 on Build GRCh38
rs556409224 Chr.16: 88658172-88658172 on Build GRCh38
rs372796451 Chr.16: 88658043-88658043 on Build GRCh38
rs746569295 Chr.16: 88657980-88657980 on Build GRCh38
rs756843054 Chr.16: 88658058-88658058 on Build GRCh38
rs571494555 Chr.16: 88658210-88658210 on Build GRCh38
rs748975666 Chr.16: 88657861-88657861 on Build GRCh38
rs1040694745 Chr.16: 88658159-88658159 on Build GRCh38
rs977362195 Chr.16: 88658036-88658036 on Build GRCh38
rs1049432348 Chr.16: 88658315-88658315 on Build GRCh38
rs997613135 Chr.16: 88658266-88658266 on Build GRCh38
rs773279830 Chr.16: 88658001-88658001 on Build GRCh38
rs200806905 Chr.16: 88658067-88658067 on Build GRCh38
rs527686536 Chr.16: 88658060-88658060 on Build GRCh38
rs150594289 Chr.16: 88657921-88657921 on Build GRCh38
rs763386948 Chr.16: 88657893-88657893 on Build GRCh38
rs2279258 Chr.16: 88657939-88657939 on Build GRCh38
rs756041753 Chr.16: 88657906-88657906 on Build GRCh38
rs200508667 Chr.16: 88657920-88657920 on Build GRCh38
rs1018458004 Chr.16: 88658287-88658287 on Build GRCh38
rs368790531 Chr.16: 88657865-88657865 on Build GRCh38
rs762031727 Chr.16: 88657941-88657941 on Build GRCh38
rs911300577 Chr.16: 88658095-88658095 on Build GRCh38
rs916067099 Chr.16: 88657903-88657903 on Build GRCh38
rs369603174 Chr.16: 88658002-88658002 on Build GRCh38
rs772306222 Chr.16: 88658076-88658076 on Build GRCh38
rs371763667 Chr.16: 88657968-88657968 on Build GRCh38
rs577551073 Chr.16: 88658156-88658156 on Build GRCh38
rs202087443 Chr.16: 88657989-88657989 on Build GRCh38
rs1014974195 Chr.16: 88658259-88658259 on Build GRCh38
rs1014593633 Chr.16: 88658033-88658033 on Build GRCh38
rs139545454 Chr.16: 88657922-88657922 on Build GRCh38
rs149300368 Chr.16: 88657933-88657933 on Build GRCh38
rs979555976 Chr.16: 88658312-88658312 on Build GRCh38
rs779850940 Chr.16: 88657910-88657910 on Build GRCh38
rs976997400 Chr.16: 88658306-88658306 on Build GRCh38
rs567141920 Chr.16: 88658062-88658062 on Build GRCh38
rs899733 Chr.16: 88658321-88658321 on Build GRCh38
rs200435526 Chr.16: 88658008-88658008 on Build GRCh38
rs769240449 Chr.16: 88658065-88658065 on Build GRCh38
rs1020798120 Chr.16: 88658186-88658186 on Build GRCh38
rs574893603 Chr.16: 88658176-88658176 on Build GRCh38
rs961657047 Chr.16: 88657917-88657917 on Build GRCh38
rs768586565 Chr.16: 88658130-88658131 on Build GRCh38
rs897476833 Chr.16: 88658260-88658260 on Build GRCh38
rs372523184 Chr.16: 88658064-88658064 on Build GRCh38
rs961905152 Chr.16: 88658042-88658042 on Build GRCh38
rs766367485 Chr.16: 88657950-88657950 on Build GRCh38
rs778700499 Chr.16: 88657966-88657966 on Build GRCh38
rs774779770 Chr.16: 88657962-88657962 on Build GRCh38
rs528393793 Chr.16: 88657938-88657938 on Build GRCh38
rs116232108 Chr.16: 88657940-88657940 on Build GRCh38
rs991544324 Chr.16: 88658246-88658246 on Build GRCh38
rs541832090 Chr.16: 88658258-88658258 on Build GRCh38
rs776110807 Chr.16: 88657934-88657934 on Build GRCh38
rs758096190 Chr.16: 88658030-88658030 on Build GRCh38
rs188461772 Chr.16: 88658308-88658308 on Build GRCh38
rs770561548 Chr.16: 88657981-88657981 on Build GRCh38
rs751801850 Chr.16: 88657898-88657898 on Build GRCh38
rs749757442 Chr.16: 88657982-88657982 on Build GRCh38
rs771486207 Chr.16: 88657923-88657923 on Build GRCh38
rs746384333 Chr.16: 88657932-88657932 on Build GRCh38
rs754868962 Chr.16: 88658009-88658009 on Build GRCh38
rs1012064332 Chr.16: 88657987-88657987 on Build GRCh38
rs56812649 Chr.16: 88657867-88657867 on Build GRCh38
rs868222 Chr.16: 88658276-88658276 on Build GRCh38
rs753061445 Chr.16: 88657884-88657884 on Build GRCh38
rs764452317 Chr.16: 88657894-88657894 on Build GRCh38
rs147999959 Chr.16: 88657905-88657905 on Build GRCh38
rs137930849 Chr.16: 88658022-88658023 on Build GRCh38
rs141636956 Chr.16: 88658155-88658155 on Build GRCh38
rs764314318 Chr.16: 88658184-88658184 on Build GRCh38
rs1043766216 Chr.16: 88657952-88657952 on Build GRCh38
rs902412967 Chr.16: 88658200-88658200 on Build GRCh38
rs940222282 Chr.16: 88658300-88658300 on Build GRCh38
rs750470646 Chr.16: 88657946-88657946 on Build GRCh38
rs1049941872 Chr.16: 88658171-88658171 on Build GRCh38
rs768253124 Chr.16: 88657862-88657862 on Build GRCh38
rs568814170 Chr.16: 88657988-88657988 on Build GRCh38
rs542229683 Chr.16: 88658105-88658105 on Build GRCh38

View Full Product Details