Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88731605-88732103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100289580
Entrez Gene ID
100289580
Gene Name
uncharacterized LOC100289580
Gene Aliases
-
UniGene
Hs.437712
Gene Chromosome Location
Chr.16: 88731180-88741425 on Build GRCh38
Transcripts
NR_103774.1 NM_001142864.3

Gene Symbol
PIEZO1
Entrez Gene ID
9780
Gene Name
piezo type mechanosensitive ion channel component 1
Gene Aliases
DHS, FAM38A, LMPH3, Mib
UniGene
Hs.377001
Gene Chromosome Location
Chr.16: 88715338-88785220 on Build GRCh38
Transcripts
NR_103774.1 NM_001142864.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5771171 c.3196G>T p.D1066Y Chr.16: 88731706-88731706 on Build GRCh38
3721720 c.3107G>A p.R1036H Chr.16: 88731795-88731795 on Build GRCh38
7352051 c.2992-1G>C p.? Chr.16: 88731911-88731911 on Build GRCh38
4884383 c.3168C>A p.C1056* Chr.16: 88731734-88731734 on Build GRCh38
239792 c.3106C>G p.R1036G Chr.16: 88731796-88731796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750862705 Chr.16: 88731819-88731819 on Build GRCh38
rs1051273700 Chr.16: 88731953-88731953 on Build GRCh38
rs142870142 Chr.16: 88731913-88731917 on Build GRCh38
rs963260252 Chr.16: 88732046-88732046 on Build GRCh38
rs535516669 Chr.16: 88731651-88731651 on Build GRCh38
rs1029340637 Chr.16: 88732057-88732057 on Build GRCh38
rs746719928 Chr.16: 88731953-88731953 on Build GRCh38
rs374777922 Chr.16: 88731954-88731955 on Build GRCh38
rs182634817 Chr.16: 88731964-88731964 on Build GRCh38
rs1013624205 Chr.16: 88732026-88732026 on Build GRCh38
rs747246644 Chr.16: 88731956-88731957 on Build GRCh38
rs562632417 Chr.16: 88731746-88731746 on Build GRCh38
rs184229578 Chr.16: 88731675-88731675 on Build GRCh38
rs999319090 Chr.16: 88731818-88731818 on Build GRCh38
rs113313581 Chr.16: 88731928-88731928 on Build GRCh38
rs755141831 Chr.16: 88731709-88731709 on Build GRCh38
rs778084729 Chr.16: 88731924-88731925 on Build GRCh38
rs796713147 Chr.16: 88731953-88731954 on Build GRCh38
rs61742732 Chr.16: 88731773-88731773 on Build GRCh38
rs953736672 Chr.16: 88732059-88732059 on Build GRCh38
rs978048890 Chr.16: 88731997-88731997 on Build GRCh38
rs1031105512 Chr.16: 88732020-88732020 on Build GRCh38
rs757156165 Chr.16: 88731946-88731946 on Build GRCh38
rs550074132 Chr.16: 88731901-88731901 on Build GRCh38
rs924595608 Chr.16: 88731998-88731998 on Build GRCh38
rs775928653 Chr.16: 88732049-88732049 on Build GRCh38
rs758103840 Chr.16: 88731658-88731659 on Build GRCh38
rs1057462689 Chr.16: 88732013-88732013 on Build GRCh38
rs762537822 Chr.16: 88731798-88731798 on Build GRCh38
rs895869805 Chr.16: 88732016-88732016 on Build GRCh38
rs1007010581 Chr.16: 88731955-88731955 on Build GRCh38
rs978397789 Chr.16: 88732074-88732074 on Build GRCh38
rs1044518025 Chr.16: 88732023-88732023 on Build GRCh38
rs531840770 Chr.16: 88731892-88731892 on Build GRCh38
rs1006203767 Chr.16: 88732034-88732034 on Build GRCh38
rs867665287 Chr.16: 88731914-88731914 on Build GRCh38
rs748155949 Chr.16: 88731721-88731721 on Build GRCh38
rs941701205 Chr.16: 88731743-88731743 on Build GRCh38
rs878968614 Chr.16: 88731783-88731783 on Build GRCh38
rs541530110 Chr.16: 88731769-88731769 on Build GRCh38
rs190814667 Chr.16: 88731928-88731928 on Build GRCh38
rs745985077 Chr.16: 88731952-88731952 on Build GRCh38
rs754261447 Chr.16: 88731955-88731956 on Build GRCh38
rs922414644 Chr.16: 88731748-88731748 on Build GRCh38
rs772144581 Chr.16: 88731953-88731954 on Build GRCh38
rs151293846 Chr.16: 88731957-88731958 on Build GRCh38
rs757829396 Chr.16: 88731934-88731935 on Build GRCh38
rs770613391 Chr.16: 88731765-88731765 on Build GRCh38
rs972205686 Chr.16: 88732048-88732048 on Build GRCh38
rs150852918 Chr.16: 88731820-88731820 on Build GRCh38
rs748845956 Chr.16: 88731887-88731887 on Build GRCh38
rs955551496 Chr.16: 88731629-88731629 on Build GRCh38
rs754188492 Chr.16: 88731836-88731836 on Build GRCh38
rs932494887 Chr.16: 88731752-88731752 on Build GRCh38
rs369167809 Chr.16: 88731676-88731677 on Build GRCh38
rs968578513 Chr.16: 88731804-88731804 on Build GRCh38
rs910214556 Chr.16: 88732071-88732071 on Build GRCh38
rs752844197 Chr.16: 88731853-88731853 on Build GRCh38
rs1044821981 Chr.16: 88731719-88731719 on Build GRCh38
rs76070043 Chr.16: 88731933-88731933 on Build GRCh38
rs934705023 Chr.16: 88732012-88732012 on Build GRCh38
rs758574762 Chr.16: 88731931-88731931 on Build GRCh38
rs577256993 Chr.16: 88731694-88731694 on Build GRCh38
rs1036246258 Chr.16: 88731652-88731652 on Build GRCh38
rs865984438 Chr.16: 88731924-88731924 on Build GRCh38
rs148039351 Chr.16: 88731687-88731687 on Build GRCh38
rs765381181 Chr.16: 88731847-88731847 on Build GRCh38
rs140450778 Chr.16: 88731639-88731639 on Build GRCh38
rs759294984 Chr.16: 88731779-88731779 on Build GRCh38
rs751591176 Chr.16: 88731934-88731934 on Build GRCh38
rs980691083 Chr.16: 88732038-88732038 on Build GRCh38
rs1023198679 Chr.16: 88731667-88731667 on Build GRCh38
rs746946494 Chr.16: 88731873-88731873 on Build GRCh38
rs368019686 Chr.16: 88731880-88731880 on Build GRCh38
rs775153962 Chr.16: 88731796-88731796 on Build GRCh38
rs558085832 Chr.16: 88732015-88732015 on Build GRCh38
rs769506340 Chr.16: 88731795-88731795 on Build GRCh38
rs917866420 Chr.16: 88731848-88731848 on Build GRCh38
rs768204845 Chr.16: 88731810-88731810 on Build GRCh38
rs574697533 Chr.16: 88731767-88731767 on Build GRCh38
rs923923118 Chr.16: 88732080-88732080 on Build GRCh38
rs891412672 Chr.16: 88731954-88731954 on Build GRCh38
rs755940638 Chr.16: 88731960-88731960 on Build GRCh38
rs1005818843 Chr.16: 88731889-88731889 on Build GRCh38
rs373215260 Chr.16: 88731917-88731918 on Build GRCh38
rs969252917 Chr.16: 88732036-88732036 on Build GRCh38
rs745725883 Chr.16: 88731958-88731958 on Build GRCh38
rs560218689 Chr.16: 88731771-88731771 on Build GRCh38
rs1036125081 Chr.16: 88731929-88731929 on Build GRCh38
rs758445295 Chr.16: 88731854-88731854 on Build GRCh38
rs72811497 Chr.16: 88731922-88731922 on Build GRCh38
rs960212732 Chr.16: 88731659-88731666 on Build GRCh38
rs985257623 Chr.16: 88731974-88731974 on Build GRCh38
rs1036396772 Chr.16: 88731755-88731755 on Build GRCh38
rs775803222 Chr.16: 88731952-88731953 on Build GRCh38
rs775275185 Chr.16: 88731886-88731886 on Build GRCh38
rs749802104 Chr.16: 88731931-88731932 on Build GRCh38
rs922384407 Chr.16: 88731910-88731910 on Build GRCh38
rs755193254 Chr.16: 88731841-88731841 on Build GRCh38
rs746865902 Chr.16: 88731764-88731764 on Build GRCh38
rs1013080220 Chr.16: 88731840-88731840 on Build GRCh38
rs759215680 Chr.16: 88731960-88731961 on Build GRCh38
rs188605848 Chr.16: 88732021-88732021 on Build GRCh38
rs564623447 Chr.16: 88731876-88731876 on Build GRCh38
rs773861214 Chr.16: 88731893-88731893 on Build GRCh38
rs1051416948 Chr.16: 88732060-88732060 on Build GRCh38
rs979876480 Chr.16: 88731679-88731679 on Build GRCh38
rs969200104 Chr.16: 88731852-88731852 on Build GRCh38
rs932980553 Chr.16: 88732058-88732058 on Build GRCh38
rs772007163 Chr.16: 88731727-88731727 on Build GRCh38
rs568280615 Chr.16: 88731902-88731902 on Build GRCh38
rs759877212 Chr.16: 88731908-88731908 on Build GRCh38
rs757180228 Chr.16: 88731874-88731874 on Build GRCh38
rs8055958 Chr.16: 88731957-88731957 on Build GRCh38
rs780361976 Chr.16: 88731704-88731704 on Build GRCh38
rs750856502 Chr.16: 88731697-88731697 on Build GRCh38
rs1029888460 Chr.16: 88731766-88731766 on Build GRCh38
rs544520218 Chr.16: 88731699-88731699 on Build GRCh38
rs765485880 Chr.16: 88731919-88731919 on Build GRCh38
rs778993785 Chr.16: 88731715-88731715 on Build GRCh38
rs547552421 Chr.16: 88731945-88731945 on Build GRCh38
rs946624926 Chr.16: 88731786-88731786 on Build GRCh38
rs904030992 Chr.16: 88731996-88731996 on Build GRCh38
rs933011598 Chr.16: 88731940-88731940 on Build GRCh38
rs915081276 Chr.16: 88731978-88731978 on Build GRCh38
rs576678562 Chr.16: 88731834-88731834 on Build GRCh38
rs147256227 Chr.16: 88732045-88732045 on Build GRCh38
rs777818544 Chr.16: 88731860-88731860 on Build GRCh38
rs753984091 Chr.16: 88731708-88731708 on Build GRCh38
rs143911952 Chr.16: 88732030-88732030 on Build GRCh38
rs377171513 Chr.16: 88731923-88731923 on Build GRCh38
rs554257960 Chr.16: 88731662-88731662 on Build GRCh38
rs546208424 Chr.16: 88731680-88731680 on Build GRCh38
rs539445849 Chr.16: 88731991-88731991 on Build GRCh38
rs992086362 Chr.16: 88731692-88731692 on Build GRCh38
rs769629472 Chr.16: 88731881-88731881 on Build GRCh38
rs113010486 Chr.16: 88731990-88731990 on Build GRCh38
rs752967733 Chr.16: 88731922-88731923 on Build GRCh38
rs947889466 Chr.16: 88731717-88731717 on Build GRCh38

View Full Product Details