Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88881202-88881705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBFA2T3
Entrez Gene ID
863
Gene Name
CBFA2/RUNX1 translocation partner 3
Gene Aliases
ETO2, MTG16, MTGR2, RUNX1T3, ZMYND4
UniGene
Hs.513811
Gene Chromosome Location
Chr.16: 88874855-88977198 on Build GRCh38
Transcripts
NM_005187.5 NM_175931.2 XM_005256323.4 XM_011523419.2 XM_017023811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2922682 c.1240C>T p.R414W Chr.16: 88881453-88881453 on Build GRCh38
2922683 c.1238C>T p.T413M Chr.16: 88881455-88881455 on Build GRCh38
3957925 c.1377C>T p.S459S Chr.16: 88881316-88881316 on Build GRCh38
2922680 c.1262G>A p.R421H Chr.16: 88881431-88881431 on Build GRCh38
7400448 c.1231G>T p.E411* Chr.16: 88881462-88881462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770381912 Chr.16: 88881287-88881287 on Build GRCh38
rs938243766 Chr.16: 88881597-88881597 on Build GRCh38
rs754765577 Chr.16: 88881278-88881278 on Build GRCh38
rs776378149 Chr.16: 88881484-88881484 on Build GRCh38
rs781508322 Chr.16: 88881409-88881409 on Build GRCh38
rs773516482 Chr.16: 88881309-88881309 on Build GRCh38
rs76551644 Chr.16: 88881295-88881295 on Build GRCh38
rs376300984 Chr.16: 88881531-88881531 on Build GRCh38
rs768700399 Chr.16: 88881305-88881306 on Build GRCh38
rs953247967 Chr.16: 88881540-88881540 on Build GRCh38
rs572508410 Chr.16: 88881578-88881578 on Build GRCh38
rs776663362 Chr.16: 88881374-88881374 on Build GRCh38
rs1000313491 Chr.16: 88881461-88881461 on Build GRCh38
rs747801573 Chr.16: 88881242-88881242 on Build GRCh38
rs200173287 Chr.16: 88881332-88881332 on Build GRCh38
rs760401870 Chr.16: 88881366-88881366 on Build GRCh38
rs199758456 Chr.16: 88881498-88881498 on Build GRCh38
rs776009718 Chr.16: 88881254-88881254 on Build GRCh38
rs762112802 Chr.16: 88881270-88881270 on Build GRCh38
rs963111182 Chr.16: 88881629-88881629 on Build GRCh38
rs202023439 Chr.16: 88881327-88881327 on Build GRCh38
rs768592867 Chr.16: 88881418-88881418 on Build GRCh38
rs767181401 Chr.16: 88881431-88881431 on Build GRCh38
rs747418254 Chr.16: 88881408-88881408 on Build GRCh38
rs964608128 Chr.16: 88881542-88881542 on Build GRCh38
rs770291986 Chr.16: 88881253-88881253 on Build GRCh38
rs757681495 Chr.16: 88881452-88881452 on Build GRCh38
rs372853568 Chr.16: 88881387-88881387 on Build GRCh38
rs369868070 Chr.16: 88881384-88881384 on Build GRCh38
rs759685635 Chr.16: 88881375-88881375 on Build GRCh38
rs200201856 Chr.16: 88881439-88881439 on Build GRCh38
rs770602306 Chr.16: 88881323-88881323 on Build GRCh38
rs953310137 Chr.16: 88881681-88881681 on Build GRCh38
rs762088947 Chr.16: 88881494-88881497 on Build GRCh38
rs201101361 Chr.16: 88881339-88881339 on Build GRCh38
rs779121744 Chr.16: 88881538-88881538 on Build GRCh38
rs761066677 Chr.16: 88881534-88881534 on Build GRCh38
rs746047153 Chr.16: 88881410-88881410 on Build GRCh38
rs911827014 Chr.16: 88881561-88881561 on Build GRCh38
rs769427504 Chr.16: 88881306-88881306 on Build GRCh38
rs374570965 Chr.16: 88881349-88881349 on Build GRCh38
rs559181450 Chr.16: 88881657-88881664 on Build GRCh38
rs748009384 Chr.16: 88881468-88881468 on Build GRCh38
rs1047929811 Chr.16: 88881621-88881621 on Build GRCh38
rs751972114 Chr.16: 88881451-88881451 on Build GRCh38
rs764810186 Chr.16: 88881489-88881489 on Build GRCh38
rs771624644 Chr.16: 88881245-88881245 on Build GRCh38
rs199953961 Chr.16: 88881372-88881372 on Build GRCh38
rs770813127 Chr.16: 88881526-88881526 on Build GRCh38
rs142010820 Chr.16: 88881257-88881257 on Build GRCh38
rs2272434 Chr.16: 88881607-88881607 on Build GRCh38
rs931948694 Chr.16: 88881324-88881324 on Build GRCh38
rs762770559 Chr.16: 88881348-88881348 on Build GRCh38
rs754482001 Chr.16: 88881445-88881445 on Build GRCh38
rs2272437 Chr.16: 88881316-88881316 on Build GRCh38
rs887346041 Chr.16: 88881602-88881602 on Build GRCh38
rs886168375 Chr.16: 88881671-88881671 on Build GRCh38
rs756757934 Chr.16: 88881326-88881326 on Build GRCh38
rs779591431 Chr.16: 88881369-88881369 on Build GRCh38
rs546198726 Chr.16: 88881584-88881584 on Build GRCh38
rs9934287 Chr.16: 88881280-88881280 on Build GRCh38
rs766247590 Chr.16: 88881275-88881275 on Build GRCh38
rs748180411 Chr.16: 88881503-88881503 on Build GRCh38
rs200535158 Chr.16: 88881315-88881315 on Build GRCh38
rs751263024 Chr.16: 88881352-88881352 on Build GRCh38
rs779686048 Chr.16: 88881393-88881393 on Build GRCh38
rs371447156 Chr.16: 88881342-88881342 on Build GRCh38
rs370798291 Chr.16: 88881386-88881386 on Build GRCh38
rs747338837 Chr.16: 88881284-88881284 on Build GRCh38
rs201472610 Chr.16: 88881502-88881502 on Build GRCh38
rs540574674 Chr.16: 88881405-88881405 on Build GRCh38
rs373074041 Chr.16: 88881530-88881530 on Build GRCh38
rs892697542 Chr.16: 88881283-88881283 on Build GRCh38
rs753311916 Chr.16: 88881399-88881399 on Build GRCh38
rs2272435 Chr.16: 88881573-88881573 on Build GRCh38
rs542005739 Chr.16: 88881572-88881572 on Build GRCh38
rs755186829 Chr.16: 88881536-88881536 on Build GRCh38
rs1028746008 Chr.16: 88881676-88881676 on Build GRCh38
rs376412751 Chr.16: 88881333-88881333 on Build GRCh38
rs182111221 Chr.16: 88881286-88881286 on Build GRCh38
rs753406165 Chr.16: 88881444-88881444 on Build GRCh38
rs774382828 Chr.16: 88881463-88881463 on Build GRCh38
rs758972950 Chr.16: 88881400-88881400 on Build GRCh38
rs370799116 Chr.16: 88881535-88881535 on Build GRCh38
rs751936699 Chr.16: 88881678-88881678 on Build GRCh38
rs748631154 Chr.16: 88881370-88881370 on Build GRCh38
rs201612351 Chr.16: 88881337-88881337 on Build GRCh38
rs771905396 Chr.16: 88881504-88881504 on Build GRCh38
rs759501623 Chr.16: 88881328-88881328 on Build GRCh38
rs780189929 Chr.16: 88881455-88881455 on Build GRCh38
rs369377378 Chr.16: 88881500-88881500 on Build GRCh38
rs936079912 Chr.16: 88881582-88881582 on Build GRCh38
rs764366886 Chr.16: 88881390-88881390 on Build GRCh38
rs2272436 Chr.16: 88881565-88881565 on Build GRCh38
rs772522304 Chr.16: 88881334-88881334 on Build GRCh38
rs774772837 Chr.16: 88881305-88881305 on Build GRCh38
rs749772901 Chr.16: 88881296-88881296 on Build GRCh38
rs777712181 Chr.16: 88881505-88881505 on Build GRCh38
rs370102475 Chr.16: 88881281-88881281 on Build GRCh38
rs778360042 Chr.16: 88881403-88881403 on Build GRCh38
rs772698882 Chr.16: 88881246-88881246 on Build GRCh38
rs775893654 Chr.16: 88881291-88881291 on Build GRCh38
rs146302322 Chr.16: 88881560-88881560 on Build GRCh38
rs534264933 Chr.16: 88881273-88881273 on Build GRCh38
rs536310335 Chr.16: 88881255-88881255 on Build GRCh38
rs749297800 Chr.16: 88881414-88881414 on Build GRCh38
rs776914052 Chr.16: 88881356-88881359 on Build GRCh38
rs771957137 Chr.16: 88881470-88881470 on Build GRCh38
rs769912125 Chr.16: 88881411-88881411 on Build GRCh38
rs377105849 Chr.16: 88881454-88881454 on Build GRCh38
rs749960652 Chr.16: 88881367-88881367 on Build GRCh38
rs1053526 Chr.16: 88881407-88881407 on Build GRCh38
rs558848178 Chr.16: 88881527-88881527 on Build GRCh38
rs113459673 Chr.16: 88881591-88881591 on Build GRCh38
rs554277604 Chr.16: 88881256-88881256 on Build GRCh38
rs761698622 Chr.16: 88881430-88881430 on Build GRCh38
rs759151097 Chr.16: 88881486-88881486 on Build GRCh38
rs780548645 Chr.16: 88881606-88881606 on Build GRCh38
rs1014596179 Chr.16: 88881619-88881619 on Build GRCh38
rs931888674 Chr.16: 88881401-88881401 on Build GRCh38
rs201916925 Chr.16: 88881354-88881354 on Build GRCh38
rs1018633827 Chr.16: 88881541-88881541 on Build GRCh38
rs763942453 Chr.16: 88881383-88881383 on Build GRCh38
rs755529310 Chr.16: 88881368-88881368 on Build GRCh38
rs764724363 Chr.16: 88881446-88881446 on Build GRCh38
rs1055309118 Chr.16: 88881604-88881604 on Build GRCh38
rs766144998 Chr.16: 88881473-88881473 on Build GRCh38
rs11642210 Chr.16: 88881432-88881432 on Build GRCh38
rs970359780 Chr.16: 88881525-88881525 on Build GRCh38
rs767770896 Chr.16: 88881272-88881272 on Build GRCh38
rs755685805 Chr.16: 88881389-88881389 on Build GRCh38
rs369233901 Chr.16: 88881313-88881313 on Build GRCh38
rs771190748 Chr.16: 88881373-88881373 on Build GRCh38
rs762856664 Chr.16: 88881378-88881378 on Build GRCh38
rs994962092 Chr.16: 88881644-88881644 on Build GRCh38
rs750000298 Chr.16: 88881388-88881388 on Build GRCh38
rs367606571 Chr.16: 88881340-88881340 on Build GRCh38
rs760105812 Chr.16: 88881248-88881248 on Build GRCh38
rs767994473 Chr.16: 88881495-88881495 on Build GRCh38
rs377146698 Chr.16: 88881376-88881376 on Build GRCh38
rs775388886 Chr.16: 88881257-88881258 on Build GRCh38
rs1055916833 Chr.16: 88881583-88881583 on Build GRCh38
rs772875272 Chr.16: 88881472-88881472 on Build GRCh38
rs1014445057 Chr.16: 88881590-88881590 on Build GRCh38
rs930800585 Chr.16: 88881609-88881609 on Build GRCh38
rs781390631 Chr.16: 88881453-88881453 on Build GRCh38
rs560491264 Chr.16: 88881338-88881338 on Build GRCh38
rs775535175 Chr.16: 88881413-88881413 on Build GRCh38
rs529754752 Chr.16: 88881494-88881494 on Build GRCh38
rs767863623 Chr.16: 88881307-88881307 on Build GRCh38
rs925801998 Chr.16: 88881424-88881424 on Build GRCh38
rs750888629 Chr.16: 88881532-88881532 on Build GRCh38
rs189102389 Chr.16: 88881460-88881460 on Build GRCh38
rs772795593 Chr.16: 88881312-88881312 on Build GRCh38
rs766050818 Chr.16: 88881440-88881440 on Build GRCh38
rs768959796 Chr.16: 88881303-88881303 on Build GRCh38
rs564438686 Chr.16: 88881588-88881588 on Build GRCh38
rs746824719 Chr.16: 88881513-88881513 on Build GRCh38
rs760826925 Chr.16: 88881270-88881270 on Build GRCh38
rs750764995 Chr.16: 88881497-88881497 on Build GRCh38
rs369662071 Chr.16: 88881239-88881239 on Build GRCh38
rs752582705 Chr.16: 88881343-88881343 on Build GRCh38
rs375924138 Chr.16: 88881421-88881421 on Build GRCh38

View Full Product Details