Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89281106-89281607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD11
Entrez Gene ID
29123
Gene Name
ankyrin repeat domain 11
Gene Aliases
ANCO-1, ANCO1, LZ16, T13
UniGene
Hs.335003
Gene Chromosome Location
Chr.16: 89267619-89490561 on Build GRCh38
Transcripts
NM_001256182.1 NM_001256183.1 NM_013275.5 XM_006721181.1 XM_006721184.2 XM_011523051.2 XM_011523053.2 XM_011523054.2 XM_011523055.2 XM_011523056.2 XM_011523057.2 XM_017023182.1 XM_017023183.1 XM_017023184.1 XM_017023185.1 XM_017023186.1 XM_017023187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6464616 c.4976C>T p.P1659L Chr.16: 89281566-89281566 on Build GRCh38
3387678 c.5369C>T p.S1790L Chr.16: 89281173-89281173 on Build GRCh38
3889146 c.4938C>T p.D1646D Chr.16: 89281604-89281604 on Build GRCh38
6968954 c.5219C>T p.A1740V Chr.16: 89281323-89281323 on Build GRCh38
435934 c.5317G>T p.E1773* Chr.16: 89281225-89281225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960304355 Chr.16: 89281289-89281289 on Build GRCh38
rs143189255 Chr.16: 89281325-89281325 on Build GRCh38
rs746723349 Chr.16: 89281418-89281418 on Build GRCh38
rs761377016 Chr.16: 89281173-89281173 on Build GRCh38
rs146205997 Chr.16: 89281454-89281454 on Build GRCh38
rs773850236 Chr.16: 89281316-89281316 on Build GRCh38
rs757264535 Chr.16: 89281326-89281326 on Build GRCh38
rs761701060 Chr.16: 89281555-89281556 on Build GRCh38
rs768368719 Chr.16: 89281464-89281464 on Build GRCh38
rs199787658 Chr.16: 89281448-89281448 on Build GRCh38
rs771001061 Chr.16: 89281543-89281543 on Build GRCh38
rs202042792 Chr.16: 89281512-89281512 on Build GRCh38
rs1035902808 Chr.16: 89281382-89281382 on Build GRCh38
rs201160642 Chr.16: 89281551-89281551 on Build GRCh38
rs142473716 Chr.16: 89281504-89281504 on Build GRCh38
rs748736120 Chr.16: 89281354-89281354 on Build GRCh38
rs945400694 Chr.16: 89281304-89281304 on Build GRCh38
rs758053289 Chr.16: 89281185-89281185 on Build GRCh38
rs765241145 Chr.16: 89281425-89281425 on Build GRCh38
rs755369887 Chr.16: 89281394-89281394 on Build GRCh38
rs746152599 Chr.16: 89281300-89281300 on Build GRCh38
rs767704252 Chr.16: 89281435-89281435 on Build GRCh38
rs369357756 Chr.16: 89281433-89281433 on Build GRCh38
rs955262955 Chr.16: 89281359-89281359 on Build GRCh38
rs781136937 Chr.16: 89281195-89281195 on Build GRCh38
rs765765688 Chr.16: 89281562-89281562 on Build GRCh38
rs137953940 Chr.16: 89281499-89281499 on Build GRCh38
rs766737603 Chr.16: 89281381-89281381 on Build GRCh38
rs770892853 Chr.16: 89281578-89281578 on Build GRCh38
rs140556973 Chr.16: 89281145-89281145 on Build GRCh38
rs148243995 Chr.16: 89281344-89281344 on Build GRCh38
rs749114664 Chr.16: 89281572-89281572 on Build GRCh38
rs147706206 Chr.16: 89281207-89281207 on Build GRCh38
rs866101421 Chr.16: 89281467-89281467 on Build GRCh38
rs778974115 Chr.16: 89281295-89281295 on Build GRCh38
rs376431701 Chr.16: 89281567-89281567 on Build GRCh38
rs764002055 Chr.16: 89281368-89281368 on Build GRCh38
rs752613308 Chr.16: 89281279-89281279 on Build GRCh38
rs966980049 Chr.16: 89281483-89281483 on Build GRCh38
rs938057375 Chr.16: 89281582-89281582 on Build GRCh38
rs377264059 Chr.16: 89281517-89281517 on Build GRCh38
rs369218399 Chr.16: 89281144-89281144 on Build GRCh38
rs764052859 Chr.16: 89281324-89281324 on Build GRCh38
rs146303178 Chr.16: 89281242-89281242 on Build GRCh38
rs946642356 Chr.16: 89281481-89281481 on Build GRCh38
rs779109308 Chr.16: 89281330-89281330 on Build GRCh38
rs745418159 Chr.16: 89281409-89281409 on Build GRCh38
rs551558075 Chr.16: 89281392-89281392 on Build GRCh38
rs775105168 Chr.16: 89281416-89281416 on Build GRCh38
rs749845953 Chr.16: 89281447-89281447 on Build GRCh38
rs751869374 Chr.16: 89281390-89281390 on Build GRCh38
rs750529678 Chr.16: 89281378-89281378 on Build GRCh38
rs748594316 Chr.16: 89281404-89281404 on Build GRCh38
rs1051158762 Chr.16: 89281271-89281271 on Build GRCh38
rs771739210 Chr.16: 89281363-89281363 on Build GRCh38
rs142332018 Chr.16: 89281226-89281226 on Build GRCh38
rs776489753 Chr.16: 89281422-89281422 on Build GRCh38
rs781519117 Chr.16: 89281352-89281352 on Build GRCh38
rs761081410 Chr.16: 89281490-89281490 on Build GRCh38
rs762258920 Chr.16: 89281559-89281559 on Build GRCh38
rs762398457 Chr.16: 89281311-89281311 on Build GRCh38
rs778242103 Chr.16: 89281408-89281408 on Build GRCh38
rs751326964 Chr.16: 89281246-89281246 on Build GRCh38
rs750932626 Chr.16: 89281564-89281564 on Build GRCh38
rs986683756 Chr.16: 89281548-89281548 on Build GRCh38
rs769960740 Chr.16: 89281135-89281135 on Build GRCh38
rs771612732 Chr.16: 89281415-89281415 on Build GRCh38
rs770331336 Chr.16: 89281355-89281355 on Build GRCh38
rs759616625 Chr.16: 89281484-89281484 on Build GRCh38
rs776477817 Chr.16: 89281217-89281217 on Build GRCh38
rs777649520 Chr.16: 89281569-89281569 on Build GRCh38
rs1057518578 Chr.16: 89281511-89281512 on Build GRCh38
rs1043713257 Chr.16: 89281420-89281420 on Build GRCh38
rs367764283 Chr.16: 89281268-89281268 on Build GRCh38
rs760034709 Chr.16: 89281163-89281163 on Build GRCh38
rs764951610 Chr.16: 89281494-89281494 on Build GRCh38
rs377219970 Chr.16: 89281275-89281275 on Build GRCh38
rs776244135 Chr.16: 89281172-89281172 on Build GRCh38
rs549029103 Chr.16: 89281127-89281127 on Build GRCh38
rs938676909 Chr.16: 89281141-89281141 on Build GRCh38
rs764834945 Chr.16: 89281235-89281235 on Build GRCh38
rs769244635 Chr.16: 89281349-89281349 on Build GRCh38
rs759287339 Chr.16: 89281317-89281317 on Build GRCh38
rs766131389 Chr.16: 89281187-89281187 on Build GRCh38
rs764364528 Chr.16: 89281150-89281152 on Build GRCh38
rs772552969 Chr.16: 89281301-89281301 on Build GRCh38
rs745853521 Chr.16: 89281581-89281581 on Build GRCh38
rs267604681 Chr.16: 89281369-89281369 on Build GRCh38
rs371566102 Chr.16: 89281197-89281197 on Build GRCh38
rs373208578 Chr.16: 89281439-89281439 on Build GRCh38
rs760434985 Chr.16: 89281367-89281367 on Build GRCh38
rs750086920 Chr.16: 89281183-89281183 on Build GRCh38
rs768334377 Chr.16: 89281421-89281421 on Build GRCh38
rs139814582 Chr.16: 89281552-89281552 on Build GRCh38
rs768481687 Chr.16: 89281130-89281130 on Build GRCh38
rs368667754 Chr.16: 89281357-89281357 on Build GRCh38
rs565890803 Chr.16: 89281580-89281580 on Build GRCh38
rs757768089 Chr.16: 89281495-89281495 on Build GRCh38
rs376034086 Chr.16: 89281473-89281473 on Build GRCh38
rs371204323 Chr.16: 89281419-89281419 on Build GRCh38
rs149420132 Chr.16: 89281553-89281553 on Build GRCh38
rs368985397 Chr.16: 89281323-89281323 on Build GRCh38
rs758643160 Chr.16: 89281379-89281379 on Build GRCh38
rs771309106 Chr.16: 89281162-89281162 on Build GRCh38
rs1035852189 Chr.16: 89281280-89281280 on Build GRCh38
rs759118611 Chr.16: 89281358-89281358 on Build GRCh38
rs149098783 Chr.16: 89281322-89281322 on Build GRCh38
rs200783129 Chr.16: 89281364-89281364 on Build GRCh38
rs767934146 Chr.16: 89281164-89281164 on Build GRCh38
rs781188924 Chr.16: 89281132-89281132 on Build GRCh38
rs753707381 Chr.16: 89281371-89281371 on Build GRCh38
rs561668093 Chr.16: 89281305-89281305 on Build GRCh38
rs139057694 Chr.16: 89281274-89281274 on Build GRCh38
rs746251878 Chr.16: 89281220-89281220 on Build GRCh38
rs551427181 Chr.16: 89281423-89281423 on Build GRCh38
rs955466792 Chr.16: 89281529-89281529 on Build GRCh38
rs1048947362 Chr.16: 89281176-89281176 on Build GRCh38
rs754486992 Chr.16: 89281566-89281566 on Build GRCh38
rs749465194 Chr.16: 89281214-89281214 on Build GRCh38
rs776119333 Chr.16: 89281227-89281227 on Build GRCh38
rs778316360 Chr.16: 89281443-89281443 on Build GRCh38
rs200482809 Chr.16: 89281375-89281375 on Build GRCh38
rs749167592 Chr.16: 89281533-89281533 on Build GRCh38
rs751093305 Chr.16: 89281500-89281500 on Build GRCh38
rs755829951 Chr.16: 89281568-89281568 on Build GRCh38
rs779607218 Chr.16: 89281461-89281461 on Build GRCh38
rs1013967207 Chr.16: 89281198-89281198 on Build GRCh38
rs145931801 Chr.16: 89281550-89281550 on Build GRCh38
rs567432230 Chr.16: 89281346-89281346 on Build GRCh38
rs977964461 Chr.16: 89281278-89281278 on Build GRCh38
rs765968892 Chr.16: 89281245-89281245 on Build GRCh38
rs748247992 Chr.16: 89281133-89281133 on Build GRCh38
rs958706615 Chr.16: 89281178-89281178 on Build GRCh38
rs770511968 Chr.16: 89281312-89281312 on Build GRCh38
rs774573314 Chr.16: 89281477-89281477 on Build GRCh38
rs746598883 Chr.16: 89281463-89281463 on Build GRCh38
rs756644111 Chr.16: 89281406-89281406 on Build GRCh38
rs780632720 Chr.16: 89281303-89281303 on Build GRCh38
rs763092437 Chr.16: 89281426-89281426 on Build GRCh38
rs756106781 Chr.16: 89281282-89281282 on Build GRCh38
rs141292414 Chr.16: 89281430-89281430 on Build GRCh38
rs531636092 Chr.16: 89281436-89281436 on Build GRCh38
rs769299642 Chr.16: 89281308-89281308 on Build GRCh38
rs780866636 Chr.16: 89281515-89281515 on Build GRCh38
rs755812080 Chr.16: 89281508-89281508 on Build GRCh38
rs751323481 Chr.16: 89281191-89281191 on Build GRCh38
rs781661922 Chr.16: 89281402-89281402 on Build GRCh38
rs772853156 Chr.16: 89281237-89281237 on Build GRCh38
rs866609551 Chr.16: 89281203-89281203 on Build GRCh38
rs754695851 Chr.16: 89281259-89281259 on Build GRCh38
rs541046005 Chr.16: 89281321-89281321 on Build GRCh38
rs764889984 Chr.16: 89281175-89281175 on Build GRCh38
rs201278036 Chr.16: 89281291-89281291 on Build GRCh38
rs761323554 Chr.16: 89281231-89281231 on Build GRCh38
rs192176841 Chr.16: 89281469-89281469 on Build GRCh38
rs931266122 Chr.16: 89281310-89281310 on Build GRCh38
rs772727317 Chr.16: 89281309-89281309 on Build GRCh38
rs373155396 Chr.16: 89281331-89281331 on Build GRCh38
rs878921394 Chr.16: 89281285-89281285 on Build GRCh38
rs202126162 Chr.16: 89281292-89281292 on Build GRCh38
rs879065365 Chr.16: 89281200-89281200 on Build GRCh38
rs752906592 Chr.16: 89281489-89281489 on Build GRCh38
rs780508146 Chr.16: 89281345-89281345 on Build GRCh38
rs151288302 Chr.16: 89281129-89281129 on Build GRCh38
rs763039848 Chr.16: 89281470-89281470 on Build GRCh38
rs75362060 Chr.16: 89281204-89281204 on Build GRCh38
rs559832262 Chr.16: 89281338-89281338 on Build GRCh38
rs777776761 Chr.16: 89281288-89281288 on Build GRCh38
rs767816242 Chr.16: 89281487-89281487 on Build GRCh38
rs559618415 Chr.16: 89281219-89281219 on Build GRCh38
rs1021246983 Chr.16: 89281479-89281479 on Build GRCh38
rs939808667 Chr.16: 89281199-89281199 on Build GRCh38
rs776241652 Chr.16: 89281466-89281466 on Build GRCh38

View Full Product Details