Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89560968-89561472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL13
Entrez Gene ID
6137
Gene Name
ribosomal protein L13
Gene Aliases
BBC1, D16S444E, D16S44E, L13
UniGene
Hs.410817
Gene Chromosome Location
Chr.16: 89560657-89566829 on Build GRCh38
Transcripts
NM_000977.3 NM_001243130.1 NM_001243131.1 NM_033251.2 NR_002450.1

Gene Symbol
SNORD68
Entrez Gene ID
606500
Gene Name
small nucleolar RNA, C/D box 68
Gene Aliases
HBII-202
UniGene
Gene Chromosome Location
Chr.16: 89561430-89561501 on Build GRCh38
Transcripts
NM_000977.3 NM_001243130.1 NM_001243131.1 NM_033251.2 NR_002450.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6404963 c.214G>A p.A72T Chr.16: 89561336-89561336 on Build GRCh38
1380535 c.93T>G p.R31R Chr.16: 89561052-89561052 on Build GRCh38
3988730 c.58C>T p.R20W Chr.16: 89561017-89561017 on Build GRCh38
6367165 c.166C>T p.R56W Chr.16: 89561288-89561288 on Build GRCh38
5517485 c.123C>T p.A41A Chr.16: 89561245-89561245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572123645 Chr.16: 89561115-89561115 on Build GRCh38
rs1023157873 Chr.16: 89561127-89561127 on Build GRCh38
rs773644615 Chr.16: 89561256-89561256 on Build GRCh38
rs879248154 Chr.16: 89561223-89561223 on Build GRCh38
rs780661827 Chr.16: 89561062-89561062 on Build GRCh38
rs768961733 Chr.16: 89561211-89561211 on Build GRCh38
rs779965054 Chr.16: 89561311-89561311 on Build GRCh38
rs376324664 Chr.16: 89561123-89561123 on Build GRCh38
rs1021153745 Chr.16: 89561257-89561257 on Build GRCh38
rs878933268 Chr.16: 89561222-89561222 on Build GRCh38
rs773626619 Chr.16: 89561349-89561349 on Build GRCh38
rs1054695979 Chr.16: 89561395-89561395 on Build GRCh38
rs770159318 Chr.16: 89561255-89561255 on Build GRCh38
rs957018955 Chr.16: 89561249-89561249 on Build GRCh38
rs146770775 Chr.16: 89561010-89561010 on Build GRCh38
rs776328297 Chr.16: 89561431-89561431 on Build GRCh38
rs199943964 Chr.16: 89561390-89561390 on Build GRCh38
rs771208542 Chr.16: 89561423-89561423 on Build GRCh38
rs140336108 Chr.16: 89561021-89561021 on Build GRCh38
rs11546502 Chr.16: 89561237-89561237 on Build GRCh38
rs760130131 Chr.16: 89561272-89561272 on Build GRCh38
rs752289365 Chr.16: 89561113-89561113 on Build GRCh38
rs571444601 Chr.16: 89561215-89561216 on Build GRCh38
rs1034935838 Chr.16: 89561230-89561230 on Build GRCh38
rs548579412 Chr.16: 89561170-89561170 on Build GRCh38
rs768266156 Chr.16: 89561426-89561426 on Build GRCh38
rs764522448 Chr.16: 89561043-89561043 on Build GRCh38
rs750370803 Chr.16: 89561305-89561305 on Build GRCh38
rs375069797 Chr.16: 89561352-89561352 on Build GRCh38
rs368371275 Chr.16: 89561376-89561376 on Build GRCh38
rs11546503 Chr.16: 89561354-89561354 on Build GRCh38
rs174035 Chr.16: 89561263-89561263 on Build GRCh38
rs781335060 Chr.16: 89561333-89561333 on Build GRCh38
rs755800828 Chr.16: 89561070-89561070 on Build GRCh38
rs537803483 Chr.16: 89561038-89561038 on Build GRCh38
rs371775201 Chr.16: 89561425-89561425 on Build GRCh38
rs545638996 Chr.16: 89561414-89561414 on Build GRCh38
rs763749740 Chr.16: 89561273-89561273 on Build GRCh38
rs1023501448 Chr.16: 89561040-89561040 on Build GRCh38
rs527849412 Chr.16: 89561195-89561195 on Build GRCh38
rs574534079 Chr.16: 89561093-89561093 on Build GRCh38
rs367674578 Chr.16: 89561404-89561404 on Build GRCh38
rs755034474 Chr.16: 89561320-89561320 on Build GRCh38
rs994782584 Chr.16: 89561169-89561169 on Build GRCh38
rs76917615 Chr.16: 89561427-89561427 on Build GRCh38
rs761525924 Chr.16: 89561433-89561433 on Build GRCh38
rs377049048 Chr.16: 89561159-89561159 on Build GRCh38
rs867542380 Chr.16: 89561326-89561326 on Build GRCh38
rs535468689 Chr.16: 89561362-89561362 on Build GRCh38
rs1003342248 Chr.16: 89561229-89561229 on Build GRCh38
rs962244148 Chr.16: 89561245-89561245 on Build GRCh38
rs766927697 Chr.16: 89561262-89561262 on Build GRCh38
rs989679393 Chr.16: 89561119-89561119 on Build GRCh38
rs570406920 Chr.16: 89561205-89561205 on Build GRCh38
rs760160948 Chr.16: 89561396-89561396 on Build GRCh38
rs754293506 Chr.16: 89561046-89561046 on Build GRCh38
rs779407112 Chr.16: 89561424-89561424 on Build GRCh38
rs761614064 Chr.16: 89561391-89561391 on Build GRCh38
rs370017335 Chr.16: 89561020-89561020 on Build GRCh38
rs994688653 Chr.16: 89561049-89561049 on Build GRCh38
rs903407054 Chr.16: 89561161-89561161 on Build GRCh38
rs930419568 Chr.16: 89561130-89561130 on Build GRCh38
rs11548711 Chr.16: 89561367-89561367 on Build GRCh38
rs1050118848 Chr.16: 89561139-89561139 on Build GRCh38
rs1032669069 Chr.16: 89561231-89561231 on Build GRCh38
rs373447378 Chr.16: 89561051-89561051 on Build GRCh38
rs560428678 Chr.16: 89561114-89561114 on Build GRCh38
rs376321802 Chr.16: 89561450-89561450 on Build GRCh38
rs371748985 Chr.16: 89561412-89561412 on Build GRCh38
rs765100472 Chr.16: 89561394-89561394 on Build GRCh38
rs772288690 Chr.16: 89561086-89561086 on Build GRCh38
rs760169834 Chr.16: 89561363-89561363 on Build GRCh38
rs760356367 Chr.16: 89561039-89561039 on Build GRCh38
rs879051217 Chr.16: 89561372-89561372 on Build GRCh38
rs961743809 Chr.16: 89561090-89561090 on Build GRCh38
rs199905422 Chr.16: 89561389-89561389 on Build GRCh38
rs751466278 Chr.16: 89561411-89561411 on Build GRCh38
rs781318388 Chr.16: 89561413-89561413 on Build GRCh38
rs763521734 Chr.16: 89561261-89561261 on Build GRCh38
rs757043374 Chr.16: 89561285-89561285 on Build GRCh38
rs372835705 Chr.16: 89561071-89561071 on Build GRCh38
rs1018702323 Chr.16: 89561202-89561202 on Build GRCh38
rs985878275 Chr.16: 89561287-89561287 on Build GRCh38
rs2280370 Chr.16: 89561052-89561052 on Build GRCh38
rs371172227 Chr.16: 89561177-89561177 on Build GRCh38
rs530071153 Chr.16: 89561162-89561162 on Build GRCh38
rs758409828 Chr.16: 89561307-89561307 on Build GRCh38
rs748375059 Chr.16: 89561336-89561336 on Build GRCh38
rs969244368 Chr.16: 89561218-89561218 on Build GRCh38
rs749228635 Chr.16: 89561078-89561078 on Build GRCh38
rs893123194 Chr.16: 89561197-89561197 on Build GRCh38
rs910761287 Chr.16: 89561144-89561144 on Build GRCh38
rs367902498 Chr.16: 89561417-89561417 on Build GRCh38
rs565200586 Chr.16: 89561125-89561125 on Build GRCh38
rs766094129 Chr.16: 89561446-89561446 on Build GRCh38
rs1059175 Chr.16: 89561003-89561003 on Build GRCh38
rs376294933 Chr.16: 89561396-89561396 on Build GRCh38
rs924902783 Chr.16: 89561385-89561385 on Build GRCh38
rs769530919 Chr.16: 89561437-89561437 on Build GRCh38
rs370274734 Chr.16: 89561059-89561059 on Build GRCh38
rs747177382 Chr.16: 89561200-89561200 on Build GRCh38
rs765190138 Chr.16: 89561288-89561288 on Build GRCh38
rs990905539 Chr.16: 89561323-89561323 on Build GRCh38
rs542134202 Chr.16: 89561112-89561112 on Build GRCh38
rs765694106 Chr.16: 89561097-89561097 on Build GRCh38
rs762687755 Chr.16: 89561402-89561402 on Build GRCh38
rs780237060 Chr.16: 89561198-89561198 on Build GRCh38
rs552749278 Chr.16: 89561204-89561204 on Build GRCh38
rs756380843 Chr.16: 89561416-89561416 on Build GRCh38
rs762752798 Chr.16: 89561441-89561441 on Build GRCh38
rs747072649 Chr.16: 89561319-89561319 on Build GRCh38
rs762277388 Chr.16: 89561094-89561094 on Build GRCh38
rs561209226 Chr.16: 89561142-89561143 on Build GRCh38
rs1010753405 Chr.16: 89561022-89561022 on Build GRCh38
rs544152230 Chr.16: 89561148-89561148 on Build GRCh38
rs745801659 Chr.16: 89561191-89561191 on Build GRCh38
rs1023051922 Chr.16: 89561213-89561213 on Build GRCh38
rs556332790 Chr.16: 89561267-89561267 on Build GRCh38
rs73264924 Chr.16: 89561145-89561145 on Build GRCh38
rs745891520 Chr.16: 89561085-89561085 on Build GRCh38
rs779021877 Chr.16: 89561082-89561082 on Build GRCh38
rs1055509728 Chr.16: 89561152-89561152 on Build GRCh38
rs772762714 Chr.16: 89561439-89561439 on Build GRCh38
rs564290531 Chr.16: 89561281-89561281 on Build GRCh38
rs1047687822 Chr.16: 89561380-89561380 on Build GRCh38
rs771029382 Chr.16: 89561079-89561079 on Build GRCh38
rs752410356 Chr.16: 89561069-89561069 on Build GRCh38
rs910269617 Chr.16: 89561300-89561300 on Build GRCh38
rs11546505 Chr.16: 89561044-89561044 on Build GRCh38
rs11546497 Chr.16: 89561234-89561234 on Build GRCh38
rs774634578 Chr.16: 89560995-89560995 on Build GRCh38
rs78090204 Chr.16: 89561430-89561430 on Build GRCh38
rs757136774 Chr.16: 89561187-89561187 on Build GRCh38
rs758990036 Chr.16: 89561100-89561100 on Build GRCh38
rs11546499 Chr.16: 89561271-89561271 on Build GRCh38
rs141550848 Chr.16: 89561347-89561347 on Build GRCh38
rs752144883 Chr.16: 89561078-89561079 on Build GRCh38
rs775507423 Chr.16: 89561089-89561089 on Build GRCh38
rs776869972 Chr.16: 89561228-89561228 on Build GRCh38

View Full Product Details