Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89561489-89561998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL13
Entrez Gene ID
6137
Gene Name
ribosomal protein L13
Gene Aliases
BBC1, D16S444E, D16S44E, L13
UniGene
Hs.410817
Gene Chromosome Location
Chr.16: 89560657-89566829 on Build GRCh38
Transcripts
NM_000977.3 NM_001243130.1 NM_001243131.1 NM_033251.2 NR_002450.1

Gene Symbol
SNORD68
Entrez Gene ID
606500
Gene Name
small nucleolar RNA, C/D box 68
Gene Aliases
HBII-202
UniGene
Gene Chromosome Location
Chr.16: 89561430-89561501 on Build GRCh38
Transcripts
NM_000977.3 NM_001243130.1 NM_001243131.1 NM_033251.2 NR_002450.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5651307 c.357G>C p.E119D Chr.16: 89561688-89561688 on Build GRCh38
6839178 c.419C>T p.S140F Chr.16: 89561750-89561750 on Build GRCh38
5676034 c.274C>T p.R92W Chr.16: 89561605-89561605 on Build GRCh38
1380536 c.298C>A p.P100T Chr.16: 89561629-89561629 on Build GRCh38
6769164 c.416G>A p.S139N Chr.16: 89561747-89561747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553737668 Chr.16: 89561760-89561760 on Build GRCh38
rs775975084 Chr.16: 89561792-89561792 on Build GRCh38
rs1044392998 Chr.16: 89561563-89561563 on Build GRCh38
rs901550710 Chr.16: 89561885-89561885 on Build GRCh38
rs906902162 Chr.16: 89561859-89561859 on Build GRCh38
rs775112440 Chr.16: 89561730-89561730 on Build GRCh38
rs770533708 Chr.16: 89561538-89561538 on Build GRCh38
rs748184221 Chr.16: 89561715-89561715 on Build GRCh38
rs761982775 Chr.16: 89561559-89561559 on Build GRCh38
rs150117825 Chr.16: 89561636-89561636 on Build GRCh38
rs370584829 Chr.16: 89561546-89561546 on Build GRCh38
rs144100837 Chr.16: 89561733-89561733 on Build GRCh38
rs371100236 Chr.16: 89561565-89561565 on Build GRCh38
rs773475885 Chr.16: 89561724-89561724 on Build GRCh38
rs779871150 Chr.16: 89561604-89561604 on Build GRCh38
rs914335975 Chr.16: 89561829-89561829 on Build GRCh38
rs952083645 Chr.16: 89561732-89561732 on Build GRCh38
rs1048859693 Chr.16: 89561853-89561853 on Build GRCh38
rs750052874 Chr.16: 89561680-89561680 on Build GRCh38
rs1021821040 Chr.16: 89561925-89561925 on Build GRCh38
rs748003385 Chr.16: 89561764-89561764 on Build GRCh38
rs1012434791 Chr.16: 89561931-89561931 on Build GRCh38
rs3751691 Chr.16: 89561973-89561973 on Build GRCh38
rs765604668 Chr.16: 89561526-89561526 on Build GRCh38
rs760971239 Chr.16: 89561798-89561798 on Build GRCh38
rs887598824 Chr.16: 89561856-89561856 on Build GRCh38
rs766830590 Chr.16: 89561639-89561639 on Build GRCh38
rs767866411 Chr.16: 89561658-89561658 on Build GRCh38
rs747523671 Chr.16: 89561529-89561529 on Build GRCh38
rs1002510848 Chr.16: 89561881-89561881 on Build GRCh38
rs769750166 Chr.16: 89561722-89561722 on Build GRCh38
rs1012170364 Chr.16: 89561914-89561914 on Build GRCh38
rs975844443 Chr.16: 89561703-89561703 on Build GRCh38
rs568180302 Chr.16: 89561695-89561695 on Build GRCh38
rs745583340 Chr.16: 89561602-89561602 on Build GRCh38
rs746172851 Chr.16: 89561786-89561786 on Build GRCh38
rs761740327 Chr.16: 89561630-89561630 on Build GRCh38
rs761022115 Chr.16: 89561841-89561841 on Build GRCh38
rs746915628 Chr.16: 89561605-89561605 on Build GRCh38
rs771837591 Chr.16: 89561603-89561603 on Build GRCh38
rs750713568 Chr.16: 89561527-89561527 on Build GRCh38
rs956410912 Chr.16: 89561735-89561735 on Build GRCh38
rs11546501 Chr.16: 89561694-89561694 on Build GRCh38
rs761914975 Chr.16: 89561960-89561960 on Build GRCh38
rs780925358 Chr.16: 89561706-89561706 on Build GRCh38
rs1054192201 Chr.16: 89561822-89561822 on Build GRCh38
rs938775554 Chr.16: 89561572-89561572 on Build GRCh38
rs993312271 Chr.16: 89561638-89561638 on Build GRCh38
rs759979623 Chr.16: 89561642-89561642 on Build GRCh38
rs376881592 Chr.16: 89561776-89561776 on Build GRCh38
rs756818063 Chr.16: 89561595-89561595 on Build GRCh38
rs376241207 Chr.16: 89561541-89561541 on Build GRCh38
rs775398146 Chr.16: 89561545-89561545 on Build GRCh38
rs764546200 Chr.16: 89561802-89561802 on Build GRCh38
rs761287889 Chr.16: 89561741-89561741 on Build GRCh38
rs970193135 Chr.16: 89561933-89561933 on Build GRCh38
rs372620293 Chr.16: 89561766-89561766 on Build GRCh38
rs117042786 Chr.16: 89561759-89561759 on Build GRCh38
rs764206171 Chr.16: 89561909-89561909 on Build GRCh38
rs768436769 Chr.16: 89561617-89561617 on Build GRCh38
rs771241892 Chr.16: 89561729-89561729 on Build GRCh38
rs777308740 Chr.16: 89561587-89561587 on Build GRCh38
rs767933386 Chr.16: 89561737-89561737 on Build GRCh38
rs772296058 Chr.16: 89561791-89561791 on Build GRCh38
rs754028917 Chr.16: 89561932-89561932 on Build GRCh38
rs945827782 Chr.16: 89561850-89561850 on Build GRCh38
rs952174900 Chr.16: 89561944-89561946 on Build GRCh38
rs997155725 Chr.16: 89561898-89561898 on Build GRCh38
rs1027856568 Chr.16: 89561936-89561936 on Build GRCh38
rs749889303 Chr.16: 89561754-89561754 on Build GRCh38
rs955791194 Chr.16: 89561911-89561911 on Build GRCh38
rs9930567 Chr.16: 89561665-89561665 on Build GRCh38
rs753250021 Chr.16: 89561590-89561590 on Build GRCh38
rs990293728 Chr.16: 89561750-89561750 on Build GRCh38
rs147112593 Chr.16: 89561660-89561660 on Build GRCh38
rs754075884 Chr.16: 89561523-89561523 on Build GRCh38
rs1038925075 Chr.16: 89561884-89561884 on Build GRCh38
rs776651716 Chr.16: 89561618-89561618 on Build GRCh38
rs754716275 Chr.16: 89561762-89561762 on Build GRCh38
rs767406624 Chr.16: 89561516-89561516 on Build GRCh38
rs200606302 Chr.16: 89561525-89561525 on Build GRCh38
rs929150646 Chr.16: 89561831-89561831 on Build GRCh38
rs752100648 Chr.16: 89561583-89561583 on Build GRCh38
rs764707113 Chr.16: 89561745-89561745 on Build GRCh38
rs766918538 Chr.16: 89561582-89561582 on Build GRCh38
rs774653132 Chr.16: 89561777-89561777 on Build GRCh38
rs201874909 Chr.16: 89561585-89561585 on Build GRCh38
rs527994266 Chr.16: 89561528-89561528 on Build GRCh38
rs1007455375 Chr.16: 89561880-89561880 on Build GRCh38
rs770496322 Chr.16: 89561551-89561551 on Build GRCh38
rs751238922 Chr.16: 89561761-89561761 on Build GRCh38
rs781107883 Chr.16: 89561763-89561763 on Build GRCh38
rs758769626 Chr.16: 89561578-89561578 on Build GRCh38
rs763239166 Chr.16: 89561726-89561726 on Build GRCh38
rs145267447 Chr.16: 89561956-89561956 on Build GRCh38
rs753495582 Chr.16: 89561556-89561557 on Build GRCh38
rs777872544 Chr.16: 89561773-89561773 on Build GRCh38
rs778448451 Chr.16: 89561667-89561667 on Build GRCh38
rs965665092 Chr.16: 89561691-89561691 on Build GRCh38
rs758084824 Chr.16: 89561683-89561683 on Build GRCh38
rs542224537 Chr.16: 89561519-89561519 on Build GRCh38
rs759441527 Chr.16: 89561514-89561514 on Build GRCh38
rs777209348 Chr.16: 89561531-89561531 on Build GRCh38
rs138372665 Chr.16: 89561640-89561640 on Build GRCh38
rs372886044 Chr.16: 89561561-89561561 on Build GRCh38
rs980386883 Chr.16: 89561947-89561947 on Build GRCh38
rs184644131 Chr.16: 89561520-89561520 on Build GRCh38
rs931817006 Chr.16: 89561803-89561803 on Build GRCh38
rs753143699 Chr.16: 89561739-89561739 on Build GRCh38
rs990840880 Chr.16: 89561547-89561547 on Build GRCh38
rs35249649 Chr.16: 89561621-89561622 on Build GRCh38
rs748950826 Chr.16: 89561536-89561536 on Build GRCh38
rs11266880 Chr.16: 89561598-89561598 on Build GRCh38
rs867557543 Chr.16: 89561861-89561861 on Build GRCh38
rs371877236 Chr.16: 89561687-89561687 on Build GRCh38
rs763325864 Chr.16: 89561637-89561637 on Build GRCh38
rs565789678 Chr.16: 89561805-89561805 on Build GRCh38
rs918641815 Chr.16: 89561813-89561813 on Build GRCh38
rs369616655 Chr.16: 89561609-89561609 on Build GRCh38
rs906855100 Chr.16: 89561622-89561622 on Build GRCh38
rs759497289 Chr.16: 89561635-89561635 on Build GRCh38
rs776612321 Chr.16: 89561555-89561555 on Build GRCh38
rs1037532398 Chr.16: 89561788-89561788 on Build GRCh38
rs771853960 Chr.16: 89561542-89561542 on Build GRCh38
rs370011382 Chr.16: 89561701-89561701 on Build GRCh38

View Full Product Details