Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89564396-89564907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL13
Entrez Gene ID
6137
Gene Name
ribosomal protein L13
Gene Aliases
BBC1, D16S444E, D16S44E, L13
UniGene
Hs.410817
Gene Chromosome Location
Chr.16: 89560657-89566829 on Build GRCh38
Transcripts
NM_000977.3 NM_001243130.1 NM_001243131.1 NM_033251.2

SNP Detail

SNP ID Chromosome Location
rs1040878415 Chr.16: 89564589-89564589 on Build GRCh38
rs1019786213 Chr.16: 89564494-89564494 on Build GRCh38
rs1047465362 Chr.16: 89564581-89564581 on Build GRCh38
rs554723877 Chr.16: 89564784-89564812 on Build GRCh38
rs1040041907 Chr.16: 89564737-89564737 on Build GRCh38
rs570442541 Chr.16: 89564607-89564607 on Build GRCh38
rs374826899 Chr.16: 89564843-89564843 on Build GRCh38
rs920800624 Chr.16: 89564702-89564702 on Build GRCh38
rs944500345 Chr.16: 89564828-89564828 on Build GRCh38
rs553927482 Chr.16: 89564768-89564768 on Build GRCh38
rs906374394 Chr.16: 89564440-89564440 on Build GRCh38
rs886161198 Chr.16: 89564714-89564714 on Build GRCh38
rs944686010 Chr.16: 89564588-89564588 on Build GRCh38
rs1047953837 Chr.16: 89564707-89564707 on Build GRCh38
rs894026338 Chr.16: 89564860-89564860 on Build GRCh38
rs34434037 Chr.16: 89564600-89564600 on Build GRCh38
rs1047587139 Chr.16: 89564817-89564817 on Build GRCh38
rs930486304 Chr.16: 89564801-89564801 on Build GRCh38
rs386794125 Chr.16: 89564777-89564778 on Build GRCh38
rs937934243 Chr.16: 89564650-89564650 on Build GRCh38
rs1046340384 Chr.16: 89564427-89564427 on Build GRCh38
rs541611683 Chr.16: 89564767-89564767 on Build GRCh38
rs889125813 Chr.16: 89564571-89564571 on Build GRCh38
rs762571361 Chr.16: 89564799-89564799 on Build GRCh38
rs116468855 Chr.16: 89564508-89564508 on Build GRCh38
rs961798613 Chr.16: 89564662-89564662 on Build GRCh38
rs910697923 Chr.16: 89564877-89564877 on Build GRCh38
rs145066302 Chr.16: 89564570-89564570 on Build GRCh38
rs952204894 Chr.16: 89564721-89564721 on Build GRCh38
rs1001996737 Chr.16: 89564465-89564465 on Build GRCh38
rs989980999 Chr.16: 89564619-89564620 on Build GRCh38
rs1023762305 Chr.16: 89564434-89564434 on Build GRCh38
rs74589409 Chr.16: 89564765-89564765 on Build GRCh38
rs952232401 Chr.16: 89564848-89564848 on Build GRCh38
rs986287965 Chr.16: 89564866-89564866 on Build GRCh38
rs982275665 Chr.16: 89564462-89564462 on Build GRCh38
rs553277379 Chr.16: 89564497-89564497 on Build GRCh38
rs1055043560 Chr.16: 89564668-89564668 on Build GRCh38
rs903238992 Chr.16: 89564596-89564596 on Build GRCh38
rs778914654 Chr.16: 89564618-89564618 on Build GRCh38
rs905078215 Chr.16: 89564793-89564793 on Build GRCh38
rs537863823 Chr.16: 89564636-89564636 on Build GRCh38
rs944717754 Chr.16: 89564766-89564766 on Build GRCh38
rs1021468165 Chr.16: 89564774-89564774 on Build GRCh38
rs986792293 Chr.16: 89564756-89564756 on Build GRCh38
rs945326448 Chr.16: 89564422-89564422 on Build GRCh38
rs906423301 Chr.16: 89564535-89564535 on Build GRCh38
rs994582387 Chr.16: 89564485-89564485 on Build GRCh38
rs570332429 Chr.16: 89564606-89564606 on Build GRCh38
rs142816940 Chr.16: 89564432-89564432 on Build GRCh38
rs373035023 Chr.16: 89564791-89564791 on Build GRCh38
rs567600003 Chr.16: 89564486-89564487 on Build GRCh38
rs386794124 Chr.16: 89564765-89564768 on Build GRCh38
rs1027321926 Chr.16: 89564837-89564837 on Build GRCh38
rs1034715121 Chr.16: 89564827-89564827 on Build GRCh38
rs149952536 Chr.16: 89564548-89564548 on Build GRCh38
rs1040404732 Chr.16: 89564771-89564771 on Build GRCh38
rs556465295 Chr.16: 89564641-89564641 on Build GRCh38
rs1028949736 Chr.16: 89564614-89564614 on Build GRCh38
rs898978093 Chr.16: 89564550-89564550 on Build GRCh38
rs924591471 Chr.16: 89564772-89564772 on Build GRCh38
rs913928081 Chr.16: 89564521-89564521 on Build GRCh38
rs927935313 Chr.16: 89564642-89564642 on Build GRCh38
rs1008589209 Chr.16: 89564733-89564733 on Build GRCh38
rs1008494281 Chr.16: 89564595-89564595 on Build GRCh38
rs567000478 Chr.16: 89564573-89564573 on Build GRCh38
rs544595738 Chr.16: 89564649-89564649 on Build GRCh38
rs950410259 Chr.16: 89564491-89564493 on Build GRCh38
rs150008739 Chr.16: 89564623-89564623 on Build GRCh38
rs116123689 Chr.16: 89564790-89564790 on Build GRCh38
rs1003857385 Chr.16: 89564796-89564796 on Build GRCh38
rs564926098 Chr.16: 89564478-89564478 on Build GRCh38
rs920457026 Chr.16: 89564693-89564693 on Build GRCh38
rs550839162 Chr.16: 89564518-89564518 on Build GRCh38
rs147350517 Chr.16: 89564474-89564474 on Build GRCh38
rs958453071 Chr.16: 89564768-89564769 on Build GRCh38
rs769145561 Chr.16: 89564601-89564601 on Build GRCh38
rs971769557 Chr.16: 89564701-89564701 on Build GRCh38
rs569230626 Chr.16: 89564522-89564522 on Build GRCh38
rs185466988 Chr.16: 89564831-89564831 on Build GRCh38
rs936775871 Chr.16: 89564514-89564514 on Build GRCh38
rs755923102 Chr.16: 89564529-89564529 on Build GRCh38
rs931612489 Chr.16: 89564845-89564845 on Build GRCh38
rs983473236 Chr.16: 89564492-89564492 on Build GRCh38
rs572185274 Chr.16: 89564777-89564777 on Build GRCh38
rs528587314 Chr.16: 89564464-89564464 on Build GRCh38
rs772209030 Chr.16: 89564639-89564639 on Build GRCh38
rs969072052 Chr.16: 89564447-89564447 on Build GRCh38
rs59013041 Chr.16: 89564778-89564778 on Build GRCh38
rs35116344 Chr.16: 89564783-89564783 on Build GRCh38
rs777521137 Chr.16: 89564555-89564555 on Build GRCh38
rs562999768 Chr.16: 89564838-89564838 on Build GRCh38
rs398102501 Chr.16: 89564842-89564842 on Build GRCh38
rs975525063 Chr.16: 89564503-89564503 on Build GRCh38
rs1030865627 Chr.16: 89564603-89564603 on Build GRCh38
rs993239938 Chr.16: 89564836-89564836 on Build GRCh38
rs111657461 Chr.16: 89564840-89564840 on Build GRCh38
rs945365264 Chr.16: 89564526-89564526 on Build GRCh38
rs147392592 Chr.16: 89564537-89564537 on Build GRCh38

View Full Product Details