Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89565865-89566363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL13
Entrez Gene ID
6137
Gene Name
ribosomal protein L13
Gene Aliases
BBC1, D16S444E, D16S44E, L13
UniGene
Hs.410817
Gene Chromosome Location
Chr.16: 89560657-89566829 on Build GRCh38
Transcripts
NM_000977.3 NM_001243130.1 NM_001243131.1 NM_033251.2

SNP Detail

SNP ID Chromosome Location
rs1023967850 Chr.16: 89566191-89566191 on Build GRCh38
rs934125283 Chr.16: 89566144-89566144 on Build GRCh38
rs113678876 Chr.16: 89566171-89566171 on Build GRCh38
rs140189346 Chr.16: 89565920-89565923 on Build GRCh38
rs963170034 Chr.16: 89566273-89566273 on Build GRCh38
rs1020174597 Chr.16: 89566012-89566012 on Build GRCh38
rs12919536 Chr.16: 89566015-89566015 on Build GRCh38
rs540747913 Chr.16: 89566187-89566187 on Build GRCh38
rs377576737 Chr.16: 89566226-89566226 on Build GRCh38
rs921262544 Chr.16: 89566122-89566122 on Build GRCh38
rs576671811 Chr.16: 89566057-89566057 on Build GRCh38
rs879655839 Chr.16: 89565905-89565906 on Build GRCh38
rs909875103 Chr.16: 89566126-89566126 on Build GRCh38
rs374507743 Chr.16: 89566310-89566310 on Build GRCh38
rs1034056324 Chr.16: 89566181-89566181 on Build GRCh38
rs1051637524 Chr.16: 89565992-89565992 on Build GRCh38
rs935964762 Chr.16: 89566232-89566232 on Build GRCh38
rs1003899514 Chr.16: 89566259-89566259 on Build GRCh38
rs145019018 Chr.16: 89565936-89565936 on Build GRCh38
rs917097661 Chr.16: 89566007-89566007 on Build GRCh38
rs890286508 Chr.16: 89565995-89565995 on Build GRCh38
rs979915619 Chr.16: 89566307-89566307 on Build GRCh38
rs890296720 Chr.16: 89566161-89566161 on Build GRCh38
rs1007242287 Chr.16: 89566282-89566282 on Build GRCh38
rs955372976 Chr.16: 89566284-89566284 on Build GRCh38
rs1057066211 Chr.16: 89565997-89565997 on Build GRCh38
rs552797315 Chr.16: 89566322-89566322 on Build GRCh38
rs904471475 Chr.16: 89566172-89566172 on Build GRCh38
rs1010166280 Chr.16: 89566009-89566009 on Build GRCh38
rs1013850886 Chr.16: 89566246-89566246 on Build GRCh38
rs530693285 Chr.16: 89565945-89565945 on Build GRCh38
rs534941203 Chr.16: 89566107-89566107 on Build GRCh38
rs12919518 Chr.16: 89565977-89565977 on Build GRCh38
rs975694239 Chr.16: 89566279-89566279 on Build GRCh38
rs544616388 Chr.16: 89566230-89566230 on Build GRCh38
rs917149437 Chr.16: 89566135-89566135 on Build GRCh38
rs933728880 Chr.16: 89566266-89566266 on Build GRCh38
rs192056302 Chr.16: 89566314-89566314 on Build GRCh38
rs992748762 Chr.16: 89566133-89566133 on Build GRCh38
rs1024714844 Chr.16: 89566216-89566216 on Build GRCh38
rs941372243 Chr.16: 89566120-89566120 on Build GRCh38
rs1051530800 Chr.16: 89566268-89566268 on Build GRCh38
rs77369830 Chr.16: 89565922-89565925 on Build GRCh38
rs945431546 Chr.16: 89566228-89566228 on Build GRCh38
rs899885197 Chr.16: 89565973-89565973 on Build GRCh38
rs890171731 Chr.16: 89566280-89566280 on Build GRCh38
rs963015593 Chr.16: 89566206-89566206 on Build GRCh38
rs184170703 Chr.16: 89566169-89566169 on Build GRCh38
rs1039736368 Chr.16: 89566121-89566121 on Build GRCh38
rs1017137190 Chr.16: 89566265-89566265 on Build GRCh38
rs61432033 Chr.16: 89566330-89566330 on Build GRCh38
rs921282824 Chr.16: 89566213-89566213 on Build GRCh38
rs924586513 Chr.16: 89565990-89565990 on Build GRCh38
rs79424940 Chr.16: 89566329-89566329 on Build GRCh38
rs886772443 Chr.16: 89566326-89566326 on Build GRCh38
rs1010051737 Chr.16: 89566163-89566163 on Build GRCh38
rs986901640 Chr.16: 89566224-89566224 on Build GRCh38
rs188714219 Chr.16: 89566254-89566254 on Build GRCh38
rs1017507193 Chr.16: 89566196-89566196 on Build GRCh38
rs1007696727 Chr.16: 89566095-89566095 on Build GRCh38
rs562245283 Chr.16: 89566193-89566193 on Build GRCh38
rs955062517 Chr.16: 89566077-89566077 on Build GRCh38
rs530040008 Chr.16: 89566038-89566038 on Build GRCh38
rs1024020333 Chr.16: 89566248-89566248 on Build GRCh38
rs140481355 Chr.16: 89566048-89566048 on Build GRCh38
rs138800827 Chr.16: 89566017-89566017 on Build GRCh38
rs1034420268 Chr.16: 89566023-89566023 on Build GRCh38
rs943665756 Chr.16: 89566295-89566295 on Build GRCh38
rs1030500910 Chr.16: 89566077-89566078 on Build GRCh38
rs532068834 Chr.16: 89566042-89566042 on Build GRCh38
rs1036680321 Chr.16: 89566040-89566040 on Build GRCh38
rs116732212 Chr.16: 89566188-89566188 on Build GRCh38
rs1018175160 Chr.16: 89566101-89566101 on Build GRCh38
rs945485271 Chr.16: 89566305-89566305 on Build GRCh38
rs549302901 Chr.16: 89566296-89566296 on Build GRCh38
rs1003927207 Chr.16: 89566331-89566331 on Build GRCh38
rs114427526 Chr.16: 89566192-89566192 on Build GRCh38
rs543949051 Chr.16: 89566106-89566106 on Build GRCh38
rs543948199 Chr.16: 89566186-89566186 on Build GRCh38
rs764106286 Chr.16: 89566058-89566058 on Build GRCh38
rs986829496 Chr.16: 89566288-89566288 on Build GRCh38
rs115147205 Chr.16: 89566167-89566167 on Build GRCh38
rs373914888 Chr.16: 89566056-89566056 on Build GRCh38
rs866403354 Chr.16: 89566309-89566309 on Build GRCh38
rs933942047 Chr.16: 89566223-89566223 on Build GRCh38
rs976062957 Chr.16: 89566211-89566211 on Build GRCh38
rs1044340520 Chr.16: 89566113-89566113 on Build GRCh38
rs941648759 Chr.16: 89566143-89566143 on Build GRCh38
rs910111156 Chr.16: 89566116-89566116 on Build GRCh38
rs12932628 Chr.16: 89566317-89566317 on Build GRCh38
rs547900419 Chr.16: 89566238-89566238 on Build GRCh38
rs111940523 Chr.16: 89566106-89566106 on Build GRCh38
rs993693243 Chr.16: 89565914-89565914 on Build GRCh38
rs909892458 Chr.16: 89565922-89565922 on Build GRCh38
rs1051148745 Chr.16: 89566146-89566146 on Build GRCh38
rs889446122 Chr.16: 89566289-89566289 on Build GRCh38
rs530714698 Chr.16: 89566295-89566295 on Build GRCh38
rs1000505502 Chr.16: 89566178-89566178 on Build GRCh38
rs1050740587 Chr.16: 89566274-89566274 on Build GRCh38
rs796539046 Chr.16: 89566315-89566317 on Build GRCh38
rs969134906 Chr.16: 89565956-89565956 on Build GRCh38
rs934601179 Chr.16: 89565994-89565994 on Build GRCh38
rs567133060 Chr.16: 89566240-89566240 on Build GRCh38
rs189615457 Chr.16: 89566043-89566043 on Build GRCh38
rs1031556944 Chr.16: 89566324-89566324 on Build GRCh38
rs1047294022 Chr.16: 89565910-89565910 on Build GRCh38
rs904354285 Chr.16: 89566231-89566231 on Build GRCh38
rs982883137 Chr.16: 89566195-89566195 on Build GRCh38
rs77770697 Chr.16: 89566315-89566315 on Build GRCh38
rs747878564 Chr.16: 89565919-89565920 on Build GRCh38
rs888175392 Chr.16: 89566031-89566031 on Build GRCh38

View Full Product Details