Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 90022522-90023030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS8
Entrez Gene ID
2622
Gene Name
growth arrest specific 8
Gene Aliases
CILD33, DRC4, GAS11
UniGene
Hs.431792
Gene Chromosome Location
Chr.16: 90019629-90044971 on Build GRCh38
Transcripts
NM_001286205.1 NM_001286208.1 NM_001286209.1 NM_001481.2 XM_005256304.4 XM_005256309.4 XM_006721175.3 XM_011522990.2 XM_011522991.1 XM_011522992.2 XM_017023122.1 XM_017023123.1 XM_017023124.1 XM_017023125.1

SNP Detail

SNP ID Chromosome Location
rs936645266 Chr.16: 90022613-90022613 on Build GRCh38
rs377600781 Chr.16: 90022713-90022713 on Build GRCh38
rs1057484078 Chr.16: 90022722-90022722 on Build GRCh38
rs544667020 Chr.16: 90022597-90022597 on Build GRCh38
rs940832941 Chr.16: 90022856-90022856 on Build GRCh38
rs1046549210 Chr.16: 90022670-90022670 on Build GRCh38
rs953237223 Chr.16: 90022959-90022959 on Build GRCh38
rs577392738 Chr.16: 90022565-90022565 on Build GRCh38
rs74684186 Chr.16: 90022585-90022585 on Build GRCh38
rs778879338 Chr.16: 90022701-90022701 on Build GRCh38
rs373637203 Chr.16: 90022744-90022744 on Build GRCh38
rs977765441 Chr.16: 90022598-90022598 on Build GRCh38
rs796369792 Chr.16: 90022546-90022546 on Build GRCh38
rs987288041 Chr.16: 90022568-90022568 on Build GRCh38
rs747039502 Chr.16: 90022737-90022737 on Build GRCh38
rs775634500 Chr.16: 90022798-90022798 on Build GRCh38
rs112094183 Chr.16: 90022899-90022899 on Build GRCh38
rs376210528 Chr.16: 90022865-90022865 on Build GRCh38
rs925848539 Chr.16: 90022793-90022793 on Build GRCh38
rs1041945431 Chr.16: 90022863-90022863 on Build GRCh38
rs1005115025 Chr.16: 90022693-90022693 on Build GRCh38
rs976563618 Chr.16: 90022691-90022691 on Build GRCh38
rs991402524 Chr.16: 90022920-90022920 on Build GRCh38
rs911747908 Chr.16: 90022589-90022589 on Build GRCh38
rs923929184 Chr.16: 90022607-90022607 on Build GRCh38
rs35989985 Chr.16: 90022601-90022602 on Build GRCh38
rs889387977 Chr.16: 90022996-90022996 on Build GRCh38
rs951713920 Chr.16: 90022749-90022749 on Build GRCh38
rs59443037 Chr.16: 90022927-90022927 on Build GRCh38
rs80287915 Chr.16: 90022715-90022715 on Build GRCh38
rs1046939814 Chr.16: 90022833-90022833 on Build GRCh38
rs1030153694 Chr.16: 90022957-90022957 on Build GRCh38
rs1011983680 Chr.16: 90022908-90022908 on Build GRCh38
rs1050695949 Chr.16: 90022991-90022991 on Build GRCh38
rs371127020 Chr.16: 90022554-90022554 on Build GRCh38
rs986208901 Chr.16: 90022818-90022818 on Build GRCh38
rs770194881 Chr.16: 90022766-90022766 on Build GRCh38
rs1008634738 Chr.16: 90022896-90022896 on Build GRCh38
rs750354117 Chr.16: 90022703-90022703 on Build GRCh38
rs887432311 Chr.16: 90022923-90022923 on Build GRCh38
rs112372427 Chr.16: 90022960-90022960 on Build GRCh38
rs753579631 Chr.16: 90022683-90022683 on Build GRCh38
rs552480254 Chr.16: 90022730-90022730 on Build GRCh38
rs372451257 Chr.16: 90022650-90022650 on Build GRCh38
rs562863688 Chr.16: 90022614-90022614 on Build GRCh38
rs902949538 Chr.16: 90022902-90022902 on Build GRCh38
rs1032814569 Chr.16: 90022907-90022907 on Build GRCh38
rs923349334 Chr.16: 90022692-90022692 on Build GRCh38
rs916612412 Chr.16: 90022827-90022827 on Build GRCh38
rs916623967 Chr.16: 90022616-90022616 on Build GRCh38
rs548867232 Chr.16: 90022669-90022669 on Build GRCh38
rs760268882 Chr.16: 90022679-90022679 on Build GRCh38
rs189240134 Chr.16: 90022895-90022895 on Build GRCh38
rs755183270 Chr.16: 90022758-90022758 on Build GRCh38
rs561120751 Chr.16: 90022948-90022948 on Build GRCh38
rs1023572032 Chr.16: 90022926-90022926 on Build GRCh38
rs948158924 Chr.16: 90022830-90022830 on Build GRCh38
rs141176595 Chr.16: 90022941-90022941 on Build GRCh38
rs1023292011 Chr.16: 90022742-90022742 on Build GRCh38
rs899383583 Chr.16: 90022875-90022875 on Build GRCh38
rs1020381458 Chr.16: 90022577-90022577 on Build GRCh38
rs753612743 Chr.16: 90022892-90022892 on Build GRCh38
rs989403820 Chr.16: 90022799-90022799 on Build GRCh38
rs374122554 Chr.16: 90022835-90022835 on Build GRCh38
rs994519870 Chr.16: 90022547-90022547 on Build GRCh38
rs546551738 Chr.16: 90022699-90022699 on Build GRCh38
rs747122133 Chr.16: 90022628-90022628 on Build GRCh38
rs781404014 Chr.16: 90022760-90022760 on Build GRCh38
rs960745343 Chr.16: 90022637-90022637 on Build GRCh38
rs570554098 Chr.16: 90022750-90022750 on Build GRCh38
rs763762363 Chr.16: 90022681-90022681 on Build GRCh38
rs984732314 Chr.16: 90022674-90022674 on Build GRCh38
rs747093538 Chr.16: 90022985-90022985 on Build GRCh38
rs977293062 Chr.16: 90022790-90022790 on Build GRCh38
rs893352443 Chr.16: 90022567-90022567 on Build GRCh38
rs982155390 Chr.16: 90022928-90022928 on Build GRCh38
rs928001372 Chr.16: 90022933-90022933 on Build GRCh38
rs866245574 Chr.16: 90022632-90022632 on Build GRCh38
rs999664274 Chr.16: 90022617-90022617 on Build GRCh38
rs1028671573 Chr.16: 90022717-90022717 on Build GRCh38
rs763825189 Chr.16: 90022887-90022887 on Build GRCh38
rs537544290 Chr.16: 90022784-90022784 on Build GRCh38
rs1034815603 Chr.16: 90022622-90022622 on Build GRCh38
rs551244363 Chr.16: 90022910-90022910 on Build GRCh38
rs748522706 Chr.16: 90022765-90022765 on Build GRCh38
rs928085395 Chr.16: 90022847-90022847 on Build GRCh38
rs1003078193 Chr.16: 90022716-90022716 on Build GRCh38
rs923119009 Chr.16: 90022800-90022800 on Build GRCh38
rs549405587 Chr.16: 90022825-90022825 on Build GRCh38
rs990461682 Chr.16: 90022640-90022640 on Build GRCh38
rs930865207 Chr.16: 90022965-90022965 on Build GRCh38
rs903381534 Chr.16: 90022594-90022594 on Build GRCh38
rs58490907 Chr.16: 90022905-90022905 on Build GRCh38
rs757018550 Chr.16: 90022696-90022696 on Build GRCh38
rs560595280 Chr.16: 90022712-90022712 on Build GRCh38
rs1050364990 Chr.16: 90022707-90022707 on Build GRCh38
rs972220720 Chr.16: 90022950-90022950 on Build GRCh38
rs953462701 Chr.16: 90022806-90022806 on Build GRCh38

View Full Product Details