Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1744900-1745400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINF2
Entrez Gene ID
5345
Gene Name
serpin family F member 2
Gene Aliases
A2AP, AAP, ALPHA-2-PI, API, PLI
UniGene
Hs.159509
Gene Chromosome Location
Chr.17: 1742808-1755265 on Build GRCh38
Transcripts
NM_000934.3 NM_001165920.1 NM_001165921.1 XM_005256701.4 XM_017024765.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4725083 c.131T>C p.V44A Chr.17: 1745361-1745361 on Build GRCh38
6775663 c.165+2T>C p.? Chr.17: 1745397-1745397 on Build GRCh38
4129625 c.98G>A p.R33Q Chr.17: 1745209-1745209 on Build GRCh38
3691382 c.4G>A p.A2T Chr.17: 1744999-1744999 on Build GRCh38
5468859 c.5C>T p.A2V Chr.17: 1745000-1745000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768970078 Chr.17: 1745232-1745232 on Build GRCh38
rs373323372 Chr.17: 1745158-1745158 on Build GRCh38
rs767986188 Chr.17: 1745240-1745240 on Build GRCh38
rs976043138 Chr.17: 1745050-1745050 on Build GRCh38
rs914849811 Chr.17: 1745112-1745112 on Build GRCh38
rs148022156 Chr.17: 1745190-1745190 on Build GRCh38
rs370280422 Chr.17: 1745055-1745055 on Build GRCh38
rs544596507 Chr.17: 1744927-1744927 on Build GRCh38
rs761481781 Chr.17: 1744979-1744979 on Build GRCh38
rs2070862 Chr.17: 1745000-1745000 on Build GRCh38
rs770737376 Chr.17: 1745248-1745248 on Build GRCh38
rs751614553 Chr.17: 1745016-1745016 on Build GRCh38
rs766407419 Chr.17: 1745008-1745008 on Build GRCh38
rs199686844 Chr.17: 1745218-1745218 on Build GRCh38
rs561282969 Chr.17: 1745329-1745329 on Build GRCh38
rs752826496 Chr.17: 1744924-1744924 on Build GRCh38
rs373572476 Chr.17: 1744943-1744943 on Build GRCh38
rs770683914 Chr.17: 1745314-1745314 on Build GRCh38
rs553501683 Chr.17: 1745254-1745254 on Build GRCh38
rs775841802 Chr.17: 1744972-1744972 on Build GRCh38
rs767370481 Chr.17: 1745031-1745031 on Build GRCh38
rs914737056 Chr.17: 1744922-1744922 on Build GRCh38
rs536866023 Chr.17: 1744923-1744923 on Build GRCh38
rs755522783 Chr.17: 1745303-1745303 on Build GRCh38
rs759505499 Chr.17: 1745161-1745161 on Build GRCh38
rs769466657 Chr.17: 1745142-1745142 on Build GRCh38
rs377325935 Chr.17: 1745148-1745148 on Build GRCh38
rs186870227 Chr.17: 1745209-1745209 on Build GRCh38
rs551505236 Chr.17: 1745081-1745081 on Build GRCh38
rs760662355 Chr.17: 1745255-1745255 on Build GRCh38
rs531196259 Chr.17: 1744925-1744925 on Build GRCh38
rs17852960 Chr.17: 1744999-1744999 on Build GRCh38
rs373385252 Chr.17: 1744956-1744956 on Build GRCh38
rs139885061 Chr.17: 1745347-1745347 on Build GRCh38
rs753693885 Chr.17: 1745282-1745282 on Build GRCh38
rs191703810 Chr.17: 1745283-1745283 on Build GRCh38
rs201339551 Chr.17: 1745328-1745328 on Build GRCh38
rs1009594279 Chr.17: 1745356-1745356 on Build GRCh38
rs764369234 Chr.17: 1745189-1745189 on Build GRCh38
rs747387759 Chr.17: 1745222-1745222 on Build GRCh38
rs771450051 Chr.17: 1744960-1744960 on Build GRCh38
rs34712321 Chr.17: 1744939-1744940 on Build GRCh38
rs560323542 Chr.17: 1745150-1745150 on Build GRCh38
rs765434085 Chr.17: 1745374-1745374 on Build GRCh38
rs775794693 Chr.17: 1745097-1745097 on Build GRCh38
rs887589788 Chr.17: 1744988-1744988 on Build GRCh38
rs369436547 Chr.17: 1745162-1745162 on Build GRCh38
rs376816385 Chr.17: 1745165-1745165 on Build GRCh38
rs542258934 Chr.17: 1745338-1745338 on Build GRCh38
rs185025710 Chr.17: 1745341-1745341 on Build GRCh38
rs758161056 Chr.17: 1745199-1745199 on Build GRCh38
rs746320572 Chr.17: 1745068-1745068 on Build GRCh38
rs944433307 Chr.17: 1745311-1745311 on Build GRCh38
rs773966273 Chr.17: 1745251-1745251 on Build GRCh38
rs753167648 Chr.17: 1745317-1745317 on Build GRCh38
rs369804932 Chr.17: 1745113-1745113 on Build GRCh38
rs1056029442 Chr.17: 1745180-1745180 on Build GRCh38
rs752464909 Chr.17: 1745037-1745037 on Build GRCh38
rs758875204 Chr.17: 1745213-1745213 on Build GRCh38
rs765921034 Chr.17: 1745156-1745156 on Build GRCh38
rs777059312 Chr.17: 1745240-1745240 on Build GRCh38
rs2070863 Chr.17: 1745208-1745208 on Build GRCh38
rs893852610 Chr.17: 1745140-1745140 on Build GRCh38
rs372981803 Chr.17: 1745056-1745056 on Build GRCh38
rs779797130 Chr.17: 1745045-1745045 on Build GRCh38
rs980330383 Chr.17: 1745115-1745115 on Build GRCh38
rs201320264 Chr.17: 1744948-1744948 on Build GRCh38
rs754979455 Chr.17: 1745024-1745024 on Build GRCh38
rs781139539 Chr.17: 1744947-1744947 on Build GRCh38
rs71359127 Chr.17: 1745106-1745106 on Build GRCh38
rs200885603 Chr.17: 1745331-1745331 on Build GRCh38
rs747375857 Chr.17: 1745139-1745139 on Build GRCh38
rs377055860 Chr.17: 1744992-1744992 on Build GRCh38
rs758533164 Chr.17: 1745288-1745288 on Build GRCh38
rs144621260 Chr.17: 1745186-1745186 on Build GRCh38
rs759163354 Chr.17: 1745253-1745253 on Build GRCh38
rs937739448 Chr.17: 1745376-1745376 on Build GRCh38
rs748843459 Chr.17: 1745242-1745242 on Build GRCh38
rs757487763 Chr.17: 1745199-1745199 on Build GRCh38
rs759893506 Chr.17: 1744964-1744964 on Build GRCh38
rs368679826 Chr.17: 1745264-1745264 on Build GRCh38
rs926583692 Chr.17: 1744928-1744928 on Build GRCh38
rs764831217 Chr.17: 1744980-1744980 on Build GRCh38
rs373510687 Chr.17: 1744944-1744944 on Build GRCh38
rs777871727 Chr.17: 1744953-1744953 on Build GRCh38
rs75500393 Chr.17: 1745270-1745270 on Build GRCh38
rs748561939 Chr.17: 1745313-1745313 on Build GRCh38
rs375847980 Chr.17: 1745350-1745350 on Build GRCh38
rs772352419 Chr.17: 1744967-1744967 on Build GRCh38
rs565475587 Chr.17: 1745054-1745054 on Build GRCh38
rs751945657 Chr.17: 1745295-1745295 on Build GRCh38
rs956245824 Chr.17: 1745074-1745074 on Build GRCh38
rs145959786 Chr.17: 1745342-1745342 on Build GRCh38
rs980636917 Chr.17: 1745346-1745346 on Build GRCh38
rs575772412 Chr.17: 1745304-1745304 on Build GRCh38
rs1018100074 Chr.17: 1745027-1745027 on Build GRCh38
rs371084929 Chr.17: 1745196-1745196 on Build GRCh38
rs368253719 Chr.17: 1745378-1745378 on Build GRCh38
rs367681012 Chr.17: 1745285-1745285 on Build GRCh38
rs780607415 Chr.17: 1745293-1745293 on Build GRCh38
rs752512440 Chr.17: 1745177-1745177 on Build GRCh38

View Full Product Details