Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 2417038-2417580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL16
Entrez Gene ID
79066
Gene Name
methyltransferase like 16
Gene Aliases
METT10D
UniGene
Hs.632237
Gene Chromosome Location
Chr.17: 2416049-2511906 on Build GRCh38
Transcripts
NM_024086.3 XM_005256801.2 XM_017025045.1 XM_017025046.1

SNP Detail

SNP ID Chromosome Location
rs537867572 Chr.17: 2417408-2417408 on Build GRCh38
rs890239482 Chr.17: 2417079-2417079 on Build GRCh38
rs982397002 Chr.17: 2417354-2417354 on Build GRCh38
rs962685005 Chr.17: 2417392-2417392 on Build GRCh38
rs1019693499 Chr.17: 2417137-2417137 on Build GRCh38
rs948385647 Chr.17: 2417224-2417224 on Build GRCh38
rs1041374287 Chr.17: 2417301-2417301 on Build GRCh38
rs775275622 Chr.17: 2417128-2417129 on Build GRCh38
rs11868007 Chr.17: 2417250-2417250 on Build GRCh38
rs538327044 Chr.17: 2417240-2417240 on Build GRCh38
rs931132455 Chr.17: 2417403-2417403 on Build GRCh38
rs911726346 Chr.17: 2417531-2417531 on Build GRCh38
rs1040987614 Chr.17: 2417486-2417486 on Build GRCh38
rs558061759 Chr.17: 2417058-2417059 on Build GRCh38
rs974516978 Chr.17: 2417422-2417422 on Build GRCh38
rs562707380 Chr.17: 2417125-2417125 on Build GRCh38
rs1050696424 Chr.17: 2417253-2417253 on Build GRCh38
rs1026925852 Chr.17: 2417082-2417086 on Build GRCh38
rs1002627008 Chr.17: 2417060-2417060 on Build GRCh38
rs1033708547 Chr.17: 2417132-2417132 on Build GRCh38
rs531767090 Chr.17: 2417140-2417140 on Build GRCh38
rs1024384845 Chr.17: 2417179-2417179 on Build GRCh38
rs149731991 Chr.17: 2417261-2417261 on Build GRCh38
rs958220266 Chr.17: 2417194-2417194 on Build GRCh38
rs1006815259 Chr.17: 2417302-2417302 on Build GRCh38
rs989645124 Chr.17: 2417341-2417341 on Build GRCh38
rs985955960 Chr.17: 2417517-2417517 on Build GRCh38
rs989674542 Chr.17: 2417196-2417196 on Build GRCh38
rs752361117 Chr.17: 2417331-2417331 on Build GRCh38
rs973277382 Chr.17: 2417211-2417211 on Build GRCh38
rs995399949 Chr.17: 2417078-2417087 on Build GRCh38
rs944455050 Chr.17: 2417295-2417295 on Build GRCh38
rs763479422 Chr.17: 2417512-2417512 on Build GRCh38
rs938384942 Chr.17: 2417383-2417384 on Build GRCh38
rs192036644 Chr.17: 2417514-2417514 on Build GRCh38
rs11868221 Chr.17: 2417504-2417504 on Build GRCh38
rs902836718 Chr.17: 2417306-2417306 on Build GRCh38
rs941394679 Chr.17: 2417192-2417192 on Build GRCh38
rs866204525 Chr.17: 2417086-2417086 on Build GRCh38
rs547543528 Chr.17: 2417220-2417220 on Build GRCh38
rs1024911705 Chr.17: 2417344-2417344 on Build GRCh38
rs575113382 Chr.17: 2417264-2417264 on Build GRCh38
rs1008531259 Chr.17: 2417084-2417084 on Build GRCh38
rs1048335309 Chr.17: 2417300-2417300 on Build GRCh38
rs867074934 Chr.17: 2417087-2417087 on Build GRCh38
rs969576560 Chr.17: 2417353-2417353 on Build GRCh38
rs901189650 Chr.17: 2417307-2417307 on Build GRCh38
rs1033261025 Chr.17: 2417330-2417330 on Build GRCh38
rs1048218104 Chr.17: 2417416-2417416 on Build GRCh38
rs1006931446 Chr.17: 2417133-2417133 on Build GRCh38
rs568462791 Chr.17: 2417180-2417180 on Build GRCh38
rs967597913 Chr.17: 2417105-2417105 on Build GRCh38
rs931161950 Chr.17: 2417296-2417297 on Build GRCh38
rs867332503 Chr.17: 2417085-2417085 on Build GRCh38
rs867646200 Chr.17: 2417166-2417166 on Build GRCh38
rs920491124 Chr.17: 2417444-2417444 on Build GRCh38
rs762501713 Chr.17: 2417289-2417293 on Build GRCh38
rs942585242 Chr.17: 2417459-2417459 on Build GRCh38
rs1015656965 Chr.17: 2417370-2417370 on Build GRCh38
rs548451664 Chr.17: 2417143-2417143 on Build GRCh38
rs754968119 Chr.17: 2417061-2417062 on Build GRCh38
rs75810416 Chr.17: 2417545-2417545 on Build GRCh38
rs958882198 Chr.17: 2417159-2417159 on Build GRCh38
rs982817581 Chr.17: 2417364-2417364 on Build GRCh38
rs999885680 Chr.17: 2417309-2417309 on Build GRCh38
rs896971760 Chr.17: 2417292-2417292 on Build GRCh38
rs931829145 Chr.17: 2417236-2417236 on Build GRCh38
rs554896936 Chr.17: 2417511-2417511 on Build GRCh38
rs908564420 Chr.17: 2417188-2417188 on Build GRCh38
rs1037236334 Chr.17: 2417072-2417087 on Build GRCh38
rs1013051666 Chr.17: 2417342-2417342 on Build GRCh38
rs999996586 Chr.17: 2417325-2417325 on Build GRCh38
rs918374085 Chr.17: 2417399-2417399 on Build GRCh38
rs571028054 Chr.17: 2417225-2417225 on Build GRCh38
rs1031561050 Chr.17: 2417191-2417191 on Build GRCh38
rs971426373 Chr.17: 2417359-2417359 on Build GRCh38
rs777290662 Chr.17: 2417518-2417518 on Build GRCh38
rs534116851 Chr.17: 2417184-2417184 on Build GRCh38
rs780772070 Chr.17: 2417210-2417210 on Build GRCh38
rs978210830 Chr.17: 2417182-2417182 on Build GRCh38
rs869134753 Chr.17: 2417085-2417086 on Build GRCh38
rs11868006 Chr.17: 2417248-2417248 on Build GRCh38
rs916862695 Chr.17: 2417206-2417206 on Build GRCh38
rs59854056 Chr.17: 2417085-2417085 on Build GRCh38
rs896999871 Chr.17: 2417077-2417077 on Build GRCh38

View Full Product Details