Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3898688-3899188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P2RX1
Entrez Gene ID
5023
Gene Name
purinergic receptor P2X 1
Gene Aliases
P2X1
UniGene
Hs.41735
Gene Chromosome Location
Chr.17: 3896591-3916666 on Build GRCh38
Transcripts
NM_002558.3 XM_006721529.2 XM_011523896.2 XM_011523897.2 XM_011523898.2 XM_011523899.2 XM_011523900.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6080921 c.963C>T p.G321G Chr.17: 3898937-3898937 on Build GRCh38
7284102 c.961G>A p.G321S Chr.17: 3898939-3898939 on Build GRCh38
3701205 c.961G>T p.G321C Chr.17: 3898939-3898939 on Build GRCh38
5386228 c.876G>A p.R292R Chr.17: 3899024-3899024 on Build GRCh38
2838307 c.913C>T p.R305C Chr.17: 3898987-3898987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044019246 Chr.17: 3898779-3898779 on Build GRCh38
rs375360882 Chr.17: 3898994-3898994 on Build GRCh38
rs765817392 Chr.17: 3898740-3898740 on Build GRCh38
rs1035052448 Chr.17: 3898734-3898734 on Build GRCh38
rs554415872 Chr.17: 3898714-3898714 on Build GRCh38
rs757102707 Chr.17: 3899048-3899048 on Build GRCh38
rs548171524 Chr.17: 3899056-3899056 on Build GRCh38
rs770151439 Chr.17: 3898999-3898999 on Build GRCh38
rs964802471 Chr.17: 3899147-3899147 on Build GRCh38
rs748666418 Chr.17: 3898997-3898997 on Build GRCh38
rs376505620 Chr.17: 3899008-3899008 on Build GRCh38
rs745324962 Chr.17: 3898967-3898967 on Build GRCh38
rs976134642 Chr.17: 3898867-3898867 on Build GRCh38
rs776898877 Chr.17: 3898987-3898987 on Build GRCh38
rs201771436 Chr.17: 3899017-3899017 on Build GRCh38
rs972022013 Chr.17: 3898749-3898749 on Build GRCh38
rs752360415 Chr.17: 3898940-3898940 on Build GRCh38
rs1026243987 Chr.17: 3899072-3899072 on Build GRCh38
rs370596965 Chr.17: 3899059-3899059 on Build GRCh38
rs200385031 Chr.17: 3898986-3898986 on Build GRCh38
rs759791296 Chr.17: 3899015-3899015 on Build GRCh38
rs1026131402 Chr.17: 3898750-3898750 on Build GRCh38
rs1017473439 Chr.17: 3899123-3899123 on Build GRCh38
rs771558716 Chr.17: 3898770-3898770 on Build GRCh38
rs766521686 Chr.17: 3898944-3898944 on Build GRCh38
rs960896469 Chr.17: 3898735-3898735 on Build GRCh38
rs567491886 Chr.17: 3899000-3899000 on Build GRCh38
rs146814691 Chr.17: 3899004-3899004 on Build GRCh38
rs766502947 Chr.17: 3899003-3899003 on Build GRCh38
rs143374859 Chr.17: 3898898-3898898 on Build GRCh38
rs753523752 Chr.17: 3899025-3899025 on Build GRCh38
rs894575206 Chr.17: 3899146-3899146 on Build GRCh38
rs755892151 Chr.17: 3898951-3898951 on Build GRCh38
rs763946439 Chr.17: 3898952-3898952 on Build GRCh38
rs933829917 Chr.17: 3898743-3898743 on Build GRCh38
rs913821940 Chr.17: 3898754-3898754 on Build GRCh38
rs1052654607 Chr.17: 3898900-3898900 on Build GRCh38
rs534086275 Chr.17: 3898712-3898712 on Build GRCh38
rs369912006 Chr.17: 3898979-3898979 on Build GRCh38
rs762882710 Chr.17: 3899002-3899002 on Build GRCh38
rs141434587 Chr.17: 3898884-3898884 on Build GRCh38
rs1038210332 Chr.17: 3899061-3899073 on Build GRCh38
rs751227763 Chr.17: 3899066-3899066 on Build GRCh38
rs779792097 Chr.17: 3898977-3898977 on Build GRCh38
rs1002606496 Chr.17: 3898730-3898730 on Build GRCh38
rs763852321 Chr.17: 3899020-3899020 on Build GRCh38
rs113856591 Chr.17: 3898860-3898860 on Build GRCh38
rs559939433 Chr.17: 3898920-3898920 on Build GRCh38
rs201217413 Chr.17: 3898888-3898888 on Build GRCh38
rs540006968 Chr.17: 3898901-3898901 on Build GRCh38
rs757983137 Chr.17: 3898968-3898968 on Build GRCh38
rs892593523 Chr.17: 3898919-3898919 on Build GRCh38
rs887374719 Chr.17: 3898953-3898953 on Build GRCh38
rs377714821 Chr.17: 3899051-3899051 on Build GRCh38
rs754571109 Chr.17: 3898960-3898960 on Build GRCh38
rs556573573 Chr.17: 3898847-3898847 on Build GRCh38
rs185009352 Chr.17: 3898959-3898959 on Build GRCh38
rs200488831 Chr.17: 3898950-3898950 on Build GRCh38
rs750391725 Chr.17: 3899052-3899052 on Build GRCh38
rs753786640 Chr.17: 3898958-3898958 on Build GRCh38
rs528005711 Chr.17: 3899151-3899151 on Build GRCh38
rs139414872 Chr.17: 3898940-3898940 on Build GRCh38
rs984621002 Chr.17: 3899091-3899091 on Build GRCh38
rs773543766 Chr.17: 3898846-3898846 on Build GRCh38
rs769152780 Chr.17: 3898894-3898894 on Build GRCh38
rs755765271 Chr.17: 3898982-3898999 on Build GRCh38
rs539959490 Chr.17: 3898842-3898842 on Build GRCh38
rs557685559 Chr.17: 3899090-3899090 on Build GRCh38
rs144650752 Chr.17: 3898934-3898934 on Build GRCh38
rs945289953 Chr.17: 3898772-3898772 on Build GRCh38
rs374742243 Chr.17: 3898729-3898729 on Build GRCh38

View Full Product Details