Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 5442697-5443207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX33
Entrez Gene ID
56919
Gene Name
DEAH-box helicase 33
Gene Aliases
DDX33
UniGene
Hs.250456
Gene Chromosome Location
Chr.17: 5440912-5469060 on Build GRCh38
Transcripts
NM_001199699.1 NM_020162.3 XM_017024877.1

SNP Detail

SNP ID Chromosome Location
rs773813697 Chr.17: 5443069-5443069 on Build GRCh38
rs564204318 Chr.17: 5443145-5443145 on Build GRCh38
rs973705467 Chr.17: 5442941-5442941 on Build GRCh38
rs901291983 Chr.17: 5443060-5443065 on Build GRCh38
rs77195858 Chr.17: 5443126-5443126 on Build GRCh38
rs1011158889 Chr.17: 5443138-5443138 on Build GRCh38
rs749570359 Chr.17: 5443007-5443007 on Build GRCh38
rs897049558 Chr.17: 5442934-5442934 on Build GRCh38
rs928916028 Chr.17: 5443006-5443006 on Build GRCh38
rs894542863 Chr.17: 5443049-5443049 on Build GRCh38
rs894572312 Chr.17: 5442776-5442776 on Build GRCh38
rs868529429 Chr.17: 5443120-5443120 on Build GRCh38
rs199589198 Chr.17: 5442894-5442894 on Build GRCh38
rs1031361430 Chr.17: 5442875-5442875 on Build GRCh38
rs909104918 Chr.17: 5443129-5443129 on Build GRCh38
rs999294614 Chr.17: 5443128-5443128 on Build GRCh38
rs1021601396 Chr.17: 5442836-5442836 on Build GRCh38
rs62072696 Chr.17: 5442902-5442902 on Build GRCh38
rs990028208 Chr.17: 5442855-5442855 on Build GRCh38
rs770618079 Chr.17: 5443041-5443041 on Build GRCh38
rs1048177770 Chr.17: 5442946-5442946 on Build GRCh38
rs563579293 Chr.17: 5442822-5442822 on Build GRCh38
rs910932272 Chr.17: 5442907-5442907 on Build GRCh38
rs915432698 Chr.17: 5442858-5442858 on Build GRCh38
rs748596480 Chr.17: 5443022-5443024 on Build GRCh38
rs184594316 Chr.17: 5443177-5443177 on Build GRCh38
rs1034389668 Chr.17: 5442737-5442737 on Build GRCh38
rs929654086 Chr.17: 5442961-5442961 on Build GRCh38
rs1047308773 Chr.17: 5442973-5442973 on Build GRCh38
rs1011632446 Chr.17: 5443086-5443086 on Build GRCh38
rs559321609 Chr.17: 5442958-5442958 on Build GRCh38
rs1031655729 Chr.17: 5442795-5442795 on Build GRCh38
rs1056823589 Chr.17: 5442913-5442914 on Build GRCh38
rs180850458 Chr.17: 5443124-5443124 on Build GRCh38
rs372857509 Chr.17: 5443034-5443037 on Build GRCh38
rs947078989 Chr.17: 5442863-5442863 on Build GRCh38
rs976819484 Chr.17: 5442780-5442780 on Build GRCh38
rs967912331 Chr.17: 5442867-5442867 on Build GRCh38
rs929922297 Chr.17: 5442940-5442940 on Build GRCh38
rs942443413 Chr.17: 5442910-5442910 on Build GRCh38
rs955472183 Chr.17: 5443174-5443174 on Build GRCh38
rs201214796 Chr.17: 5443031-5443034 on Build GRCh38
rs978023998 Chr.17: 5442869-5442869 on Build GRCh38
rs1031016537 Chr.17: 5443122-5443122 on Build GRCh38
rs940392466 Chr.17: 5443133-5443133 on Build GRCh38
rs1018197127 Chr.17: 5442744-5442744 on Build GRCh38
rs979854764 Chr.17: 5442878-5442878 on Build GRCh38
rs138715319 Chr.17: 5443156-5443156 on Build GRCh38
rs998582855 Chr.17: 5443068-5443069 on Build GRCh38
rs879781810 Chr.17: 5443128-5443129 on Build GRCh38
rs955225655 Chr.17: 5442876-5442876 on Build GRCh38

View Full Product Details