Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6120914-6121413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WSCD1
Entrez Gene ID
23302
Gene Name
WSC domain containing 1
Gene Aliases
-
UniGene
Hs.370166
Gene Chromosome Location
Chr.17: 6069106-6124427 on Build GRCh38
Transcripts
NM_015253.1 XM_005256572.1 XM_005256573.1

SNP Detail

SNP ID Chromosome Location
rs543657075 Chr.17: 6121103-6121103 on Build GRCh38
rs1035719290 Chr.17: 6121128-6121128 on Build GRCh38
rs893399398 Chr.17: 6121075-6121075 on Build GRCh38
rs1002473069 Chr.17: 6121222-6121222 on Build GRCh38
rs932579203 Chr.17: 6121143-6121143 on Build GRCh38
rs536561875 Chr.17: 6121013-6121013 on Build GRCh38
rs913630339 Chr.17: 6121083-6121083 on Build GRCh38
rs141980371 Chr.17: 6121221-6121221 on Build GRCh38
rs1038109224 Chr.17: 6121351-6121351 on Build GRCh38
rs540825861 Chr.17: 6121009-6121009 on Build GRCh38
rs929387730 Chr.17: 6121364-6121364 on Build GRCh38
rs145817538 Chr.17: 6121063-6121063 on Build GRCh38
rs949066115 Chr.17: 6120957-6120957 on Build GRCh38
rs926288770 Chr.17: 6121071-6121071 on Build GRCh38
rs1040563699 Chr.17: 6120936-6120936 on Build GRCh38
rs966435284 Chr.17: 6121282-6121282 on Build GRCh38
rs936440288 Chr.17: 6120982-6120982 on Build GRCh38
rs766449717 Chr.17: 6121298-6121298 on Build GRCh38
rs774159980 Chr.17: 6120992-6120992 on Build GRCh38
rs976437997 Chr.17: 6121291-6121291 on Build GRCh38
rs909780932 Chr.17: 6121306-6121306 on Build GRCh38
rs901981275 Chr.17: 6120948-6120948 on Build GRCh38
rs1040971522 Chr.17: 6120995-6120995 on Build GRCh38
rs571139365 Chr.17: 6121344-6121344 on Build GRCh38
rs1025153614 Chr.17: 6121198-6121198 on Build GRCh38
rs1053399483 Chr.17: 6121069-6121069 on Build GRCh38
rs941786098 Chr.17: 6121368-6121368 on Build GRCh38
rs957917944 Chr.17: 6121077-6121077 on Build GRCh38
rs943143979 Chr.17: 6121040-6121040 on Build GRCh38
rs989544927 Chr.17: 6121247-6121247 on Build GRCh38
rs1049660520 Chr.17: 6121303-6121303 on Build GRCh38
rs765326607 Chr.17: 6121188-6121188 on Build GRCh38
rs576357183 Chr.17: 6120944-6120944 on Build GRCh38
rs572874929 Chr.17: 6121275-6121275 on Build GRCh38
rs748967101 Chr.17: 6120964-6120964 on Build GRCh38
rs943008469 Chr.17: 6121168-6121168 on Build GRCh38
rs1046491644 Chr.17: 6121367-6121367 on Build GRCh38
rs897789105 Chr.17: 6121362-6121362 on Build GRCh38
rs3087809 Chr.17: 6120968-6120968 on Build GRCh38
rs190477276 Chr.17: 6121299-6121299 on Build GRCh38
rs942081979 Chr.17: 6121317-6121317 on Build GRCh38
rs369338447 Chr.17: 6121242-6121242 on Build GRCh38
rs777190990 Chr.17: 6120954-6120954 on Build GRCh38
rs539667364 Chr.17: 6121328-6121328 on Build GRCh38
rs1016742365 Chr.17: 6121035-6121035 on Build GRCh38
rs113346968 Chr.17: 6121372-6121372 on Build GRCh38
rs777224556 Chr.17: 6121113-6121113 on Build GRCh38
rs913693071 Chr.17: 6121258-6121258 on Build GRCh38
rs375038087 Chr.17: 6120983-6120983 on Build GRCh38
rs144975226 Chr.17: 6121034-6121034 on Build GRCh38
rs1045030551 Chr.17: 6121244-6121244 on Build GRCh38
rs1037823435 Chr.17: 6121163-6121163 on Build GRCh38
rs1012201581 Chr.17: 6121076-6121076 on Build GRCh38
rs541872977 Chr.17: 6120961-6120961 on Build GRCh38
rs563460122 Chr.17: 6121123-6121123 on Build GRCh38
rs892319386 Chr.17: 6120988-6120988 on Build GRCh38
rs889142982 Chr.17: 6121153-6121153 on Build GRCh38
rs981910425 Chr.17: 6120994-6120994 on Build GRCh38
rs3826522 Chr.17: 6121373-6121373 on Build GRCh38
rs8066328 Chr.17: 6121232-6121232 on Build GRCh38
rs905831224 Chr.17: 6121118-6121118 on Build GRCh38
rs185803018 Chr.17: 6121283-6121283 on Build GRCh38
rs960515921 Chr.17: 6121001-6121001 on Build GRCh38
rs999135378 Chr.17: 6120958-6120958 on Build GRCh38
rs989269775 Chr.17: 6121079-6121079 on Build GRCh38
rs572236678 Chr.17: 6120979-6120979 on Build GRCh38
rs549035703 Chr.17: 6121154-6121154 on Build GRCh38
rs1025269436 Chr.17: 6121059-6121059 on Build GRCh38
rs962264590 Chr.17: 6121039-6121039 on Build GRCh38
rs889081528 Chr.17: 6121016-6121016 on Build GRCh38
rs1006319702 Chr.17: 6121026-6121026 on Build GRCh38
rs868162180 Chr.17: 6121062-6121062 on Build GRCh38
rs1034330410 Chr.17: 6121226-6121226 on Build GRCh38
rs893286315 Chr.17: 6121231-6121231 on Build GRCh38
rs1049604476 Chr.17: 6121150-6121150 on Build GRCh38
rs980984381 Chr.17: 6121067-6121067 on Build GRCh38
rs78004699 Chr.17: 6121134-6121134 on Build GRCh38
rs748972191 Chr.17: 6120997-6120997 on Build GRCh38
rs930347162 Chr.17: 6121017-6121017 on Build GRCh38
rs993541935 Chr.17: 6121192-6121192 on Build GRCh38
rs532991517 Chr.17: 6121078-6121078 on Build GRCh38
rs1011568958 Chr.17: 6120971-6120971 on Build GRCh38
rs747310735 Chr.17: 6121050-6121050 on Build GRCh38

View Full Product Details