Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 11558431-11558921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHISA6
Entrez Gene ID
388336
Gene Name
shisa family member 6
Gene Aliases
-
UniGene
Hs.592124
Gene Chromosome Location
Chr.17: 11241423-11564063 on Build GRCh38
Transcripts
NM_001173461.1 NM_001173462.1 NM_207386.3 XM_017024618.1

SNP Detail

SNP ID Chromosome Location
rs958674023 Chr.17: 11558481-11558481 on Build GRCh38
rs540997320 Chr.17: 11558459-11558459 on Build GRCh38
rs907585135 Chr.17: 11558881-11558881 on Build GRCh38
rs188446410 Chr.17: 11558612-11558612 on Build GRCh38
rs537307690 Chr.17: 11558814-11558814 on Build GRCh38
rs977045144 Chr.17: 11558643-11558643 on Build GRCh38
rs933050921 Chr.17: 11558662-11558662 on Build GRCh38
rs777771377 Chr.17: 11558493-11558493 on Build GRCh38
rs968381021 Chr.17: 11558633-11558633 on Build GRCh38
rs964634370 Chr.17: 11558765-11558765 on Build GRCh38
rs562226417 Chr.17: 11558656-11558656 on Build GRCh38
rs970780308 Chr.17: 11558570-11558570 on Build GRCh38
rs1025161977 Chr.17: 11558565-11558565 on Build GRCh38
rs977349414 Chr.17: 11558499-11558499 on Build GRCh38
rs989461415 Chr.17: 11558751-11558751 on Build GRCh38
rs867410165 Chr.17: 11558754-11558754 on Build GRCh38
rs923926081 Chr.17: 11558599-11558599 on Build GRCh38
rs1021823287 Chr.17: 11558787-11558789 on Build GRCh38
rs116990566 Chr.17: 11558774-11558774 on Build GRCh38
rs571579529 Chr.17: 11558631-11558631 on Build GRCh38
rs767045230 Chr.17: 11558581-11558581 on Build GRCh38
rs574788646 Chr.17: 11558895-11558895 on Build GRCh38
rs750886125 Chr.17: 11558483-11558483 on Build GRCh38
rs933185254 Chr.17: 11558524-11558524 on Build GRCh38
rs188955816 Chr.17: 11558717-11558717 on Build GRCh38
rs1031131737 Chr.17: 11558743-11558743 on Build GRCh38
rs3095679 Chr.17: 11558661-11558661 on Build GRCh38
rs947044829 Chr.17: 11558490-11558490 on Build GRCh38
rs964930700 Chr.17: 11558847-11558847 on Build GRCh38
rs200653411 Chr.17: 11558813-11558813 on Build GRCh38
rs958487583 Chr.17: 11558614-11558614 on Build GRCh38
rs745701636 Chr.17: 11558817-11558817 on Build GRCh38
rs550379318 Chr.17: 11558630-11558630 on Build GRCh38
rs927984254 Chr.17: 11558897-11558898 on Build GRCh38
rs758702499 Chr.17: 11558793-11558793 on Build GRCh38
rs987829159 Chr.17: 11558839-11558839 on Build GRCh38
rs1005923648 Chr.17: 11558675-11558675 on Build GRCh38
rs143397292 Chr.17: 11558473-11558473 on Build GRCh38
rs906729141 Chr.17: 11558736-11558736 on Build GRCh38
rs1006868918 Chr.17: 11558485-11558485 on Build GRCh38
rs973232178 Chr.17: 11558852-11558852 on Build GRCh38
rs184203270 Chr.17: 11558714-11558714 on Build GRCh38
rs1047360027 Chr.17: 11558460-11558460 on Build GRCh38
rs781046223 Chr.17: 11558894-11558894 on Build GRCh38
rs999603026 Chr.17: 11558737-11558737 on Build GRCh38
rs553039943 Chr.17: 11558835-11558835 on Build GRCh38
rs1012385305 Chr.17: 11558562-11558562 on Build GRCh38
rs755048537 Chr.17: 11558722-11558722 on Build GRCh38
rs958367119 Chr.17: 11558750-11558750 on Build GRCh38
rs79470314 Chr.17: 11558789-11558789 on Build GRCh38
rs569380455 Chr.17: 11558773-11558773 on Build GRCh38
rs929336654 Chr.17: 11558884-11558884 on Build GRCh38
rs933188224 Chr.17: 11558808-11558808 on Build GRCh38
rs151205142 Chr.17: 11558613-11558613 on Build GRCh38
rs868142334 Chr.17: 11558452-11558452 on Build GRCh38
rs1016548497 Chr.17: 11558530-11558530 on Build GRCh38
rs943409375 Chr.17: 11558559-11558559 on Build GRCh38
rs140443293 Chr.17: 11558577-11558577 on Build GRCh38
rs1037782775 Chr.17: 11558682-11558682 on Build GRCh38
rs941789345 Chr.17: 11558890-11558890 on Build GRCh38
rs1021370727 Chr.17: 11558621-11558621 on Build GRCh38
rs566257534 Chr.17: 11558698-11558698 on Build GRCh38
rs886095024 Chr.17: 11558673-11558673 on Build GRCh38
rs34294075 Chr.17: 11558765-11558766 on Build GRCh38
rs529956466 Chr.17: 11558474-11558474 on Build GRCh38
rs561319727 Chr.17: 11558606-11558606 on Build GRCh38
rs543141288 Chr.17: 11558537-11558537 on Build GRCh38
rs1047736662 Chr.17: 11558729-11558729 on Build GRCh38
rs1038814224 Chr.17: 11558632-11558632 on Build GRCh38
rs906783836 Chr.17: 11558735-11558735 on Build GRCh38
rs991679806 Chr.17: 11558617-11558617 on Build GRCh38
rs910468514 Chr.17: 11558845-11558845 on Build GRCh38
rs990471182 Chr.17: 11558607-11558607 on Build GRCh38
rs191754634 Chr.17: 11558667-11558667 on Build GRCh38
rs1052006966 Chr.17: 11558525-11558525 on Build GRCh38
rs895341292 Chr.17: 11558760-11558760 on Build GRCh38
rs140291120 Chr.17: 11558831-11558831 on Build GRCh38
rs924514355 Chr.17: 11558518-11558518 on Build GRCh38

View Full Product Details