Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15651000-15651497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM16
Entrez Gene ID
10626
Gene Name
tripartite motif containing 16
Gene Aliases
EBBP
UniGene
Hs.123534
Gene Chromosome Location
Chr.17: 15627966-15682879 on Build GRCh38
Transcripts
NM_006470.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5440041 c.338A>G p.K113R Chr.17: 15651272-15651272 on Build GRCh38
401683 c.320C>T p.P107L Chr.17: 15651290-15651290 on Build GRCh38
148197 c.363G>T p.E121D Chr.17: 15651247-15651247 on Build GRCh38
5499590 c.340C>G p.L114V Chr.17: 15651270-15651270 on Build GRCh38
7196695 c.307G>A p.E103K Chr.17: 15651303-15651303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937381211 Chr.17: 15651249-15651249 on Build GRCh38
rs770668690 Chr.17: 15651372-15651372 on Build GRCh38
rs749500109 Chr.17: 15651366-15651366 on Build GRCh38
rs766231260 Chr.17: 15651167-15651167 on Build GRCh38
rs776009860 Chr.17: 15651108-15651108 on Build GRCh38
rs377452311 Chr.17: 15651429-15651429 on Build GRCh38
rs767578705 Chr.17: 15651387-15651387 on Build GRCh38
rs1042011844 Chr.17: 15651069-15651069 on Build GRCh38
rs768380215 Chr.17: 15651376-15651376 on Build GRCh38
rs751497068 Chr.17: 15651275-15651275 on Build GRCh38
rs772418858 Chr.17: 15651206-15651206 on Build GRCh38
rs657380 Chr.17: 15651217-15651217 on Build GRCh38
rs138524166 Chr.17: 15651258-15651258 on Build GRCh38
rs749283138 Chr.17: 15651473-15651473 on Build GRCh38
rs772377200 Chr.17: 15651043-15651043 on Build GRCh38
rs746827352 Chr.17: 15651362-15651364 on Build GRCh38
rs767519631 Chr.17: 15651278-15651278 on Build GRCh38
rs781057869 Chr.17: 15651074-15651074 on Build GRCh38
rs780443884 Chr.17: 15651301-15651301 on Build GRCh38
rs149620739 Chr.17: 15651446-15651446 on Build GRCh38
rs373394688 Chr.17: 15651078-15651078 on Build GRCh38
rs140413277 Chr.17: 15651467-15651467 on Build GRCh38
rs113007976 Chr.17: 15651051-15651051 on Build GRCh38
rs776407777 Chr.17: 15651417-15651418 on Build GRCh38
rs1037290040 Chr.17: 15651363-15651363 on Build GRCh38
rs773415380 Chr.17: 15651250-15651250 on Build GRCh38
rs754444064 Chr.17: 15651066-15651066 on Build GRCh38
rs1054492375 Chr.17: 15651319-15651319 on Build GRCh38
rs4792642 Chr.17: 15651190-15651190 on Build GRCh38
rs759354072 Chr.17: 15651386-15651386 on Build GRCh38
rs148027903 Chr.17: 15651290-15651290 on Build GRCh38
rs765064432 Chr.17: 15651447-15651461 on Build GRCh38
rs1000248219 Chr.17: 15651079-15651079 on Build GRCh38
rs1026773661 Chr.17: 15651140-15651140 on Build GRCh38
rs534484297 Chr.17: 15651309-15651309 on Build GRCh38
rs779991124 Chr.17: 15651054-15651054 on Build GRCh38
rs780822886 Chr.17: 15651180-15651180 on Build GRCh38
rs751508968 Chr.17: 15651173-15651173 on Build GRCh38
rs748640378 Chr.17: 15651466-15651466 on Build GRCh38
rs1043794122 Chr.17: 15651024-15651024 on Build GRCh38
rs779387833 Chr.17: 15651091-15651091 on Build GRCh38
rs879000045 Chr.17: 15651154-15651155 on Build GRCh38
rs945420249 Chr.17: 15651245-15651245 on Build GRCh38
rs657363 Chr.17: 15651202-15651202 on Build GRCh38
rs763651571 Chr.17: 15651420-15651420 on Build GRCh38
rs752274548 Chr.17: 15651288-15651288 on Build GRCh38
rs755513301 Chr.17: 15651310-15651310 on Build GRCh38
rs140938167 Chr.17: 15651291-15651291 on Build GRCh38
rs201801259 Chr.17: 15651146-15651146 on Build GRCh38
rs755629686 Chr.17: 15651289-15651289 on Build GRCh38
rs562799935 Chr.17: 15651153-15651153 on Build GRCh38
rs762748965 Chr.17: 15651148-15651148 on Build GRCh38
rs751043235 Chr.17: 15651064-15651064 on Build GRCh38
rs560028798 Chr.17: 15651200-15651200 on Build GRCh38
rs141481228 Chr.17: 15651413-15651413 on Build GRCh38
rs145594570 Chr.17: 15651223-15651223 on Build GRCh38
rs757111961 Chr.17: 15651040-15651040 on Build GRCh38
rs754499963 Chr.17: 15651178-15651178 on Build GRCh38
rs528978044 Chr.17: 15651225-15651225 on Build GRCh38
rs772482617 Chr.17: 15651105-15651105 on Build GRCh38
rs753118418 Chr.17: 15651337-15651337 on Build GRCh38
rs200262081 Chr.17: 15651134-15651134 on Build GRCh38
rs116554938 Chr.17: 15651384-15651384 on Build GRCh38
rs759511584 Chr.17: 15651276-15651276 on Build GRCh38
rs150907471 Chr.17: 15651421-15651421 on Build GRCh38
rs1007320809 Chr.17: 15651119-15651119 on Build GRCh38
rs771467114 Chr.17: 15651088-15651088 on Build GRCh38
rs144630893 Chr.17: 15651224-15651224 on Build GRCh38
rs762722832 Chr.17: 15651252-15651252 on Build GRCh38
rs760331850 Chr.17: 15651416-15651416 on Build GRCh38
rs775534126 Chr.17: 15651077-15651077 on Build GRCh38
rs182850025 Chr.17: 15651052-15651052 on Build GRCh38
rs769858985 Chr.17: 15651353-15651353 on Build GRCh38
rs376571595 Chr.17: 15651116-15651116 on Build GRCh38
rs765739065 Chr.17: 15651156-15651156 on Build GRCh38
rs762002659 Chr.17: 15651128-15651128 on Build GRCh38
rs2074890 Chr.17: 15651247-15651247 on Build GRCh38
rs776662920 Chr.17: 15651243-15651243 on Build GRCh38
rs200484084 Chr.17: 15651459-15651459 on Build GRCh38
rs755266781 Chr.17: 15651454-15651454 on Build GRCh38
rs776896442 Chr.17: 15651125-15651125 on Build GRCh38
rs371753308 Chr.17: 15651299-15651299 on Build GRCh38
rs746201378 Chr.17: 15651104-15651104 on Build GRCh38
rs144568697 Chr.17: 15651191-15651191 on Build GRCh38
rs144663766 Chr.17: 15651294-15651294 on Build GRCh38
rs912745765 Chr.17: 15651317-15651317 on Build GRCh38
rs773363981 Chr.17: 15651355-15651355 on Build GRCh38
rs756787854 Chr.17: 15651328-15651329 on Build GRCh38
rs761798463 Chr.17: 15651429-15651430 on Build GRCh38
rs188307419 Chr.17: 15651430-15651430 on Build GRCh38
rs748229826 Chr.17: 15651352-15651352 on Build GRCh38
rs965801663 Chr.17: 15651436-15651436 on Build GRCh38
rs781620349 Chr.17: 15651321-15651321 on Build GRCh38
rs61741220 Chr.17: 15651142-15651142 on Build GRCh38
rs138309375 Chr.17: 15651445-15651445 on Build GRCh38
rs751946025 Chr.17: 15651453-15651453 on Build GRCh38
rs895650036 Chr.17: 15651021-15651021 on Build GRCh38
rs888783982 Chr.17: 15651340-15651340 on Build GRCh38
rs560522804 Chr.17: 15651082-15651082 on Build GRCh38

View Full Product Details