Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15741704-15742199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D26
Entrez Gene ID
353149
Gene Name
TBC1 domain family member 26
Gene Aliases
-
UniGene
Hs.434123
Gene Chromosome Location
Chr.17: 15732277-15744784 on Build GRCh38
Transcripts
NM_178571.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6797382 c.707_708insTC p.R237fs*>15 Chr.17: 15741982-15741983 on Build GRCh38
5820680 c.714C>A p.T238T Chr.17: 15741989-15741989 on Build GRCh38
975829 c.695C>T p.S232L Chr.17: 15741970-15741970 on Build GRCh38
3755281 c.700G>A p.G234S Chr.17: 15741975-15741975 on Build GRCh38
5572466 c.706T>A p.Y236N Chr.17: 15741981-15741981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758103295 Chr.17: 15742075-15742075 on Build GRCh38
rs543843846 Chr.17: 15741827-15741827 on Build GRCh38
rs775133285 Chr.17: 15741930-15741930 on Build GRCh38
rs763084367 Chr.17: 15741918-15741918 on Build GRCh38
rs371552821 Chr.17: 15741996-15741996 on Build GRCh38
rs192774848 Chr.17: 15741967-15741967 on Build GRCh38
rs375958701 Chr.17: 15742029-15742029 on Build GRCh38
rs761078651 Chr.17: 15741897-15741897 on Build GRCh38
rs759839354 Chr.17: 15742008-15742008 on Build GRCh38
rs748552512 Chr.17: 15742078-15742078 on Build GRCh38
rs745552552 Chr.17: 15741960-15741960 on Build GRCh38
rs375067059 Chr.17: 15741944-15741944 on Build GRCh38
rs768333815 Chr.17: 15742016-15742023 on Build GRCh38
rs777237238 Chr.17: 15741760-15741760 on Build GRCh38
rs752484963 Chr.17: 15741931-15741931 on Build GRCh38
rs916998718 Chr.17: 15741920-15741920 on Build GRCh38
rs953088133 Chr.17: 15741845-15741845 on Build GRCh38
rs765715917 Chr.17: 15741910-15741910 on Build GRCh38
rs753836103 Chr.17: 15742058-15742058 on Build GRCh38
rs746705470 Chr.17: 15741928-15741930 on Build GRCh38
rs571602964 Chr.17: 15741788-15741788 on Build GRCh38
rs533052229 Chr.17: 15741917-15741917 on Build GRCh38
rs757103448 Chr.17: 15742064-15742064 on Build GRCh38
rs575218158 Chr.17: 15741819-15741819 on Build GRCh38
rs374054402 Chr.17: 15742140-15742140 on Build GRCh38
rs773490341 Chr.17: 15742041-15742042 on Build GRCh38
rs926967789 Chr.17: 15741875-15741875 on Build GRCh38
rs372066766 Chr.17: 15742070-15742070 on Build GRCh38
rs889761126 Chr.17: 15741774-15741774 on Build GRCh38
rs1019645554 Chr.17: 15741791-15741791 on Build GRCh38
rs566345514 Chr.17: 15742147-15742147 on Build GRCh38
rs769032592 Chr.17: 15741900-15741900 on Build GRCh38
rs370843078 Chr.17: 15741904-15741904 on Build GRCh38
rs1025773051 Chr.17: 15741839-15741839 on Build GRCh38
rs551474325 Chr.17: 15742090-15742090 on Build GRCh38
rs367840777 Chr.17: 15741985-15741985 on Build GRCh38
rs529472345 Chr.17: 15741952-15741952 on Build GRCh38
rs200550508 Chr.17: 15741970-15741970 on Build GRCh38
rs535267267 Chr.17: 15742164-15742164 on Build GRCh38
rs1000084231 Chr.17: 15741731-15741731 on Build GRCh38
rs774027640 Chr.17: 15741891-15741891 on Build GRCh38
rs965552591 Chr.17: 15741812-15741812 on Build GRCh38
rs774096737 Chr.17: 15741984-15741984 on Build GRCh38
rs767841021 Chr.17: 15742026-15742026 on Build GRCh38
rs771647420 Chr.17: 15741965-15741965 on Build GRCh38
rs973311129 Chr.17: 15741824-15741824 on Build GRCh38
rs189760905 Chr.17: 15741878-15741878 on Build GRCh38
rs984362817 Chr.17: 15741867-15741867 on Build GRCh38
rs1034591177 Chr.17: 15742096-15742096 on Build GRCh38
rs756067120 Chr.17: 15741942-15741942 on Build GRCh38
rs766546411 Chr.17: 15741919-15741919 on Build GRCh38
rs369993816 Chr.17: 15742051-15742051 on Build GRCh38
rs777642006 Chr.17: 15742033-15742033 on Build GRCh38
rs762310026 Chr.17: 15741909-15741909 on Build GRCh38
rs534032712 Chr.17: 15742118-15742118 on Build GRCh38
rs763991069 Chr.17: 15742055-15742055 on Build GRCh38
rs865991437 Chr.17: 15741932-15741932 on Build GRCh38
rs201575162 Chr.17: 15741929-15741929 on Build GRCh38
rs553810440 Chr.17: 15742141-15742141 on Build GRCh38
rs17855672 Chr.17: 15741975-15741975 on Build GRCh38
rs945776390 Chr.17: 15741927-15741928 on Build GRCh38
rs773738177 Chr.17: 15741979-15741979 on Build GRCh38
rs778238190 Chr.17: 15741956-15741956 on Build GRCh38
rs571452794 Chr.17: 15742094-15742094 on Build GRCh38
rs1006243234 Chr.17: 15742089-15742089 on Build GRCh38
rs75518324 Chr.17: 15741947-15741947 on Build GRCh38
rs745322687 Chr.17: 15742071-15742071 on Build GRCh38

View Full Product Details