Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27907068-27907613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPA2
Entrez Gene ID
6118
Gene Name
replication protein A2
Gene Aliases
REPA2, RP-A p32, RP-A p34, RPA32
UniGene
Hs.79411
Gene Chromosome Location
Chr.1: 27891524-27914797 on Build GRCh38
Transcripts
NM_001286076.1 NM_001297558.1 NM_002946.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
534753 c.441G>T p.L147F Chr.1: 27907223-27907223 on Build GRCh38
6769002 c.155C>T p.S52F Chr.1: 27907245-27907245 on Build GRCh38
680132 c.199G>T p.G67W Chr.1: 27907201-27907201 on Build GRCh38
4860528 c.223G>T p.G75W Chr.1: 27907201-27907201 on Build GRCh38
5735346 c.219G>T p.Q73H Chr.1: 27907181-27907181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041712257 Chr.1: 27907172-27907172 on Build GRCh38
rs7535816 Chr.1: 27907543-27907543 on Build GRCh38
rs753513007 Chr.1: 27907296-27907296 on Build GRCh38
rs776720485 Chr.1: 27907183-27907183 on Build GRCh38
rs181135231 Chr.1: 27907489-27907489 on Build GRCh38
rs963271686 Chr.1: 27907514-27907514 on Build GRCh38
rs372538532 Chr.1: 27907297-27907297 on Build GRCh38
rs779346244 Chr.1: 27907135-27907135 on Build GRCh38
rs748920260 Chr.1: 27907440-27907440 on Build GRCh38
rs755866319 Chr.1: 27907276-27907276 on Build GRCh38
rs540849246 Chr.1: 27907249-27907249 on Build GRCh38
rs770432057 Chr.1: 27907555-27907555 on Build GRCh38
rs904825024 Chr.1: 27907579-27907579 on Build GRCh38
rs371007532 Chr.1: 27907515-27907515 on Build GRCh38
rs889276630 Chr.1: 27907256-27907256 on Build GRCh38
rs775993410 Chr.1: 27907138-27907138 on Build GRCh38
rs1010506596 Chr.1: 27907271-27907271 on Build GRCh38
rs748208522 Chr.1: 27907165-27907166 on Build GRCh38
rs367965034 Chr.1: 27907224-27907224 on Build GRCh38
rs765428859 Chr.1: 27907223-27907223 on Build GRCh38
rs5030742 Chr.1: 27907225-27907225 on Build GRCh38
rs1050563630 Chr.1: 27907227-27907227 on Build GRCh38
rs767211752 Chr.1: 27907255-27907255 on Build GRCh38
rs761941067 Chr.1: 27907211-27907211 on Build GRCh38
rs773557487 Chr.1: 27907317-27907320 on Build GRCh38
rs546369892 Chr.1: 27907552-27907552 on Build GRCh38
rs747328560 Chr.1: 27907332-27907332 on Build GRCh38
rs556399818 Chr.1: 27907457-27907457 on Build GRCh38
rs1025643736 Chr.1: 27907420-27907420 on Build GRCh38
rs951432403 Chr.1: 27907463-27907463 on Build GRCh38
rs746373645 Chr.1: 27907312-27907312 on Build GRCh38
rs768861340 Chr.1: 27907169-27907169 on Build GRCh38
rs376928504 Chr.1: 27907217-27907217 on Build GRCh38
rs28988921 Chr.1: 27907474-27907474 on Build GRCh38
rs562736368 Chr.1: 27907439-27907439 on Build GRCh38
rs921754186 Chr.1: 27907516-27907516 on Build GRCh38
rs746425118 Chr.1: 27907136-27907136 on Build GRCh38
rs546320364 Chr.1: 27907434-27907434 on Build GRCh38
rs768500130 Chr.1: 27907145-27907148 on Build GRCh38
rs960187213 Chr.1: 27907335-27907335 on Build GRCh38
rs760791137 Chr.1: 27907166-27907166 on Build GRCh38
rs866101587 Chr.1: 27907363-27907363 on Build GRCh38
rs962234142 Chr.1: 27907282-27907282 on Build GRCh38
rs1051474282 Chr.1: 27907226-27907226 on Build GRCh38
rs189697485 Chr.1: 27907375-27907375 on Build GRCh38
rs779620779 Chr.1: 27907380-27907380 on Build GRCh38
rs772399026 Chr.1: 27907137-27907137 on Build GRCh38
rs1025238217 Chr.1: 27907319-27907319 on Build GRCh38
rs1006268188 Chr.1: 27907584-27907584 on Build GRCh38
rs373150909 Chr.1: 27907506-27907506 on Build GRCh38
rs941812612 Chr.1: 27907125-27907125 on Build GRCh38
rs1050940524 Chr.1: 27907577-27907577 on Build GRCh38
rs551895891 Chr.1: 27907193-27907193 on Build GRCh38
rs752479657 Chr.1: 27907272-27907272 on Build GRCh38
rs971110715 Chr.1: 27907415-27907416 on Build GRCh38
rs770067079 Chr.1: 27907169-27907170 on Build GRCh38
rs781090246 Chr.1: 27907270-27907270 on Build GRCh38
rs532485060 Chr.1: 27907417-27907417 on Build GRCh38
rs777442773 Chr.1: 27907293-27907293 on Build GRCh38
rs781327933 Chr.1: 27907147-27907150 on Build GRCh38
rs376585215 Chr.1: 27907299-27907299 on Build GRCh38
rs578019301 Chr.1: 27907109-27907109 on Build GRCh38
rs904772611 Chr.1: 27907288-27907288 on Build GRCh38
rs780434874 Chr.1: 27907322-27907322 on Build GRCh38
rs912910638 Chr.1: 27907547-27907547 on Build GRCh38
rs933087990 Chr.1: 27907540-27907540 on Build GRCh38
rs28988920 Chr.1: 27907571-27907571 on Build GRCh38
rs755871861 Chr.1: 27907314-27907314 on Build GRCh38

View Full Product Details