Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27945125-27945631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMPDL3B
Entrez Gene ID
27293
Gene Name
sphingomyelin phosphodiesterase acid like 3B
Gene Aliases
ASML3B
UniGene
Hs.123659
Gene Chromosome Location
Chr.1: 27934955-27959157 on Build GRCh38
Transcripts
NM_001009568.2 NM_001304579.1 NM_014474.3 XM_011541259.2 XM_011541260.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6785691 c.163G>A p.D55N Chr.1: 27945333-27945333 on Build GRCh38
3487733 c.149C>T p.S50F Chr.1: 27945319-27945319 on Build GRCh38
3487732 c.149C>T p.S50F Chr.1: 27945319-27945319 on Build GRCh38
3487735 c.172C>T p.P58S Chr.1: 27945342-27945342 on Build GRCh38
6785694 c.162C>T p.P54P Chr.1: 27945332-27945332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377076689 Chr.1: 27945221-27945221 on Build GRCh38
rs143396392 Chr.1: 27945407-27945407 on Build GRCh38
rs897816429 Chr.1: 27945241-27945241 on Build GRCh38
rs76374093 Chr.1: 27945199-27945199 on Build GRCh38
rs776230924 Chr.1: 27945418-27945418 on Build GRCh38
rs1021301269 Chr.1: 27945272-27945272 on Build GRCh38
rs951535753 Chr.1: 27945369-27945369 on Build GRCh38
rs1034417100 Chr.1: 27945603-27945603 on Build GRCh38
rs750939971 Chr.1: 27945333-27945333 on Build GRCh38
rs77491746 Chr.1: 27945198-27945198 on Build GRCh38
rs547045855 Chr.1: 27945153-27945153 on Build GRCh38
rs377660078 Chr.1: 27945381-27945381 on Build GRCh38
rs1021266705 Chr.1: 27945385-27945385 on Build GRCh38
rs758755619 Chr.1: 27945222-27945222 on Build GRCh38
rs751611334 Chr.1: 27945602-27945602 on Build GRCh38
rs779424305 Chr.1: 27945194-27945194 on Build GRCh38
rs529895475 Chr.1: 27945178-27945178 on Build GRCh38
rs1009996050 Chr.1: 27945555-27945555 on Build GRCh38
rs113119629 Chr.1: 27945607-27945607 on Build GRCh38
rs538609600 Chr.1: 27945183-27945183 on Build GRCh38
rs761516460 Chr.1: 27945427-27945427 on Build GRCh38
rs145731626 Chr.1: 27945248-27945248 on Build GRCh38
rs748200088 Chr.1: 27945265-27945265 on Build GRCh38
rs763693269 Chr.1: 27945313-27945313 on Build GRCh38
rs141404787 Chr.1: 27945293-27945293 on Build GRCh38
rs767330057 Chr.1: 27945290-27945290 on Build GRCh38
rs780627580 Chr.1: 27945335-27945335 on Build GRCh38
rs775527405 Chr.1: 27945401-27945401 on Build GRCh38
rs1052018019 Chr.1: 27945246-27945246 on Build GRCh38
rs772619758 Chr.1: 27945251-27945252 on Build GRCh38
rs79111444 Chr.1: 27945196-27945196 on Build GRCh38
rs141035438 Chr.1: 27945270-27945270 on Build GRCh38
rs762573078 Chr.1: 27945252-27945253 on Build GRCh38
rs544282310 Chr.1: 27945249-27945249 on Build GRCh38
rs756232960 Chr.1: 27945484-27945484 on Build GRCh38
rs200259391 Chr.1: 27945233-27945233 on Build GRCh38
rs371271480 Chr.1: 27945334-27945334 on Build GRCh38
rs984131434 Chr.1: 27945379-27945379 on Build GRCh38
rs781719214 Chr.1: 27945481-27945481 on Build GRCh38
rs755374098 Chr.1: 27945480-27945480 on Build GRCh38
rs759457560 Chr.1: 27945288-27945288 on Build GRCh38
rs888446491 Chr.1: 27945162-27945162 on Build GRCh38
rs777773009 Chr.1: 27945371-27945371 on Build GRCh38
rs759586530 Chr.1: 27945517-27945517 on Build GRCh38
rs145282396 Chr.1: 27945482-27945482 on Build GRCh38
rs370892810 Chr.1: 27945456-27945456 on Build GRCh38
rs554773189 Chr.1: 27945513-27945513 on Build GRCh38
rs762531990 Chr.1: 27945450-27945450 on Build GRCh38
rs945826067 Chr.1: 27945146-27945146 on Build GRCh38
rs74922246 Chr.1: 27945197-27945197 on Build GRCh38
rs150797389 Chr.1: 27945332-27945332 on Build GRCh38
rs1002336053 Chr.1: 27945295-27945295 on Build GRCh38
rs989676027 Chr.1: 27945373-27945373 on Build GRCh38
rs1038948175 Chr.1: 27945449-27945449 on Build GRCh38
rs764686418 Chr.1: 27945188-27945189 on Build GRCh38
rs749849136 Chr.1: 27945446-27945446 on Build GRCh38
rs769742234 Chr.1: 27945556-27945556 on Build GRCh38
rs923114234 Chr.1: 27945162-27945162 on Build GRCh38
rs771178377 Chr.1: 27945506-27945506 on Build GRCh38
rs781517843 Chr.1: 27945349-27945349 on Build GRCh38
rs751908684 Chr.1: 27945336-27945336 on Build GRCh38
rs1001798862 Chr.1: 27945302-27945302 on Build GRCh38
rs780353245 Chr.1: 27945226-27945226 on Build GRCh38
rs760410391 Chr.1: 27945402-27945402 on Build GRCh38
rs748430948 Chr.1: 27945351-27945351 on Build GRCh38
rs910371252 Chr.1: 27945602-27945602 on Build GRCh38
rs375944325 Chr.1: 27945367-27945367 on Build GRCh38
rs764794646 Chr.1: 27945428-27945428 on Build GRCh38
rs933153354 Chr.1: 27945551-27945551 on Build GRCh38
rs760465883 Chr.1: 27945304-27945304 on Build GRCh38
rs751294134 Chr.1: 27945386-27945388 on Build GRCh38
rs753508870 Chr.1: 27945326-27945326 on Build GRCh38
rs573749895 Chr.1: 27945601-27945601 on Build GRCh38
rs904206654 Chr.1: 27945591-27945591 on Build GRCh38
rs746407046 Chr.1: 27945195-27945195 on Build GRCh38
rs774323423 Chr.1: 27945279-27945279 on Build GRCh38
rs752135504 Chr.1: 27945471-27945471 on Build GRCh38
rs778058897 Chr.1: 27945491-27945491 on Build GRCh38
rs771835908 Chr.1: 27945388-27945388 on Build GRCh38
rs765917801 Chr.1: 27945327-27945328 on Build GRCh38

View Full Product Details