Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28593566-28594071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB42
Entrez Gene ID
115273
Gene Name
RAB42, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.652321
Gene Chromosome Location
Chr.1: 28592200-28595444 on Build GRCh38
Transcripts
NM_001193532.1 NM_152304.1 XM_017000226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7115704 c.153C>T p.C51C Chr.1: 28593952-28593952 on Build GRCh38
907993 c.37C>T p.L13L Chr.1: 28593836-28593836 on Build GRCh38
680113 c.135C>T p.T45T Chr.1: 28593934-28593934 on Build GRCh38
6452301 c.97G>C p.E33Q Chr.1: 28593896-28593896 on Build GRCh38
4966799 c.250C>T p.L84F Chr.1: 28594049-28594049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007370007 Chr.1: 28593603-28593603 on Build GRCh38
rs752734250 Chr.1: 28593876-28593876 on Build GRCh38
rs750123180 Chr.1: 28593998-28593998 on Build GRCh38
rs558733882 Chr.1: 28593892-28593892 on Build GRCh38
rs753505515 Chr.1: 28593885-28593885 on Build GRCh38
rs756486630 Chr.1: 28594032-28594032 on Build GRCh38
rs1023042316 Chr.1: 28593975-28593975 on Build GRCh38
rs756944729 Chr.1: 28593887-28593887 on Build GRCh38
rs375798399 Chr.1: 28593870-28593870 on Build GRCh38
rs951257705 Chr.1: 28593654-28593654 on Build GRCh38
rs748662385 Chr.1: 28594030-28594030 on Build GRCh38
rs766539203 Chr.1: 28593873-28593873 on Build GRCh38
rs1039426552 Chr.1: 28593910-28593910 on Build GRCh38
rs369958463 Chr.1: 28594036-28594036 on Build GRCh38
rs144161680 Chr.1: 28593917-28593917 on Build GRCh38
rs148146932 Chr.1: 28593934-28593934 on Build GRCh38
rs761578781 Chr.1: 28593966-28593966 on Build GRCh38
rs146157625 Chr.1: 28593936-28593936 on Build GRCh38
rs149069590 Chr.1: 28593918-28593918 on Build GRCh38
rs976849278 Chr.1: 28593680-28593680 on Build GRCh38
rs141958895 Chr.1: 28594046-28594046 on Build GRCh38
rs758007316 Chr.1: 28593999-28593999 on Build GRCh38
rs750402725 Chr.1: 28593731-28593731 on Build GRCh38
rs74065028 Chr.1: 28593594-28593594 on Build GRCh38
rs769851968 Chr.1: 28593867-28593867 on Build GRCh38
rs778237147 Chr.1: 28594035-28594035 on Build GRCh38
rs575396798 Chr.1: 28593900-28593900 on Build GRCh38
rs372832023 Chr.1: 28593817-28593817 on Build GRCh38
rs774530195 Chr.1: 28593943-28593943 on Build GRCh38
rs1018809513 Chr.1: 28593629-28593629 on Build GRCh38
rs370099619 Chr.1: 28593893-28593893 on Build GRCh38
rs148442336 Chr.1: 28593762-28593762 on Build GRCh38
rs924746164 Chr.1: 28593824-28593824 on Build GRCh38
rs556985641 Chr.1: 28593587-28593587 on Build GRCh38
rs765392082 Chr.1: 28593710-28593710 on Build GRCh38
rs763719859 Chr.1: 28593958-28593958 on Build GRCh38
rs1035572719 Chr.1: 28593699-28593699 on Build GRCh38
rs753536519 Chr.1: 28594034-28594035 on Build GRCh38
rs771555587 Chr.1: 28593846-28593846 on Build GRCh38
rs776614667 Chr.1: 28593954-28593954 on Build GRCh38
rs924056105 Chr.1: 28593694-28593694 on Build GRCh38
rs147843526 Chr.1: 28593852-28593852 on Build GRCh38
rs765880684 Chr.1: 28594000-28594000 on Build GRCh38
rs776822894 Chr.1: 28593857-28593857 on Build GRCh38
rs745310965 Chr.1: 28593829-28593829 on Build GRCh38
rs760691136 Chr.1: 28593948-28593948 on Build GRCh38
rs551662559 Chr.1: 28593937-28593937 on Build GRCh38
rs867218443 Chr.1: 28593716-28593716 on Build GRCh38
rs370993430 Chr.1: 28593952-28593952 on Build GRCh38
rs751012787 Chr.1: 28594004-28594004 on Build GRCh38
rs111894928 Chr.1: 28593849-28593849 on Build GRCh38
rs553426672 Chr.1: 28593781-28593781 on Build GRCh38
rs767623892 Chr.1: 28593751-28593751 on Build GRCh38
rs56083542 Chr.1: 28593747-28593747 on Build GRCh38
rs781655799 Chr.1: 28594007-28594007 on Build GRCh38
rs756817119 Chr.1: 28593823-28593823 on Build GRCh38
rs762100612 Chr.1: 28593859-28593859 on Build GRCh38
rs530073440 Chr.1: 28593926-28593926 on Build GRCh38
rs771209228 Chr.1: 28594041-28594041 on Build GRCh38
rs778486046 Chr.1: 28593827-28593827 on Build GRCh38
rs760224359 Chr.1: 28593872-28593873 on Build GRCh38
rs754498996 Chr.1: 28594006-28594006 on Build GRCh38
rs777410636 Chr.1: 28593814-28593814 on Build GRCh38
rs965892823 Chr.1: 28593643-28593643 on Build GRCh38
rs770193273 Chr.1: 28593935-28593935 on Build GRCh38
rs761461909 Chr.1: 28594035-28594036 on Build GRCh38
rs763197223 Chr.1: 28593872-28593872 on Build GRCh38
rs757922679 Chr.1: 28593881-28593881 on Build GRCh38
rs765050696 Chr.1: 28593980-28593980 on Build GRCh38
rs745999616 Chr.1: 28594049-28594049 on Build GRCh38
rs759560670 Chr.1: 28593732-28593732 on Build GRCh38

View Full Product Details