Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 30936551-30937048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PUM1
Entrez Gene ID
9698
Gene Name
pumilio RNA binding family member 1
Gene Aliases
HSPUM, PUMH, PUMH1, PUML1
UniGene
Hs.281707
Gene Chromosome Location
Chr.1: 30931506-31065717 on Build GRCh38
Transcripts
NM_001020658.1 NM_014676.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3977565 c.3397G>T p.E1133* Chr.1: 30936675-30936675 on Build GRCh38
4767889 c.3390C>A p.D1130E Chr.1: 30936682-30936682 on Build GRCh38
7402362 c.3242T>C p.V1081A Chr.1: 30936830-30936830 on Build GRCh38
6040323 c.3425A>C p.H1142P Chr.1: 30936647-30936647 on Build GRCh38
2076986 c.3319G>A p.G1107S Chr.1: 30936753-30936753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201154662 Chr.1: 30936845-30936845 on Build GRCh38
rs570305938 Chr.1: 30936911-30936911 on Build GRCh38
rs907096613 Chr.1: 30936615-30936615 on Build GRCh38
rs376122983 Chr.1: 30936880-30936880 on Build GRCh38
rs772730238 Chr.1: 30936847-30936847 on Build GRCh38
rs114154027 Chr.1: 30936655-30936655 on Build GRCh38
rs369445106 Chr.1: 30936682-30936682 on Build GRCh38
rs560223801 Chr.1: 30936602-30936602 on Build GRCh38
rs373947877 Chr.1: 30936724-30936724 on Build GRCh38
rs746857009 Chr.1: 30936662-30936662 on Build GRCh38
rs150518018 Chr.1: 30936796-30936796 on Build GRCh38
rs982086752 Chr.1: 30936970-30936970 on Build GRCh38
rs572661565 Chr.1: 30936934-30936934 on Build GRCh38
rs111555425 Chr.1: 30936853-30936853 on Build GRCh38
rs1016998098 Chr.1: 30936888-30936888 on Build GRCh38
rs772139393 Chr.1: 30936654-30936654 on Build GRCh38
rs201905426 Chr.1: 30936626-30936627 on Build GRCh38
rs754499819 Chr.1: 30936922-30936922 on Build GRCh38
rs772786563 Chr.1: 30936703-30936703 on Build GRCh38
rs56387244 Chr.1: 30936880-30936881 on Build GRCh38
rs377637084 Chr.1: 30936778-30936778 on Build GRCh38
rs752935491 Chr.1: 30936635-30936635 on Build GRCh38
rs777288060 Chr.1: 30936879-30936880 on Build GRCh38
rs141661422 Chr.1: 30936808-30936808 on Build GRCh38
rs761500460 Chr.1: 30936690-30936690 on Build GRCh38
rs371056869 Chr.1: 30936789-30936789 on Build GRCh38
rs192476821 Chr.1: 30936782-30936782 on Build GRCh38
rs1051691728 Chr.1: 30936800-30936800 on Build GRCh38
rs760889372 Chr.1: 30936878-30936878 on Build GRCh38
rs959212451 Chr.1: 30936999-30936999 on Build GRCh38
rs140134045 Chr.1: 30936832-30936832 on Build GRCh38
rs759936107 Chr.1: 30936735-30936735 on Build GRCh38
rs749288838 Chr.1: 30936651-30936651 on Build GRCh38
rs537558623 Chr.1: 30936992-30936993 on Build GRCh38
rs374355042 Chr.1: 30936790-30936790 on Build GRCh38
rs774991909 Chr.1: 30936843-30936844 on Build GRCh38
rs1045555842 Chr.1: 30936609-30936609 on Build GRCh38
rs138834169 Chr.1: 30936754-30936754 on Build GRCh38
rs1038151358 Chr.1: 30936577-30936577 on Build GRCh38
rs982313508 Chr.1: 30936624-30936624 on Build GRCh38
rs918400416 Chr.1: 30936595-30936595 on Build GRCh38
rs1055737677 Chr.1: 30936936-30936936 on Build GRCh38
rs937245130 Chr.1: 30936930-30936930 on Build GRCh38
rs774888741 Chr.1: 30936868-30936868 on Build GRCh38
rs781463461 Chr.1: 30936626-30936626 on Build GRCh38
rs760060759 Chr.1: 30936870-30936870 on Build GRCh38
rs768564087 Chr.1: 30936681-30936681 on Build GRCh38
rs762395844 Chr.1: 30936715-30936715 on Build GRCh38
rs764432650 Chr.1: 30936879-30936879 on Build GRCh38
rs752060985 Chr.1: 30936733-30936733 on Build GRCh38
rs769869616 Chr.1: 30936573-30936573 on Build GRCh38
rs371510675 Chr.1: 30936637-30936637 on Build GRCh38
rs555884494 Chr.1: 30936927-30936927 on Build GRCh38
rs770400343 Chr.1: 30936855-30936855 on Build GRCh38
rs754107627 Chr.1: 30936884-30936884 on Build GRCh38
rs764155039 Chr.1: 30936781-30936781 on Build GRCh38
rs770800330 Chr.1: 30936964-30936964 on Build GRCh38
rs1035254121 Chr.1: 30936979-30936979 on Build GRCh38
rs56143291 Chr.1: 30936787-30936787 on Build GRCh38
rs115183923 Chr.1: 30936636-30936636 on Build GRCh38
rs937297455 Chr.1: 30936623-30936623 on Build GRCh38
rs777932146 Chr.1: 30936640-30936640 on Build GRCh38
rs889987766 Chr.1: 30936866-30936866 on Build GRCh38
rs188483102 Chr.1: 30936923-30936923 on Build GRCh38
rs775928132 Chr.1: 30936874-30936874 on Build GRCh38
rs773997263 Chr.1: 30936987-30936987 on Build GRCh38
rs950610583 Chr.1: 30936969-30936969 on Build GRCh38
rs769278762 Chr.1: 30936697-30936697 on Build GRCh38

View Full Product Details