Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 31180848-31181347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKAIN1
Entrez Gene ID
79570
Gene Name
Na+/K+ transporting ATPase interacting 1
Gene Aliases
FAM77C
UniGene
Hs.470259
Gene Chromosome Location
Chr.1: 31179270-31239887 on Build GRCh38
Transcripts
NM_024522.2 XM_017002320.1

SNP Detail

SNP ID Chromosome Location
rs995219797 Chr.1: 31181057-31181057 on Build GRCh38
rs968426141 Chr.1: 31181181-31181181 on Build GRCh38
rs772821058 Chr.1: 31181260-31181260 on Build GRCh38
rs890087025 Chr.1: 31181272-31181272 on Build GRCh38
rs569675127 Chr.1: 31180979-31180979 on Build GRCh38
rs535461320 Chr.1: 31180994-31180994 on Build GRCh38
rs1043986041 Chr.1: 31181236-31181236 on Build GRCh38
rs371512280 Chr.1: 31181263-31181265 on Build GRCh38
rs549348683 Chr.1: 31180893-31180893 on Build GRCh38
rs946878260 Chr.1: 31181235-31181235 on Build GRCh38
rs991594974 Chr.1: 31180936-31180936 on Build GRCh38
rs4949356 Chr.1: 31181141-31181141 on Build GRCh38
rs928408948 Chr.1: 31180932-31180932 on Build GRCh38
rs928067813 Chr.1: 31181180-31181180 on Build GRCh38
rs887061982 Chr.1: 31181325-31181325 on Build GRCh38
rs987689320 Chr.1: 31181216-31181216 on Build GRCh38
rs899235526 Chr.1: 31181257-31181257 on Build GRCh38
rs191213303 Chr.1: 31181175-31181175 on Build GRCh38
rs895282656 Chr.1: 31180901-31180901 on Build GRCh38
rs916755829 Chr.1: 31181220-31181220 on Build GRCh38
rs959831294 Chr.1: 31181151-31181151 on Build GRCh38
rs924897145 Chr.1: 31181187-31181187 on Build GRCh38
rs1007657758 Chr.1: 31181033-31181033 on Build GRCh38
rs973740142 Chr.1: 31181109-31181109 on Build GRCh38
rs934839506 Chr.1: 31180952-31180952 on Build GRCh38
rs1009081842 Chr.1: 31181308-31181308 on Build GRCh38
rs939440425 Chr.1: 31181196-31181196 on Build GRCh38
rs964553543 Chr.1: 31181100-31181100 on Build GRCh38
rs1017857151 Chr.1: 31181089-31181089 on Build GRCh38
rs6425714 Chr.1: 31180886-31180886 on Build GRCh38
rs565169396 Chr.1: 31181261-31181263 on Build GRCh38
rs998509877 Chr.1: 31181258-31181258 on Build GRCh38
rs930681781 Chr.1: 31181286-31181286 on Build GRCh38
rs1039134379 Chr.1: 31181252-31181252 on Build GRCh38
rs1031421383 Chr.1: 31181022-31181023 on Build GRCh38
rs978816209 Chr.1: 31181186-31181186 on Build GRCh38
rs1043332233 Chr.1: 31180940-31180940 on Build GRCh38
rs148130879 Chr.1: 31181242-31181242 on Build GRCh38
rs1039491254 Chr.1: 31181042-31181042 on Build GRCh38
rs557962899 Chr.1: 31181228-31181228 on Build GRCh38
rs915587264 Chr.1: 31180937-31180937 on Build GRCh38
rs182583866 Chr.1: 31181218-31181218 on Build GRCh38
rs781008924 Chr.1: 31181064-31181064 on Build GRCh38
rs143823443 Chr.1: 31180891-31180891 on Build GRCh38
rs991255749 Chr.1: 31181160-31181160 on Build GRCh38
rs934893289 Chr.1: 31181188-31181188 on Build GRCh38
rs527573258 Chr.1: 31181251-31181251 on Build GRCh38
rs866620823 Chr.1: 31181296-31181296 on Build GRCh38
rs764564770 Chr.1: 31180938-31180938 on Build GRCh38
rs1035478088 Chr.1: 31181146-31181146 on Build GRCh38
rs899185570 Chr.1: 31181047-31181047 on Build GRCh38
rs924828392 Chr.1: 31180941-31180941 on Build GRCh38
rs890579297 Chr.1: 31181022-31181022 on Build GRCh38
rs747757920 Chr.1: 31181070-31181070 on Build GRCh38
rs959777812 Chr.1: 31180935-31180935 on Build GRCh38

View Full Product Details