Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 31908747-31909258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTP4A2
Entrez Gene ID
8073
Gene Name
protein tyrosine phosphatase type IVA, member 2
Gene Aliases
HH13, HH7-2, HU-PP-1, OV-1, PRL-2, PRL2, PTP4A, PTPCAAX2, ptp-IV1a, ptp-IV1b
UniGene
Hs.470477
Gene Chromosome Location
Chr.1: 31906421-31942635 on Build GRCh38
Transcripts
NM_001195100.1 NM_001195101.1 NM_080391.3 XM_005271231.3 XM_005271232.3 XM_017002401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1222500 c.445T>C p.Y149H Chr.1: 31908911-31908911 on Build GRCh38
4718889 c.405G>C p.R135S Chr.1: 31908951-31908951 on Build GRCh38
7110744 c.471C>T p.F157F Chr.1: 31908885-31908885 on Build GRCh38
4718888 c.448C>T p.R150* Chr.1: 31908908-31908908 on Build GRCh38
1627014 c.410C>T p.A137V Chr.1: 31908946-31908946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023810438 Chr.1: 31909132-31909132 on Build GRCh38
rs769459304 Chr.1: 31908840-31908840 on Build GRCh38
rs751911060 Chr.1: 31908824-31908824 on Build GRCh38
rs150063538 Chr.1: 31909222-31909222 on Build GRCh38
rs967563512 Chr.1: 31909157-31909157 on Build GRCh38
rs575388303 Chr.1: 31909094-31909094 on Build GRCh38
rs367554803 Chr.1: 31908831-31908831 on Build GRCh38
rs371791995 Chr.1: 31908838-31908838 on Build GRCh38
rs775361473 Chr.1: 31909012-31909012 on Build GRCh38
rs200712857 Chr.1: 31909000-31909000 on Build GRCh38
rs765327705 Chr.1: 31908892-31908892 on Build GRCh38
rs1024202388 Chr.1: 31909017-31909017 on Build GRCh38
rs780447763 Chr.1: 31908821-31908821 on Build GRCh38
rs540600360 Chr.1: 31909208-31909208 on Build GRCh38
rs377229712 Chr.1: 31908976-31908976 on Build GRCh38
rs143857852 Chr.1: 31908879-31908879 on Build GRCh38
rs373023781 Chr.1: 31908994-31908994 on Build GRCh38
rs764148402 Chr.1: 31909147-31909147 on Build GRCh38
rs913358730 Chr.1: 31908808-31908808 on Build GRCh38
rs748946683 Chr.1: 31908850-31908850 on Build GRCh38
rs149882515 Chr.1: 31908874-31908874 on Build GRCh38
rs1004908538 Chr.1: 31909048-31909048 on Build GRCh38
rs570123945 Chr.1: 31909212-31909212 on Build GRCh38
rs9782995 Chr.1: 31909106-31909106 on Build GRCh38
rs74065984 Chr.1: 31909198-31909198 on Build GRCh38
rs755406990 Chr.1: 31908945-31908945 on Build GRCh38
rs370929116 Chr.1: 31908816-31908816 on Build GRCh38
rs376380842 Chr.1: 31908896-31908896 on Build GRCh38
rs770601686 Chr.1: 31908997-31908997 on Build GRCh38
rs777242223 Chr.1: 31908843-31908843 on Build GRCh38
rs201098063 Chr.1: 31908860-31908860 on Build GRCh38
rs775406448 Chr.1: 31908888-31908888 on Build GRCh38
rs750520518 Chr.1: 31908894-31908894 on Build GRCh38
rs942144099 Chr.1: 31909224-31909224 on Build GRCh38
rs778440356 Chr.1: 31908998-31908998 on Build GRCh38
rs781737488 Chr.1: 31908982-31908982 on Build GRCh38
rs907119359 Chr.1: 31909049-31909049 on Build GRCh38
rs372563676 Chr.1: 31908946-31908946 on Build GRCh38
rs969509528 Chr.1: 31909140-31909140 on Build GRCh38
rs765175375 Chr.1: 31908867-31908878 on Build GRCh38
rs369642528 Chr.1: 31908903-31908903 on Build GRCh38
rs550344653 Chr.1: 31909176-31909176 on Build GRCh38
rs990280067 Chr.1: 31909126-31909126 on Build GRCh38
rs750294219 Chr.1: 31908993-31908994 on Build GRCh38
rs374554893 Chr.1: 31908830-31908830 on Build GRCh38
rs796598067 Chr.1: 31908773-31908773 on Build GRCh38
rs534962729 Chr.1: 31908782-31908782 on Build GRCh38
rs116733944 Chr.1: 31909069-31909069 on Build GRCh38
rs190414223 Chr.1: 31909205-31909205 on Build GRCh38
rs768021397 Chr.1: 31909107-31909107 on Build GRCh38
rs777304535 Chr.1: 31908991-31908991 on Build GRCh38
rs371968552 Chr.1: 31908986-31908986 on Build GRCh38
rs267598550 Chr.1: 31908942-31908942 on Build GRCh38
rs957153227 Chr.1: 31909108-31909108 on Build GRCh38
rs761612470 Chr.1: 31908852-31908852 on Build GRCh38
rs139226862 Chr.1: 31908788-31908788 on Build GRCh38
rs771888718 Chr.1: 31909009-31909009 on Build GRCh38
rs1013840597 Chr.1: 31909122-31909122 on Build GRCh38
rs1038218022 Chr.1: 31908977-31908977 on Build GRCh38
rs751942426 Chr.1: 31908920-31908920 on Build GRCh38
rs376335336 Chr.1: 31908988-31908988 on Build GRCh38
rs774189032 Chr.1: 31908862-31908862 on Build GRCh38
rs74065981 Chr.1: 31909194-31909194 on Build GRCh38

View Full Product Details