Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29255719-29256260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUFIP2
Entrez Gene ID
57532
Gene Name
NUFIP2, FMR1 interacting protein 2
Gene Aliases
182-FIP, 82-FIP, FIP-82, PIG1
UniGene
Hs.462598
Gene Chromosome Location
Chr.17: 29255836-29294148 on Build GRCh38
Transcripts
NM_020772.2 XM_017024896.1

SNP Detail

SNP ID Chromosome Location
rs564163803 Chr.17: 29255962-29255962 on Build GRCh38
rs760083147 Chr.17: 29256089-29256089 on Build GRCh38
rs1039484629 Chr.17: 29255959-29255959 on Build GRCh38
rs937549841 Chr.17: 29256026-29256026 on Build GRCh38
rs1038195120 Chr.17: 29255818-29255818 on Build GRCh38
rs1051681327 Chr.17: 29256138-29256138 on Build GRCh38
rs767044605 Chr.17: 29255759-29255764 on Build GRCh38
rs1006768466 Chr.17: 29255908-29255909 on Build GRCh38
rs900745403 Chr.17: 29256022-29256022 on Build GRCh38
rs750236672 Chr.17: 29256175-29256175 on Build GRCh38
rs893079073 Chr.17: 29256180-29256180 on Build GRCh38
rs749159830 Chr.17: 29256233-29256233 on Build GRCh38
rs545819856 Chr.17: 29255933-29255933 on Build GRCh38
rs1006737203 Chr.17: 29256173-29256173 on Build GRCh38
rs999706152 Chr.17: 29256067-29256067 on Build GRCh38
rs1018032320 Chr.17: 29255909-29255909 on Build GRCh38
rs962641711 Chr.17: 29256168-29256168 on Build GRCh38
rs1011603830 Chr.17: 29256031-29256031 on Build GRCh38
rs188510768 Chr.17: 29255847-29255847 on Build GRCh38
rs1011814800 Chr.17: 29256201-29256201 on Build GRCh38
rs942180914 Chr.17: 29256098-29256098 on Build GRCh38
rs777782643 Chr.17: 29256017-29256017 on Build GRCh38
rs1050888999 Chr.17: 29256177-29256177 on Build GRCh38
rs181651593 Chr.17: 29256186-29256186 on Build GRCh38
rs990496148 Chr.17: 29256125-29256125 on Build GRCh38
rs897901652 Chr.17: 29256117-29256117 on Build GRCh38
rs562097724 Chr.17: 29256220-29256220 on Build GRCh38
rs916264610 Chr.17: 29256156-29256156 on Build GRCh38
rs1008860486 Chr.17: 29255944-29255944 on Build GRCh38
rs916459515 Chr.17: 29255760-29255760 on Build GRCh38
rs375829155 Chr.17: 29256223-29256223 on Build GRCh38
rs752440075 Chr.17: 29256012-29256012 on Build GRCh38
rs535789433 Chr.17: 29256175-29256175 on Build GRCh38
rs891003136 Chr.17: 29255876-29255876 on Build GRCh38
rs3809796 Chr.17: 29255917-29255917 on Build GRCh38
rs72819452 Chr.17: 29256235-29256235 on Build GRCh38
rs1034732038 Chr.17: 29256113-29256113 on Build GRCh38
rs1030328991 Chr.17: 29255850-29255851 on Build GRCh38
rs531879265 Chr.17: 29256052-29256052 on Build GRCh38
rs1036567156 Chr.17: 29256107-29256107 on Build GRCh38
rs941311821 Chr.17: 29255816-29255816 on Build GRCh38
rs573782394 Chr.17: 29255747-29255747 on Build GRCh38
rs981067843 Chr.17: 29256000-29256000 on Build GRCh38
rs374036216 Chr.17: 29256039-29256039 on Build GRCh38
rs918518635 Chr.17: 29256174-29256174 on Build GRCh38
rs929831651 Chr.17: 29255896-29255896 on Build GRCh38
rs966843437 Chr.17: 29256061-29256061 on Build GRCh38
rs760638071 Chr.17: 29255953-29255956 on Build GRCh38
rs1036837832 Chr.17: 29255826-29255828 on Build GRCh38
rs942104774 Chr.17: 29255752-29255758 on Build GRCh38
rs960445438 Chr.17: 29256116-29256116 on Build GRCh38
rs928294950 Chr.17: 29256018-29256018 on Build GRCh38
rs990020640 Chr.17: 29255929-29255929 on Build GRCh38
rs897939358 Chr.17: 29255826-29255829 on Build GRCh38
rs753719177 Chr.17: 29256236-29256236 on Build GRCh38
rs146779011 Chr.17: 29255916-29255916 on Build GRCh38
rs767239550 Chr.17: 29255905-29255905 on Build GRCh38
rs889127290 Chr.17: 29256163-29256166 on Build GRCh38
rs974376258 Chr.17: 29256169-29256169 on Build GRCh38
rs933409268 Chr.17: 29256130-29256130 on Build GRCh38

View Full Product Details