Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32024255-32024762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC37B
Entrez Gene ID
114659
Gene Name
leucine rich repeat containing 37B
Gene Aliases
LRRC37
UniGene
Hs.514071
Gene Chromosome Location
Chr.17: 32007872-32053500 on Build GRCh38
Transcripts
NM_001321350.1 NM_052888.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4530356 c.1690G>A p.G564R Chr.17: 32024721-32024721 on Build GRCh38
4939352 c.1690G>T p.G564* Chr.17: 32024721-32024721 on Build GRCh38
4699075 c.1720G>A p.V574I Chr.17: 32024751-32024751 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs184184550 Chr.17: 32024628-32024628 on Build GRCh38
rs754236149 Chr.17: 32024719-32024719 on Build GRCh38
rs879160134 Chr.17: 32024303-32024303 on Build GRCh38
rs560732178 Chr.17: 32024414-32024414 on Build GRCh38
rs958089262 Chr.17: 32024553-32024553 on Build GRCh38
rs577050416 Chr.17: 32024312-32024312 on Build GRCh38
rs553941079 Chr.17: 32024298-32024298 on Build GRCh38
rs556780900 Chr.17: 32024353-32024353 on Build GRCh38
rs544148691 Chr.17: 32024399-32024399 on Build GRCh38
rs766776004 Chr.17: 32024707-32024707 on Build GRCh38
rs375318220 Chr.17: 32024665-32024665 on Build GRCh38
rs112648041 Chr.17: 32024706-32024706 on Build GRCh38
rs552418245 Chr.17: 32024505-32024505 on Build GRCh38
rs775276610 Chr.17: 32024674-32024674 on Build GRCh38
rs560370164 Chr.17: 32024446-32024446 on Build GRCh38
rs548551176 Chr.17: 32024597-32024597 on Build GRCh38
rs978217063 Chr.17: 32024532-32024532 on Build GRCh38
rs199756150 Chr.17: 32024496-32024496 on Build GRCh38
rs954667264 Chr.17: 32024510-32024510 on Build GRCh38
rs527756849 Chr.17: 32024635-32024635 on Build GRCh38
rs574575167 Chr.17: 32024416-32024416 on Build GRCh38
rs755543429 Chr.17: 32024723-32024723 on Build GRCh38
rs1026454002 Chr.17: 32024381-32024381 on Build GRCh38
rs188732402 Chr.17: 32024673-32024673 on Build GRCh38
rs989470651 Chr.17: 32024576-32024576 on Build GRCh38
rs191491355 Chr.17: 32024336-32024336 on Build GRCh38
rs568881 Chr.17: 32024618-32024618 on Build GRCh38
rs531515906 Chr.17: 32024538-32024538 on Build GRCh38
rs549174705 Chr.17: 32024281-32024281 on Build GRCh38
rs773871830 Chr.17: 32024678-32024678 on Build GRCh38
rs565918748 Chr.17: 32024314-32024314 on Build GRCh38
rs773620619 Chr.17: 32024672-32024672 on Build GRCh38
rs780952202 Chr.17: 32024695-32024698 on Build GRCh38
rs1037327258 Chr.17: 32024735-32024735 on Build GRCh38
rs1030124665 Chr.17: 32024508-32024508 on Build GRCh38
rs576685757 Chr.17: 32024386-32024386 on Build GRCh38
rs562753096 Chr.17: 32024506-32024506 on Build GRCh38
rs767122600 Chr.17: 32024697-32024697 on Build GRCh38
rs371923939 Chr.17: 32024699-32024699 on Build GRCh38
rs763472124 Chr.17: 32024675-32024675 on Build GRCh38
rs144144091 Chr.17: 32024729-32024729 on Build GRCh38
rs369447706 Chr.17: 32024286-32024286 on Build GRCh38
rs916873155 Chr.17: 32024603-32024603 on Build GRCh38
rs540377684 Chr.17: 32024445-32024445 on Build GRCh38
rs1046648221 Chr.17: 32024633-32024633 on Build GRCh38
rs761360726 Chr.17: 32024690-32024690 on Build GRCh38
rs879065187 Chr.17: 32024347-32024347 on Build GRCh38
rs1016162994 Chr.17: 32024422-32024422 on Build GRCh38

View Full Product Details