Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18121927-18122429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO15A
Entrez Gene ID
51168
Gene Name
myosin XVA
Gene Aliases
DFNB3, MYO15
UniGene
Hs.462390
Gene Chromosome Location
Chr.17: 18108706-18180227 on Build GRCh38
Transcripts
NM_016239.3 XM_011523918.2 XM_017024714.1 XM_017024715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
182142 c.3316C>T p.R1106W Chr.17: 18122116-18122116 on Build GRCh38
3795322 c.3210G>C p.Q1070H Chr.17: 18122010-18122010 on Build GRCh38
1609915 c.3338G>A p.R1113H Chr.17: 18122138-18122138 on Build GRCh38
6080426 c.3412C>T p.Q1138* Chr.17: 18122212-18122212 on Build GRCh38
4517390 c.3330_3331CC>TT p.P1111S Chr.17: 18122130-18122131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757368406 Chr.17: 18122007-18122007 on Build GRCh38
rs760164088 Chr.17: 18122068-18122068 on Build GRCh38
rs770121419 Chr.17: 18122232-18122232 on Build GRCh38
rs770547097 Chr.17: 18122188-18122188 on Build GRCh38
rs763512706 Chr.17: 18121965-18121965 on Build GRCh38
rs992088282 Chr.17: 18122292-18122292 on Build GRCh38
rs763119472 Chr.17: 18122344-18122344 on Build GRCh38
rs114052634 Chr.17: 18122311-18122311 on Build GRCh38
rs773409705 Chr.17: 18122233-18122233 on Build GRCh38
rs368275759 Chr.17: 18122390-18122390 on Build GRCh38
rs199851548 Chr.17: 18122192-18122192 on Build GRCh38
rs376630473 Chr.17: 18122039-18122039 on Build GRCh38
rs759771939 Chr.17: 18121979-18121979 on Build GRCh38
rs777184129 Chr.17: 18121976-18121984 on Build GRCh38
rs199537186 Chr.17: 18121996-18121996 on Build GRCh38
rs866595552 Chr.17: 18122033-18122033 on Build GRCh38
rs746651600 Chr.17: 18122055-18122055 on Build GRCh38
rs751475570 Chr.17: 18122161-18122161 on Build GRCh38
rs121908971 Chr.17: 18122113-18122113 on Build GRCh38
rs766730482 Chr.17: 18121973-18121973 on Build GRCh38
rs754109028 Chr.17: 18122086-18122086 on Build GRCh38
rs539382039 Chr.17: 18122062-18122062 on Build GRCh38
rs768363883 Chr.17: 18122059-18122059 on Build GRCh38
rs776352434 Chr.17: 18122061-18122061 on Build GRCh38
rs200093968 Chr.17: 18121983-18121983 on Build GRCh38
rs370973719 Chr.17: 18122258-18122258 on Build GRCh38
rs373661688 Chr.17: 18122119-18122119 on Build GRCh38
rs557393326 Chr.17: 18122402-18122402 on Build GRCh38
rs774683207 Chr.17: 18122067-18122067 on Build GRCh38
rs761946533 Chr.17: 18122224-18122224 on Build GRCh38
rs868716354 Chr.17: 18122034-18122034 on Build GRCh38
rs769884586 Chr.17: 18122134-18122134 on Build GRCh38
rs368083374 Chr.17: 18122351-18122351 on Build GRCh38
rs747533694 Chr.17: 18122298-18122298 on Build GRCh38
rs1023023222 Chr.17: 18122300-18122300 on Build GRCh38
rs748603406 Chr.17: 18122306-18122306 on Build GRCh38
rs1026017968 Chr.17: 18122107-18122107 on Build GRCh38
rs200222411 Chr.17: 18122391-18122391 on Build GRCh38
rs945403409 Chr.17: 18121957-18121957 on Build GRCh38
rs148356427 Chr.17: 18122001-18122001 on Build GRCh38
rs776059784 Chr.17: 18122070-18122070 on Build GRCh38
rs748868741 Chr.17: 18122185-18122185 on Build GRCh38
rs752122074 Chr.17: 18121975-18121975 on Build GRCh38
rs776149914 Chr.17: 18122138-18122138 on Build GRCh38
rs780997753 Chr.17: 18122175-18122175 on Build GRCh38
rs375451997 Chr.17: 18122158-18122158 on Build GRCh38
rs766414503 Chr.17: 18122245-18122245 on Build GRCh38
rs942014701 Chr.17: 18122106-18122106 on Build GRCh38
rs774353733 Chr.17: 18122364-18122364 on Build GRCh38
rs764161170 Chr.17: 18122080-18122080 on Build GRCh38
rs780749414 Chr.17: 18122194-18122194 on Build GRCh38
rs199505716 Chr.17: 18122382-18122382 on Build GRCh38
rs778519450 Chr.17: 18122284-18122284 on Build GRCh38
rs767226283 Chr.17: 18122260-18122260 on Build GRCh38
rs769141382 Chr.17: 18122302-18122302 on Build GRCh38
rs773462695 Chr.17: 18122340-18122340 on Build GRCh38
rs756457030 Chr.17: 18122000-18122000 on Build GRCh38
rs376438000 Chr.17: 18122257-18122257 on Build GRCh38
rs794729637 Chr.17: 18122111-18122112 on Build GRCh38
rs779871596 Chr.17: 18122047-18122047 on Build GRCh38
rs535308861 Chr.17: 18122184-18122184 on Build GRCh38
rs973338524 Chr.17: 18122326-18122326 on Build GRCh38
rs752465669 Chr.17: 18122265-18122265 on Build GRCh38
rs776997964 Chr.17: 18122218-18122218 on Build GRCh38
rs757242173 Chr.17: 18122087-18122087 on Build GRCh38
rs762048139 Chr.17: 18122155-18122155 on Build GRCh38
rs755853662 Chr.17: 18122269-18122269 on Build GRCh38
rs747480812 Chr.17: 18122197-18122197 on Build GRCh38
rs771821256 Chr.17: 18122041-18122041 on Build GRCh38
rs919809633 Chr.17: 18122110-18122110 on Build GRCh38
rs367781635 Chr.17: 18122065-18122065 on Build GRCh38
rs377301443 Chr.17: 18122109-18122109 on Build GRCh38
rs189061214 Chr.17: 18122003-18122003 on Build GRCh38
rs371576029 Chr.17: 18122076-18122076 on Build GRCh38
rs764491646 Chr.17: 18122152-18122152 on Build GRCh38
rs781546107 Chr.17: 18122305-18122305 on Build GRCh38
rs865928977 Chr.17: 18122096-18122096 on Build GRCh38
rs776751357 Chr.17: 18122154-18122154 on Build GRCh38
rs879047948 Chr.17: 18122014-18122014 on Build GRCh38
rs201197589 Chr.17: 18122163-18122163 on Build GRCh38
rs554373246 Chr.17: 18122401-18122401 on Build GRCh38
rs991578634 Chr.17: 18122074-18122074 on Build GRCh38
rs763112969 Chr.17: 18122259-18122260 on Build GRCh38
rs761174310 Chr.17: 18122145-18122145 on Build GRCh38
rs766187994 Chr.17: 18122323-18122324 on Build GRCh38
rs767554523 Chr.17: 18122386-18122386 on Build GRCh38
rs754781505 Chr.17: 18122116-18122116 on Build GRCh38
rs746368529 Chr.17: 18122120-18122120 on Build GRCh38
rs200738532 Chr.17: 18122159-18122159 on Build GRCh38
rs765396397 Chr.17: 18122095-18122095 on Build GRCh38
rs763069463 Chr.17: 18122243-18122243 on Build GRCh38
rs919257187 Chr.17: 18122341-18122341 on Build GRCh38
rs768132457 Chr.17: 18121951-18121951 on Build GRCh38
rs76468019 Chr.17: 18122213-18122213 on Build GRCh38
rs199952218 Chr.17: 18122117-18122117 on Build GRCh38
rs747775830 Chr.17: 18122181-18122181 on Build GRCh38
rs533897442 Chr.17: 18122278-18122278 on Build GRCh38
rs750304417 Chr.17: 18122108-18122108 on Build GRCh38
rs772777538 Chr.17: 18122137-18122137 on Build GRCh38
rs899479127 Chr.17: 18121981-18121981 on Build GRCh38
rs375220157 Chr.17: 18122279-18122279 on Build GRCh38
rs773355397 Chr.17: 18122252-18122261 on Build GRCh38
rs776063232 Chr.17: 18121961-18121961 on Build GRCh38

View Full Product Details