Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19378807-19379308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK7
Entrez Gene ID
5598
Gene Name
mitogen-activated protein kinase 7
Gene Aliases
BMK1, ERK4, ERK5, PRKM7
UniGene
Hs.150136
Gene Chromosome Location
Chr.17: 19377721-19383544 on Build GRCh38
Transcripts
NM_002749.3 NM_139032.2 NM_139033.2 NM_139034.2 XM_006721557.3 XM_006721558.3 XM_006721559.3 XM_011523957.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6282046 c.179C>T p.T60I Chr.17: 19379079-19379079 on Build GRCh38
343238 c.136G>A p.D46N Chr.17: 19379036-19379036 on Build GRCh38
4151497 c.177G>T p.E59D Chr.17: 19379077-19379077 on Build GRCh38
4567363 c.66_67GG>AA p.A23T Chr.17: 19378966-19378967 on Build GRCh38
4474862 c.194C>T p.A65V Chr.17: 19379094-19379094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922017856 Chr.17: 19379256-19379256 on Build GRCh38
rs904098316 Chr.17: 19379013-19379013 on Build GRCh38
rs764239408 Chr.17: 19379133-19379133 on Build GRCh38
rs1031202714 Chr.17: 19378849-19378849 on Build GRCh38
rs759494226 Chr.17: 19378999-19378999 on Build GRCh38
rs199846619 Chr.17: 19378897-19378897 on Build GRCh38
rs759673722 Chr.17: 19378928-19378928 on Build GRCh38
rs983879740 Chr.17: 19379198-19379198 on Build GRCh38
rs750972995 Chr.17: 19379065-19379065 on Build GRCh38
rs925550501 Chr.17: 19379282-19379282 on Build GRCh38
rs778393353 Chr.17: 19378969-19378969 on Build GRCh38
rs1052943323 Chr.17: 19379170-19379170 on Build GRCh38
rs1023480108 Chr.17: 19379095-19379095 on Build GRCh38
rs1030166474 Chr.17: 19379262-19379262 on Build GRCh38
rs771836793 Chr.17: 19378999-19379010 on Build GRCh38
rs764364202 Chr.17: 19379041-19379041 on Build GRCh38
rs770790042 Chr.17: 19379114-19379114 on Build GRCh38
rs544657727 Chr.17: 19378927-19378927 on Build GRCh38
rs572805130 Chr.17: 19379223-19379223 on Build GRCh38
rs955360849 Chr.17: 19379264-19379264 on Build GRCh38
rs1027348768 Chr.17: 19379178-19379178 on Build GRCh38
rs767949718 Chr.17: 19379154-19379154 on Build GRCh38
rs944771923 Chr.17: 19378982-19378982 on Build GRCh38
rs982797236 Chr.17: 19379274-19379274 on Build GRCh38
rs932176213 Chr.17: 19379260-19379260 on Build GRCh38
rs935572925 Chr.17: 19379286-19379286 on Build GRCh38
rs754399677 Chr.17: 19379070-19379070 on Build GRCh38
rs767059776 Chr.17: 19379163-19379163 on Build GRCh38
rs185779172 Chr.17: 19379156-19379156 on Build GRCh38
rs983461334 Chr.17: 19378958-19378958 on Build GRCh38
rs752946139 Chr.17: 19378937-19378937 on Build GRCh38
rs776635073 Chr.17: 19378994-19378994 on Build GRCh38
rs773113285 Chr.17: 19378979-19378979 on Build GRCh38
rs549349784 Chr.17: 19379276-19379276 on Build GRCh38
rs747827826 Chr.17: 19378972-19378972 on Build GRCh38
rs778086311 Chr.17: 19378948-19378948 on Build GRCh38
rs757997389 Chr.17: 19379171-19379171 on Build GRCh38
rs145510195 Chr.17: 19378919-19378919 on Build GRCh38
rs371411625 Chr.17: 19379081-19379081 on Build GRCh38
rs751245072 Chr.17: 19379174-19379174 on Build GRCh38
rs1021971502 Chr.17: 19378829-19378829 on Build GRCh38
rs777526691 Chr.17: 19379173-19379173 on Build GRCh38
rs763073942 Chr.17: 19379132-19379132 on Build GRCh38
rs763127575 Chr.17: 19379026-19379026 on Build GRCh38
rs999198607 Chr.17: 19378844-19378844 on Build GRCh38
rs555044170 Chr.17: 19378943-19378943 on Build GRCh38
rs767848284 Chr.17: 19379059-19379059 on Build GRCh38
rs774563799 Chr.17: 19379142-19379142 on Build GRCh38
rs774070569 Chr.17: 19378891-19378891 on Build GRCh38
rs1043887354 Chr.17: 19379268-19379268 on Build GRCh38
rs907854552 Chr.17: 19379208-19379208 on Build GRCh38
rs999628197 Chr.17: 19379036-19379036 on Build GRCh38
rs768177113 Chr.17: 19378883-19378883 on Build GRCh38
rs573586298 Chr.17: 19378855-19378855 on Build GRCh38
rs546912846 Chr.17: 19378992-19378992 on Build GRCh38
rs748692208 Chr.17: 19378961-19378961 on Build GRCh38
rs757695720 Chr.17: 19378959-19378959 on Build GRCh38
rs374947186 Chr.17: 19378968-19378968 on Build GRCh38
rs761636396 Chr.17: 19378895-19378895 on Build GRCh38
rs763642391 Chr.17: 19378925-19378933 on Build GRCh38
rs899914236 Chr.17: 19379257-19379257 on Build GRCh38
rs765309809 Chr.17: 19378934-19378934 on Build GRCh38
rs1011354245 Chr.17: 19379194-19379194 on Build GRCh38
rs571863801 Chr.17: 19379247-19379248 on Build GRCh38
rs903478339 Chr.17: 19378830-19378830 on Build GRCh38
rs992016027 Chr.17: 19378861-19378861 on Build GRCh38
rs762048562 Chr.17: 19379149-19379149 on Build GRCh38
rs781750078 Chr.17: 19378960-19378960 on Build GRCh38
rs951314422 Chr.17: 19379190-19379190 on Build GRCh38
rs367750154 Chr.17: 19379119-19379119 on Build GRCh38
rs370028907 Chr.17: 19378879-19378879 on Build GRCh38
rs752243700 Chr.17: 19379166-19379166 on Build GRCh38
rs778545740 Chr.17: 19379074-19379074 on Build GRCh38
rs973929179 Chr.17: 19379151-19379151 on Build GRCh38
rs528247418 Chr.17: 19379281-19379281 on Build GRCh38
rs747534963 Chr.17: 19378859-19378859 on Build GRCh38
rs756614601 Chr.17: 19379161-19379161 on Build GRCh38
rs751958457 Chr.17: 19378953-19378953 on Build GRCh38
rs567028407 Chr.17: 19378976-19378976 on Build GRCh38
rs769655675 Chr.17: 19379125-19379125 on Build GRCh38
rs776381725 Chr.17: 19379118-19379118 on Build GRCh38
rs976100931 Chr.17: 19379224-19379224 on Build GRCh38
rs746713787 Chr.17: 19379113-19379113 on Build GRCh38
rs745794753 Chr.17: 19379120-19379120 on Build GRCh38
rs777627048 Chr.17: 19379104-19379104 on Build GRCh38
rs771473684 Chr.17: 19378878-19378878 on Build GRCh38
rs546925729 Chr.17: 19379283-19379283 on Build GRCh38
rs932256706 Chr.17: 19379005-19379005 on Build GRCh38
rs777882299 Chr.17: 19378954-19378954 on Build GRCh38
rs757651014 Chr.17: 19379044-19379044 on Build GRCh38
rs747235015 Chr.17: 19379227-19379227 on Build GRCh38
rs531434532 Chr.17: 19378922-19378922 on Build GRCh38
rs751903543 Chr.17: 19379043-19379043 on Build GRCh38
rs1022307946 Chr.17: 19378945-19378945 on Build GRCh38
rs142428761 Chr.17: 19379131-19379131 on Build GRCh38
rs2233073 Chr.17: 19378918-19378918 on Build GRCh38
rs147694046 Chr.17: 19378990-19378990 on Build GRCh38
rs748891527 Chr.17: 19378882-19378882 on Build GRCh38
rs1044409528 Chr.17: 19379009-19379009 on Build GRCh38
rs758119969 Chr.17: 19379089-19379089 on Build GRCh38
rs771272754 Chr.17: 19379261-19379261 on Build GRCh38
rs765379731 Chr.17: 19379012-19379012 on Build GRCh38
rs529303114 Chr.17: 19379228-19379228 on Build GRCh38
rs771406354 Chr.17: 19378900-19378900 on Build GRCh38

View Full Product Details